Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95814403-95814909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRX
Entrez Gene ID
2745
Gene Name
glutaredoxin
Gene Aliases
GRX, GRX1
UniGene
Hs.28988
Gene Chromosome Location
Chr.5: 95813849-95822873 on Build GRCh38
Transcripts
NM_001118890.1 NM_001243658.1 NM_001243659.1 NM_002064.2

SNP Detail

SNP ID Chromosome Location
rs29000870 Chr.5: 95814719-95814719 on Build GRCh38
rs913926738 Chr.5: 95814685-95814685 on Build GRCh38
rs973574436 Chr.5: 95814693-95814693 on Build GRCh38
rs1049085384 Chr.5: 95814559-95814559 on Build GRCh38
rs991491334 Chr.5: 95814668-95814668 on Build GRCh38
rs1047421 Chr.5: 95814428-95814428 on Build GRCh38
rs556582479 Chr.5: 95814843-95814843 on Build GRCh38
rs899760988 Chr.5: 95814605-95814605 on Build GRCh38
rs1047419 Chr.5: 95814765-95814765 on Build GRCh38
rs1018143063 Chr.5: 95814592-95814592 on Build GRCh38
rs890775110 Chr.5: 95814536-95814536 on Build GRCh38
rs899728634 Chr.5: 95814801-95814801 on Build GRCh38
rs1047245793 Chr.5: 95814708-95814708 on Build GRCh38
rs548133210 Chr.5: 95814789-95814789 on Build GRCh38
rs929521239 Chr.5: 95814702-95814702 on Build GRCh38
rs905407249 Chr.5: 95814533-95814533 on Build GRCh38
rs763787799 Chr.5: 95814660-95814660 on Build GRCh38
rs1048570297 Chr.5: 95814774-95814774 on Build GRCh38
rs1014623125 Chr.5: 95814629-95814629 on Build GRCh38
rs775868886 Chr.5: 95814481-95814481 on Build GRCh38
rs888887094 Chr.5: 95814884-95814884 on Build GRCh38
rs997192905 Chr.5: 95814868-95814868 on Build GRCh38
rs29000871 Chr.5: 95814667-95814667 on Build GRCh38
rs967464702 Chr.5: 95814649-95814649 on Build GRCh38
rs912348736 Chr.5: 95814718-95814718 on Build GRCh38
rs750314510 Chr.5: 95814786-95814786 on Build GRCh38
rs1047420 Chr.5: 95814541-95814541 on Build GRCh38
rs1006274132 Chr.5: 95814601-95814601 on Build GRCh38
rs916131478 Chr.5: 95814677-95814677 on Build GRCh38
rs938140168 Chr.5: 95814723-95814723 on Build GRCh38
rs29000872 Chr.5: 95814631-95814631 on Build GRCh38
rs761372564 Chr.5: 95814684-95814684 on Build GRCh38
rs929736098 Chr.5: 95814451-95814451 on Build GRCh38
rs982193603 Chr.5: 95814710-95814710 on Build GRCh38
rs997307403 Chr.5: 95814638-95814638 on Build GRCh38
rs903001033 Chr.5: 95814885-95814885 on Build GRCh38
rs919403866 Chr.5: 95814697-95814697 on Build GRCh38
rs570997888 Chr.5: 95814661-95814661 on Build GRCh38
rs1036437481 Chr.5: 95814796-95814796 on Build GRCh38
rs539943244 Chr.5: 95814663-95814663 on Build GRCh38
rs953158639 Chr.5: 95814656-95814656 on Build GRCh38
rs888704880 Chr.5: 95814580-95814580 on Build GRCh38
rs181120744 Chr.5: 95814664-95814664 on Build GRCh38
rs1011328744 Chr.5: 95814624-95814624 on Build GRCh38
rs188956801 Chr.5: 95814874-95814874 on Build GRCh38

View Full Product Details