Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 96875857-96876363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERAP1
Entrez Gene ID
51752
Gene Name
endoplasmic reticulum aminopeptidase 1
Gene Aliases
A-LAP, ALAP, APPILS, ARTS-1, ARTS1, ERAAP, ERAAP1, PILS-AP, PILSAP
UniGene
Hs.666524
Gene Chromosome Location
Chr.5: 96759245-96935983 on Build GRCh38
Transcripts
XM_011543480.2 XM_011543481.2 XM_011543484.2 XM_011543485.2 XM_011543486.2 XM_017009580.1 XM_017009581.1

Gene Symbol
ERAP2
Entrez Gene ID
64167
Gene Name
endoplasmic reticulum aminopeptidase 2
Gene Aliases
L-RAP, LRAP
UniGene
Hs.482910
Gene Chromosome Location
Chr.5: 96875940-96919703 on Build GRCh38
Transcripts
XM_011543480.2 XM_011543481.2 XM_011543484.2 XM_011543485.2 XM_011543486.2 XM_017009580.1 XM_017009581.1

SNP Detail

SNP ID Chromosome Location
rs543599557 Chr.5: 96876145-96876145 on Build GRCh38
rs552213027 Chr.5: 96875929-96875929 on Build GRCh38
rs1230358 Chr.5: 96876037-96876037 on Build GRCh38
rs116493034 Chr.5: 96876221-96876221 on Build GRCh38
rs1046739289 Chr.5: 96875933-96875933 on Build GRCh38
rs941152037 Chr.5: 96876246-96876246 on Build GRCh38
rs896821045 Chr.5: 96876334-96876334 on Build GRCh38
rs79440765 Chr.5: 96876013-96876013 on Build GRCh38
rs549982292 Chr.5: 96876323-96876323 on Build GRCh38
rs76443575 Chr.5: 96875890-96875890 on Build GRCh38
rs191165100 Chr.5: 96876019-96876019 on Build GRCh38
rs148124557 Chr.5: 96876211-96876211 on Build GRCh38
rs17087114 Chr.5: 96876026-96876026 on Build GRCh38
rs564558427 Chr.5: 96876106-96876106 on Build GRCh38
rs977152926 Chr.5: 96875884-96875884 on Build GRCh38
rs566588762 Chr.5: 96875886-96875886 on Build GRCh38
rs74420041 Chr.5: 96876205-96876205 on Build GRCh38
rs753513123 Chr.5: 96875996-96875996 on Build GRCh38
rs1054893484 Chr.5: 96875991-96875991 on Build GRCh38
rs770012599 Chr.5: 96876261-96876261 on Build GRCh38
rs913267767 Chr.5: 96876113-96876113 on Build GRCh38
rs182637068 Chr.5: 96876223-96876223 on Build GRCh38
rs368544407 Chr.5: 96875887-96875887 on Build GRCh38
rs568595222 Chr.5: 96876047-96876047 on Build GRCh38
rs971804717 Chr.5: 96876185-96876185 on Build GRCh38
rs570519348 Chr.5: 96876230-96876230 on Build GRCh38
rs911963366 Chr.5: 96876078-96876078 on Build GRCh38
rs960259150 Chr.5: 96875939-96875939 on Build GRCh38
rs572372041 Chr.5: 96876030-96876030 on Build GRCh38
rs970228715 Chr.5: 96876302-96876302 on Build GRCh38
rs1006985715 Chr.5: 96876079-96876079 on Build GRCh38
rs747389930 Chr.5: 96876143-96876143 on Build GRCh38
rs576605235 Chr.5: 96876116-96876116 on Build GRCh38
rs529521215 Chr.5: 96876193-96876193 on Build GRCh38
rs143715161 Chr.5: 96876000-96876000 on Build GRCh38
rs755366208 Chr.5: 96876183-96876183 on Build GRCh38
rs531572772 Chr.5: 96876277-96876277 on Build GRCh38
rs185868372 Chr.5: 96876014-96876014 on Build GRCh38
rs927782589 Chr.5: 96875977-96875977 on Build GRCh38
rs562078061 Chr.5: 96876178-96876178 on Build GRCh38
rs1033076811 Chr.5: 96876081-96876081 on Build GRCh38
rs1031421715 Chr.5: 96876206-96876206 on Build GRCh38
rs554232257 Chr.5: 96876027-96876027 on Build GRCh38
rs1005694906 Chr.5: 96875997-96875997 on Build GRCh38
rs532088347 Chr.5: 96876028-96876028 on Build GRCh38
rs937844331 Chr.5: 96876220-96876220 on Build GRCh38
rs560225103 Chr.5: 96876209-96876209 on Build GRCh38
rs957501898 Chr.5: 96876088-96876088 on Build GRCh38
rs570562456 Chr.5: 96875978-96875978 on Build GRCh38

View Full Product Details