Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 96892107-96892608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERAP1
Entrez Gene ID
51752
Gene Name
endoplasmic reticulum aminopeptidase 1
Gene Aliases
A-LAP, ALAP, APPILS, ARTS-1, ARTS1, ERAAP, ERAAP1, PILS-AP, PILSAP
UniGene
Hs.666524
Gene Chromosome Location
Chr.5: 96759245-96935983 on Build GRCh38
Transcripts
XM_011543480.2 XM_011543481.2 XM_011543484.2 XM_011543485.2 XM_011543486.2 XM_017009580.1 XM_017009581.1 XM_011543544.1

Gene Symbol
ERAP2
Entrez Gene ID
64167
Gene Name
endoplasmic reticulum aminopeptidase 2
Gene Aliases
L-RAP, LRAP
UniGene
Hs.482910
Gene Chromosome Location
Chr.5: 96875940-96919703 on Build GRCh38
Transcripts
XM_011543480.2 XM_011543481.2 XM_011543484.2 XM_011543485.2 XM_011543486.2 XM_017009580.1 XM_017009581.1 XM_011543544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6211945 c.992T>C p.F331S Chr.5: 96892320-96892320 on Build GRCh38
6655655 c.1087T>C p.W363R Chr.5: 96892415-96892415 on Build GRCh38
122887 c.1073C>T p.A358V Chr.5: 96892401-96892401 on Build GRCh38
3618903 c.1019G>A p.G340D Chr.5: 96892347-96892347 on Build GRCh38
4457744 c.1058C>T p.P353L Chr.5: 96892386-96892386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769865750 Chr.5: 96892341-96892341 on Build GRCh38
rs777841760 Chr.5: 96892140-96892140 on Build GRCh38
rs747947287 Chr.5: 96892318-96892318 on Build GRCh38
rs1001183714 Chr.5: 96892208-96892208 on Build GRCh38
rs555009989 Chr.5: 96892485-96892485 on Build GRCh38
rs935849463 Chr.5: 96892129-96892129 on Build GRCh38
rs60603456 Chr.5: 96892271-96892271 on Build GRCh38
rs372076669 Chr.5: 96892279-96892279 on Build GRCh38
rs58608188 Chr.5: 96892536-96892536 on Build GRCh38
rs954400211 Chr.5: 96892296-96892296 on Build GRCh38
rs1026232772 Chr.5: 96892576-96892576 on Build GRCh38
rs754666015 Chr.5: 96892303-96892303 on Build GRCh38
rs774647815 Chr.5: 96892447-96892447 on Build GRCh38
rs773702450 Chr.5: 96892470-96892470 on Build GRCh38
rs777784116 Chr.5: 96892335-96892335 on Build GRCh38
rs770840038 Chr.5: 96892427-96892427 on Build GRCh38
rs768101203 Chr.5: 96892280-96892280 on Build GRCh38
rs1055135781 Chr.5: 96892185-96892185 on Build GRCh38
rs11744836 Chr.5: 96892507-96892507 on Build GRCh38
rs779640411 Chr.5: 96892287-96892287 on Build GRCh38
rs546243037 Chr.5: 96892325-96892325 on Build GRCh38
rs146790983 Chr.5: 96892442-96892442 on Build GRCh38
rs1004071858 Chr.5: 96892170-96892170 on Build GRCh38
rs766965697 Chr.5: 96892477-96892477 on Build GRCh38
rs758281528 Chr.5: 96892512-96892512 on Build GRCh38
rs370129609 Chr.5: 96892393-96892393 on Build GRCh38
rs910310199 Chr.5: 96892328-96892328 on Build GRCh38
rs754361180 Chr.5: 96892480-96892480 on Build GRCh38
rs139665203 Chr.5: 96892369-96892369 on Build GRCh38
rs895197091 Chr.5: 96892211-96892211 on Build GRCh38
rs140735506 Chr.5: 96892462-96892462 on Build GRCh38
rs767716707 Chr.5: 96892366-96892366 on Build GRCh38
rs375951913 Chr.5: 96892392-96892392 on Build GRCh38
rs185661936 Chr.5: 96892515-96892515 on Build GRCh38
rs763216518 Chr.5: 96892344-96892344 on Build GRCh38
rs997662533 Chr.5: 96892540-96892540 on Build GRCh38
rs765846266 Chr.5: 96892291-96892291 on Build GRCh38
rs775698713 Chr.5: 96892258-96892258 on Build GRCh38
rs369938769 Chr.5: 96892513-96892513 on Build GRCh38
rs1023953164 Chr.5: 96892154-96892154 on Build GRCh38
rs374222647 Chr.5: 96892471-96892471 on Build GRCh38
rs768900310 Chr.5: 96892345-96892345 on Build GRCh38
rs905507561 Chr.5: 96892197-96892197 on Build GRCh38
rs976154575 Chr.5: 96892326-96892326 on Build GRCh38
rs752396045 Chr.5: 96892487-96892487 on Build GRCh38
rs763094519 Chr.5: 96892261-96892261 on Build GRCh38
rs747014990 Chr.5: 96892413-96892413 on Build GRCh38
rs75263594 Chr.5: 96892368-96892368 on Build GRCh38
rs764590222 Chr.5: 96892486-96892486 on Build GRCh38
rs762260525 Chr.5: 96892359-96892359 on Build GRCh38
rs563179464 Chr.5: 96892242-96892242 on Build GRCh38
rs544616996 Chr.5: 96892150-96892150 on Build GRCh38
rs879127834 Chr.5: 96892415-96892415 on Build GRCh38
rs746888838 Chr.5: 96892339-96892339 on Build GRCh38
rs200808443 Chr.5: 96892406-96892406 on Build GRCh38
rs142595885 Chr.5: 96892310-96892310 on Build GRCh38
rs750912735 Chr.5: 96892285-96892285 on Build GRCh38
rs753379375 Chr.5: 96892386-96892386 on Build GRCh38
rs112382612 Chr.5: 96892525-96892525 on Build GRCh38
rs532733715 Chr.5: 96892146-96892146 on Build GRCh38
rs367873633 Chr.5: 96892342-96892342 on Build GRCh38
rs557002492 Chr.5: 96892382-96892382 on Build GRCh38
rs565485273 Chr.5: 96892139-96892139 on Build GRCh38
rs371076065 Chr.5: 96892254-96892254 on Build GRCh38
rs377162910 Chr.5: 96892376-96892376 on Build GRCh38
rs953456211 Chr.5: 96892573-96892573 on Build GRCh38
rs768571471 Chr.5: 96892446-96892446 on Build GRCh38
rs113463435 Chr.5: 96892405-96892405 on Build GRCh38
rs1016022484 Chr.5: 96892260-96892260 on Build GRCh38
rs370896657 Chr.5: 96892466-96892466 on Build GRCh38
rs895248945 Chr.5: 96892479-96892479 on Build GRCh38
rs187647551 Chr.5: 96892265-96892265 on Build GRCh38
rs774617844 Chr.5: 96892346-96892346 on Build GRCh38
rs780156918 Chr.5: 96892441-96892441 on Build GRCh38
rs891585982 Chr.5: 96892144-96892144 on Build GRCh38
rs1029504989 Chr.5: 96892569-96892569 on Build GRCh38
rs192506462 Chr.5: 96892288-96892288 on Build GRCh38
rs372787526 Chr.5: 96892352-96892352 on Build GRCh38

View Full Product Details