Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 107380606-107381121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNA5
Entrez Gene ID
1946
Gene Name
ephrin A5
Gene Aliases
AF1, EFL5, EPLG7, GLC1M, LERK7, RAGS
UniGene
Hs.288741
Gene Chromosome Location
Chr.5: 107376889-107672364 on Build GRCh38
Transcripts
NM_001962.2 XM_006714565.2 XM_011543250.2 XM_011543251.2 XM_017009205.1

SNP Detail

SNP ID Chromosome Location
rs982998335 Chr.5: 107380835-107380835 on Build GRCh38
rs867591834 Chr.5: 107380915-107380915 on Build GRCh38
rs1044750628 Chr.5: 107380650-107380650 on Build GRCh38
rs1018283218 Chr.5: 107380731-107380731 on Build GRCh38
rs1024778060 Chr.5: 107380909-107380909 on Build GRCh38
rs994081496 Chr.5: 107380947-107380947 on Build GRCh38
rs368717510 Chr.5: 107380660-107380660 on Build GRCh38
rs1033804632 Chr.5: 107380706-107380706 on Build GRCh38
rs878960993 Chr.5: 107381051-107381051 on Build GRCh38
rs118003949 Chr.5: 107380751-107380751 on Build GRCh38
rs537882004 Chr.5: 107380725-107380725 on Build GRCh38
rs193067158 Chr.5: 107380918-107380918 on Build GRCh38
rs561312287 Chr.5: 107381052-107381052 on Build GRCh38
rs904851818 Chr.5: 107380650-107380650 on Build GRCh38
rs564995287 Chr.5: 107380906-107380906 on Build GRCh38
rs879513485 Chr.5: 107380696-107380696 on Build GRCh38
rs903845698 Chr.5: 107380845-107380845 on Build GRCh38
rs537753416 Chr.5: 107380855-107380855 on Build GRCh38
rs1013689722 Chr.5: 107380908-107380908 on Build GRCh38
rs1020452355 Chr.5: 107380718-107380718 on Build GRCh38
rs1009789267 Chr.5: 107380673-107380673 on Build GRCh38
rs995939344 Chr.5: 107381053-107381053 on Build GRCh38
rs576602820 Chr.5: 107380820-107380820 on Build GRCh38
rs961253140 Chr.5: 107380727-107380727 on Build GRCh38
rs778160277 Chr.5: 107380952-107380952 on Build GRCh38
rs540508009 Chr.5: 107380755-107380755 on Build GRCh38
rs187676091 Chr.5: 107380648-107380648 on Build GRCh38
rs892240216 Chr.5: 107380905-107380905 on Build GRCh38
rs553701773 Chr.5: 107380805-107380805 on Build GRCh38
rs1024774305 Chr.5: 107381094-107381094 on Build GRCh38
rs571339710 Chr.5: 107380750-107380750 on Build GRCh38
rs895207587 Chr.5: 107380896-107380896 on Build GRCh38
rs545105393 Chr.5: 107380837-107380837 on Build GRCh38
rs1053554562 Chr.5: 107380883-107380883 on Build GRCh38
rs982824040 Chr.5: 107381086-107381086 on Build GRCh38
rs878976099 Chr.5: 107381056-107381056 on Build GRCh38
rs952466157 Chr.5: 107381068-107381068 on Build GRCh38
rs758732195 Chr.5: 107380914-107380914 on Build GRCh38
rs544325018 Chr.5: 107381023-107381023 on Build GRCh38
rs910895646 Chr.5: 107381078-107381078 on Build GRCh38
rs555186045 Chr.5: 107380723-107380723 on Build GRCh38
rs943805930 Chr.5: 107380779-107380779 on Build GRCh38
rs945593581 Chr.5: 107380823-107380823 on Build GRCh38
rs903856156 Chr.5: 107380701-107380701 on Build GRCh38
rs910999607 Chr.5: 107380763-107380763 on Build GRCh38
rs899841552 Chr.5: 107380972-107380972 on Build GRCh38
rs936311819 Chr.5: 107380866-107380866 on Build GRCh38
rs1008486923 Chr.5: 107380945-107380945 on Build GRCh38
rs1040492827 Chr.5: 107380782-107380782 on Build GRCh38
rs1055599206 Chr.5: 107380869-107380869 on Build GRCh38
rs879084231 Chr.5: 107381046-107381046 on Build GRCh38
rs556003756 Chr.5: 107381072-107381072 on Build GRCh38
rs951506930 Chr.5: 107381082-107381082 on Build GRCh38
rs1001356743 Chr.5: 107380703-107380703 on Build GRCh38
rs891264763 Chr.5: 107380929-107380929 on Build GRCh38
rs972650406 Chr.5: 107380739-107380739 on Build GRCh38
rs1040586989 Chr.5: 107380643-107380643 on Build GRCh38
rs757471463 Chr.5: 107380737-107380737 on Build GRCh38
rs912808332 Chr.5: 107381035-107381035 on Build GRCh38
rs959454558 Chr.5: 107380721-107380721 on Build GRCh38
rs951452645 Chr.5: 107380754-107380754 on Build GRCh38

View Full Product Details