Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 107427135-107427650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNA5
Entrez Gene ID
1946
Gene Name
ephrin A5
Gene Aliases
AF1, EFL5, EPLG7, GLC1M, LERK7, RAGS
UniGene
Hs.288741
Gene Chromosome Location
Chr.5: 107376889-107672364 on Build GRCh38
Transcripts
NM_001962.2 XM_006714565.2 XM_011543250.2 XM_011543251.2 XM_017009205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5931629 c.278C>T p.T93I Chr.5: 107427357-107427357 on Build GRCh38
3607463 c.367C>T p.P123S Chr.5: 107427268-107427268 on Build GRCh38
5975293 c.220A>G p.T74A Chr.5: 107427415-107427415 on Build GRCh38
3607464 c.310C>T p.R104W Chr.5: 107427325-107427325 on Build GRCh38
3661310 c.342C>A p.F114L Chr.5: 107427293-107427293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369274052 Chr.5: 107427191-107427191 on Build GRCh38
rs142282920 Chr.5: 107427235-107427235 on Build GRCh38
rs753881471 Chr.5: 107427442-107427442 on Build GRCh38
rs190308641 Chr.5: 107427188-107427188 on Build GRCh38
rs893172628 Chr.5: 107427161-107427161 on Build GRCh38
rs373469834 Chr.5: 107427318-107427318 on Build GRCh38
rs371145939 Chr.5: 107427351-107427351 on Build GRCh38
rs1029129100 Chr.5: 107427596-107427596 on Build GRCh38
rs150481235 Chr.5: 107427592-107427592 on Build GRCh38
rs756911153 Chr.5: 107427522-107427522 on Build GRCh38
rs775314064 Chr.5: 107427234-107427234 on Build GRCh38
rs923924806 Chr.5: 107427543-107427543 on Build GRCh38
rs200843002 Chr.5: 107427534-107427534 on Build GRCh38
rs532799683 Chr.5: 107427169-107427169 on Build GRCh38
rs534814016 Chr.5: 107427498-107427498 on Build GRCh38
rs199676207 Chr.5: 107427309-107427309 on Build GRCh38
rs778068750 Chr.5: 107427489-107427489 on Build GRCh38
rs535711947 Chr.5: 107427332-107427332 on Build GRCh38
rs529519528 Chr.5: 107427374-107427374 on Build GRCh38
rs959521055 Chr.5: 107427380-107427380 on Build GRCh38
rs1012474541 Chr.5: 107427404-107427404 on Build GRCh38
rs533453014 Chr.5: 107427589-107427589 on Build GRCh38
rs780705232 Chr.5: 107427554-107427554 on Build GRCh38
rs760794041 Chr.5: 107427268-107427268 on Build GRCh38
rs374263179 Chr.5: 107427560-107427560 on Build GRCh38
rs779367666 Chr.5: 107427317-107427317 on Build GRCh38
rs758824988 Chr.5: 107427471-107427471 on Build GRCh38
rs762156298 Chr.5: 107427194-107427194 on Build GRCh38
rs750412562 Chr.5: 107427260-107427260 on Build GRCh38
rs200830072 Chr.5: 107427522-107427523 on Build GRCh38
rs772560595 Chr.5: 107427170-107427170 on Build GRCh38
rs868787124 Chr.5: 107427281-107427281 on Build GRCh38
rs1035548678 Chr.5: 107427324-107427324 on Build GRCh38
rs377722066 Chr.5: 107427325-107427325 on Build GRCh38
rs770718022 Chr.5: 107427177-107427177 on Build GRCh38
rs374490426 Chr.5: 107427353-107427353 on Build GRCh38
rs547396648 Chr.5: 107427196-107427196 on Build GRCh38
rs760844173 Chr.5: 107427431-107427431 on Build GRCh38
rs550879331 Chr.5: 107427409-107427409 on Build GRCh38
rs780608946 Chr.5: 107427364-107427364 on Build GRCh38
rs746537018 Chr.5: 107427172-107427172 on Build GRCh38
rs767729698 Chr.5: 107427254-107427254 on Build GRCh38
rs761975926 Chr.5: 107427245-107427245 on Build GRCh38
rs759521759 Chr.5: 107427523-107427523 on Build GRCh38
rs765848016 Chr.5: 107427457-107427457 on Build GRCh38
rs775199182 Chr.5: 107427415-107427415 on Build GRCh38
rs1035996519 Chr.5: 107427257-107427257 on Build GRCh38
rs569574276 Chr.5: 107427546-107427546 on Build GRCh38
rs769661006 Chr.5: 107427555-107427555 on Build GRCh38
rs1048989524 Chr.5: 107427167-107427167 on Build GRCh38
rs766457284 Chr.5: 107427432-107427432 on Build GRCh38
rs963061379 Chr.5: 107427591-107427591 on Build GRCh38
rs368744696 Chr.5: 107427302-107427302 on Build GRCh38
rs768391866 Chr.5: 107427420-107427420 on Build GRCh38
rs201968475 Chr.5: 107427445-107427445 on Build GRCh38
rs773478586 Chr.5: 107427430-107427430 on Build GRCh38
rs140584205 Chr.5: 107427365-107427365 on Build GRCh38
rs766433848 Chr.5: 107427299-107427299 on Build GRCh38
rs78302459 Chr.5: 107427265-107427265 on Build GRCh38
rs753057345 Chr.5: 107427467-107427467 on Build GRCh38
rs764988266 Chr.5: 107427230-107427230 on Build GRCh38
rs469062 Chr.5: 107427470-107427470 on Build GRCh38
rs762750294 Chr.5: 107427418-107427418 on Build GRCh38

View Full Product Details