Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115271207-115271746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC112
Entrez Gene ID
153733
Gene Name
coiled-coil domain containing 112
Gene Aliases
MBC1
UniGene
Hs.436121
Gene Chromosome Location
Chr.5: 115267188-115296761 on Build GRCh38
Transcripts
NM_001040440.2 NM_152549.2 XM_005271908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5060574 c.1328A>G p.E443G Chr.5: 115271217-115271217 on Build GRCh38
7272239 c.1172G>A p.R391H Chr.5: 115271373-115271373 on Build GRCh38
5026368 c.932G>A p.W311* Chr.5: 115271613-115271613 on Build GRCh38
6903321 c.1196delT p.L399fs*3 Chr.5: 115271349-115271349 on Build GRCh38
324926 c.948G>T p.Q316H Chr.5: 115271597-115271597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377238982 Chr.5: 115271535-115271535 on Build GRCh38
rs113522230 Chr.5: 115271705-115271705 on Build GRCh38
rs776567418 Chr.5: 115271659-115271659 on Build GRCh38
rs768792285 Chr.5: 115271373-115271373 on Build GRCh38
rs767775346 Chr.5: 115271666-115271667 on Build GRCh38
rs148589970 Chr.5: 115271288-115271288 on Build GRCh38
rs751026782 Chr.5: 115271428-115271428 on Build GRCh38
rs762902061 Chr.5: 115271411-115271413 on Build GRCh38
rs1025511675 Chr.5: 115271423-115271423 on Build GRCh38
rs376028786 Chr.5: 115271633-115271633 on Build GRCh38
rs759927817 Chr.5: 115271539-115271539 on Build GRCh38
rs746433964 Chr.5: 115271663-115271663 on Build GRCh38
rs775478272 Chr.5: 115271408-115271412 on Build GRCh38
rs775982113 Chr.5: 115271548-115271548 on Build GRCh38
rs752435553 Chr.5: 115271413-115271413 on Build GRCh38
rs747066895 Chr.5: 115271519-115271519 on Build GRCh38
rs201580583 Chr.5: 115271245-115271245 on Build GRCh38
rs992808168 Chr.5: 115271446-115271446 on Build GRCh38
rs756043590 Chr.5: 115271587-115271587 on Build GRCh38
rs762602877 Chr.5: 115271649-115271649 on Build GRCh38
rs77125878 Chr.5: 115271520-115271520 on Build GRCh38
rs752274160 Chr.5: 115271331-115271331 on Build GRCh38
rs770634478 Chr.5: 115271256-115271264 on Build GRCh38
rs181140069 Chr.5: 115271304-115271304 on Build GRCh38
rs748438204 Chr.5: 115271466-115271466 on Build GRCh38
rs75062506 Chr.5: 115271707-115271707 on Build GRCh38
rs766919358 Chr.5: 115271576-115271576 on Build GRCh38
rs777999344 Chr.5: 115271632-115271632 on Build GRCh38
rs1037784567 Chr.5: 115271681-115271681 on Build GRCh38
rs150286179 Chr.5: 115271416-115271416 on Build GRCh38
rs758646662 Chr.5: 115271622-115271622 on Build GRCh38
rs34508935 Chr.5: 115271434-115271434 on Build GRCh38
rs201799101 Chr.5: 115271666-115271666 on Build GRCh38
rs151110926 Chr.5: 115271374-115271374 on Build GRCh38
rs768034782 Chr.5: 115271262-115271262 on Build GRCh38
rs77012548 Chr.5: 115271595-115271595 on Build GRCh38
rs141205772 Chr.5: 115271549-115271549 on Build GRCh38
rs764120657 Chr.5: 115271595-115271597 on Build GRCh38
rs202145172 Chr.5: 115271342-115271342 on Build GRCh38
rs185790295 Chr.5: 115271367-115271367 on Build GRCh38
rs768869439 Chr.5: 115271243-115271243 on Build GRCh38
rs145983307 Chr.5: 115271560-115271560 on Build GRCh38
rs765762178 Chr.5: 115271545-115271545 on Build GRCh38
rs1038909053 Chr.5: 115271667-115271667 on Build GRCh38
rs779795573 Chr.5: 115271463-115271463 on Build GRCh38
rs771082187 Chr.5: 115271655-115271655 on Build GRCh38
rs759891822 Chr.5: 115271401-115271401 on Build GRCh38
rs137897718 Chr.5: 115271697-115271697 on Build GRCh38
rs747008235 Chr.5: 115271351-115271351 on Build GRCh38
rs750979744 Chr.5: 115271278-115271278 on Build GRCh38
rs759373328 Chr.5: 115271354-115271355 on Build GRCh38
rs17856922 Chr.5: 115271273-115271273 on Build GRCh38
rs750418329 Chr.5: 115271583-115271583 on Build GRCh38
rs568043712 Chr.5: 115271437-115271437 on Build GRCh38
rs769626465 Chr.5: 115271439-115271439 on Build GRCh38
rs777486031 Chr.5: 115271427-115271427 on Build GRCh38
rs373774298 Chr.5: 115271333-115271333 on Build GRCh38
rs745613153 Chr.5: 115271368-115271368 on Build GRCh38
rs200581001 Chr.5: 115271242-115271242 on Build GRCh38
rs142993660 Chr.5: 115271322-115271322 on Build GRCh38
rs751643755 Chr.5: 115271634-115271635 on Build GRCh38
rs138859531 Chr.5: 115271594-115271594 on Build GRCh38
rs997474995 Chr.5: 115271706-115271706 on Build GRCh38
rs765670425 Chr.5: 115271405-115271405 on Build GRCh38
rs761841426 Chr.5: 115271249-115271249 on Build GRCh38
rs776207249 Chr.5: 115271306-115271308 on Build GRCh38
rs756746664 Chr.5: 115271565-115271565 on Build GRCh38
rs772180993 Chr.5: 115271375-115271375 on Build GRCh38
rs17852930 Chr.5: 115271235-115271235 on Build GRCh38
rs766841483 Chr.5: 115271385-115271385 on Build GRCh38
rs773264311 Chr.5: 115271380-115271380 on Build GRCh38
rs138427488 Chr.5: 115271395-115271395 on Build GRCh38
rs766710099 Chr.5: 115271297-115271297 on Build GRCh38
rs772221880 Chr.5: 115271477-115271477 on Build GRCh38
rs770890256 Chr.5: 115271366-115271366 on Build GRCh38
rs200498332 Chr.5: 115271234-115271234 on Build GRCh38
rs1032271890 Chr.5: 115271497-115271497 on Build GRCh38
rs769520015 Chr.5: 115271400-115271402 on Build GRCh38
rs367638591 Chr.5: 115271666-115271666 on Build GRCh38
rs773282749 Chr.5: 115271503-115271503 on Build GRCh38
rs776002768 Chr.5: 115271672-115271672 on Build GRCh38

View Full Product Details