Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 123349978-123350522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP120
Entrez Gene ID
153241
Gene Name
centrosomal protein 120
Gene Aliases
CCDC100, SRTD13
UniGene
Hs.483209
Gene Chromosome Location
Chr.5: 123344885-123423592 on Build GRCh38
Transcripts
NM_001166226.1 NM_153223.3 XM_005271901.4 XM_011543185.2 XM_011543186.2 XM_017009085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1633895 c.2581-1G>T p.? Chr.5: 123350090-123350090 on Build GRCh38
5764055 c.2647C>T p.R883C Chr.5: 123350023-123350023 on Build GRCh38
6353318 c.2637G>C p.Q879H Chr.5: 123350033-123350033 on Build GRCh38
481798 c.2655T>A p.L885L Chr.5: 123350015-123350015 on Build GRCh38
448506 c.2689C>T p.R897* Chr.5: 123349981-123349981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs398093707 Chr.5: 123350223-123350223 on Build GRCh38
rs375395610 Chr.5: 123350303-123350303 on Build GRCh38
rs963767744 Chr.5: 123350051-123350051 on Build GRCh38
rs894791404 Chr.5: 123350406-123350406 on Build GRCh38
rs368260761 Chr.5: 123350022-123350022 on Build GRCh38
rs1047437 Chr.5: 123350033-123350033 on Build GRCh38
rs945583405 Chr.5: 123350105-123350105 on Build GRCh38
rs138806663 Chr.5: 123350290-123350290 on Build GRCh38
rs989539025 Chr.5: 123350330-123350330 on Build GRCh38
rs1004730076 Chr.5: 123350260-123350260 on Build GRCh38
rs527848587 Chr.5: 123350256-123350256 on Build GRCh38
rs928199677 Chr.5: 123350385-123350385 on Build GRCh38
rs767450821 Chr.5: 123350053-123350053 on Build GRCh38
rs748088885 Chr.5: 123350009-123350009 on Build GRCh38
rs939650018 Chr.5: 123350392-123350392 on Build GRCh38
rs769997607 Chr.5: 123350188-123350188 on Build GRCh38
rs371666277 Chr.5: 123350065-123350065 on Build GRCh38
rs954089232 Chr.5: 123350324-123350324 on Build GRCh38
rs1018406690 Chr.5: 123350482-123350482 on Build GRCh38
rs796863868 Chr.5: 123350179-123350179 on Build GRCh38
rs966029062 Chr.5: 123350279-123350279 on Build GRCh38
rs935029333 Chr.5: 123350201-123350201 on Build GRCh38
rs370748677 Chr.5: 123350011-123350011 on Build GRCh38
rs894886394 Chr.5: 123350204-123350204 on Build GRCh38
rs915172132 Chr.5: 123350223-123350225 on Build GRCh38
rs952982583 Chr.5: 123350017-123350017 on Build GRCh38
rs754830593 Chr.5: 123350005-123350005 on Build GRCh38
rs895884152 Chr.5: 123350347-123350347 on Build GRCh38
rs537839972 Chr.5: 123350475-123350475 on Build GRCh38
rs945501406 Chr.5: 123350360-123350360 on Build GRCh38
rs200061679 Chr.5: 123350037-123350037 on Build GRCh38
rs758383095 Chr.5: 123350130-123350130 on Build GRCh38
rs776087538 Chr.5: 123350280-123350280 on Build GRCh38
rs763397454 Chr.5: 123350032-123350032 on Build GRCh38
rs1027379869 Chr.5: 123350232-123350232 on Build GRCh38
rs551759720 Chr.5: 123350314-123350314 on Build GRCh38
rs1044936059 Chr.5: 123350248-123350248 on Build GRCh38
rs754114492 Chr.5: 123350069-123350069 on Build GRCh38
rs751102986 Chr.5: 123350140-123350140 on Build GRCh38
rs777613540 Chr.5: 123350131-123350131 on Build GRCh38
rs778683092 Chr.5: 123350086-123350086 on Build GRCh38
rs79413938 Chr.5: 123350405-123350405 on Build GRCh38
rs886245328 Chr.5: 123350247-123350247 on Build GRCh38
rs754709593 Chr.5: 123350071-123350071 on Build GRCh38
rs930327365 Chr.5: 123350462-123350462 on Build GRCh38
rs1019239651 Chr.5: 123350269-123350269 on Build GRCh38
rs915341915 Chr.5: 123350352-123350352 on Build GRCh38
rs1054753539 Chr.5: 123350203-123350203 on Build GRCh38
rs35423173 Chr.5: 123350213-123350213 on Build GRCh38
rs485847 Chr.5: 123350107-123350107 on Build GRCh38
rs78722208 Chr.5: 123350222-123350223 on Build GRCh38
rs750490786 Chr.5: 123350063-123350063 on Build GRCh38
rs777373534 Chr.5: 123350012-123350012 on Build GRCh38
rs937414248 Chr.5: 123350287-123350287 on Build GRCh38
rs886193973 Chr.5: 123350501-123350501 on Build GRCh38
rs978286280 Chr.5: 123350380-123350380 on Build GRCh38
rs1043004775 Chr.5: 123350109-123350109 on Build GRCh38
rs746550279 Chr.5: 123350020-123350020 on Build GRCh38
rs140306974 Chr.5: 123350023-123350023 on Build GRCh38
rs994569850 Chr.5: 123350226-123350226 on Build GRCh38
rs771132137 Chr.5: 123350064-123350064 on Build GRCh38
rs762445605 Chr.5: 123350044-123350044 on Build GRCh38
rs994606272 Chr.5: 123350374-123350378 on Build GRCh38
rs566247026 Chr.5: 123350185-123350185 on Build GRCh38

View Full Product Details