Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 123587997-123588500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1G3
Entrez Gene ID
1456
Gene Name
casein kinase 1 gamma 3
Gene Aliases
CKI-gamma 3, CSNK1G3L
UniGene
Hs.129206
Gene Chromosome Location
Chr.5: 123512099-123617045 on Build GRCh38
Transcripts
NM_001031812.3 NM_001044723.2 NM_001270572.1 NM_001270573.1 NM_001270574.1 NM_004384.4 XM_005271891.1 XM_005271892.2 XM_005271894.2 XM_005271895.2 XM_005271896.1 XM_017009057.1 XM_017009058.1 XM_017009059.1 XM_017009060.1 XM_017009061.1 XM_017009062.1 XM_017009063.1 XM_017009064.1 XM_017009065.1 XM_017009066.1 XM_017009067.1 XM_017009068.1 XM_017009069.1 XM_017009070.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1060146 c.709C>A p.H237N Chr.5: 123588103-123588103 on Build GRCh38
7193038 c.771delA p.K258fs*16 Chr.5: 123588438-123588438 on Build GRCh38
236079 c.786G>T p.Q262H Chr.5: 123588453-123588453 on Build GRCh38
3608038 c.815C>A p.P272Q Chr.5: 123588482-123588482 on Build GRCh38
3608037 c.742C>T p.P248S Chr.5: 123588136-123588136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753575767 Chr.5: 123588037-123588037 on Build GRCh38
rs757775590 Chr.5: 123588057-123588057 on Build GRCh38
rs112303138 Chr.5: 123588084-123588084 on Build GRCh38
rs768015242 Chr.5: 123588381-123588381 on Build GRCh38
rs192791060 Chr.5: 123588236-123588236 on Build GRCh38
rs1044951613 Chr.5: 123588310-123588310 on Build GRCh38
rs778266325 Chr.5: 123588381-123588381 on Build GRCh38
rs923841023 Chr.5: 123588473-123588473 on Build GRCh38
rs954176248 Chr.5: 123588169-123588172 on Build GRCh38
rs936335106 Chr.5: 123588272-123588272 on Build GRCh38
rs867767855 Chr.5: 123588428-123588428 on Build GRCh38
rs992956089 Chr.5: 123588038-123588038 on Build GRCh38
rs948984554 Chr.5: 123588059-123588059 on Build GRCh38
rs561054689 Chr.5: 123588388-123588388 on Build GRCh38
rs998041845 Chr.5: 123588132-123588132 on Build GRCh38
rs751923502 Chr.5: 123588050-123588050 on Build GRCh38
rs545669054 Chr.5: 123588222-123588222 on Build GRCh38
rs376627411 Chr.5: 123588394-123588394 on Build GRCh38
rs771972676 Chr.5: 123588029-123588029 on Build GRCh38
rs758157687 Chr.5: 123588407-123588407 on Build GRCh38
rs1031758596 Chr.5: 123588353-123588353 on Build GRCh38
rs746918782 Chr.5: 123588444-123588444 on Build GRCh38
rs759248785 Chr.5: 123588187-123588187 on Build GRCh38
rs897135194 Chr.5: 123588349-123588349 on Build GRCh38
rs947869315 Chr.5: 123588285-123588285 on Build GRCh38
rs764863600 Chr.5: 123588188-123588188 on Build GRCh38
rs758821874 Chr.5: 123588158-123588158 on Build GRCh38
rs747927747 Chr.5: 123588404-123588404 on Build GRCh38
rs1027023374 Chr.5: 123588355-123588355 on Build GRCh38
rs781551880 Chr.5: 123588078-123588078 on Build GRCh38
rs1019826166 Chr.5: 123588403-123588403 on Build GRCh38
rs985568893 Chr.5: 123588398-123588398 on Build GRCh38
rs182600640 Chr.5: 123588337-123588337 on Build GRCh38
rs978004280 Chr.5: 123588244-123588244 on Build GRCh38
rs910126600 Chr.5: 123588462-123588462 on Build GRCh38
rs750902869 Chr.5: 123588382-123588382 on Build GRCh38
rs201734739 Chr.5: 123588459-123588459 on Build GRCh38
rs969133218 Chr.5: 123588315-123588315 on Build GRCh38
rs760223290 Chr.5: 123588177-123588177 on Build GRCh38
rs1022174100 Chr.5: 123588253-123588253 on Build GRCh38
rs144957358 Chr.5: 123588114-123588114 on Build GRCh38
rs769326739 Chr.5: 123588407-123588408 on Build GRCh38
rs770386858 Chr.5: 123588027-123588028 on Build GRCh38
rs1053484022 Chr.5: 123588283-123588283 on Build GRCh38
rs748615733 Chr.5: 123588026-123588027 on Build GRCh38
rs764495734 Chr.5: 123588048-123588048 on Build GRCh38
rs541069932 Chr.5: 123588103-123588103 on Build GRCh38
rs939667742 Chr.5: 123588098-123588098 on Build GRCh38
rs766079654 Chr.5: 123588035-123588035 on Build GRCh38
rs773046445 Chr.5: 123588174-123588174 on Build GRCh38
rs759301045 Chr.5: 123588039-123588039 on Build GRCh38
rs751881329 Chr.5: 123588190-123588190 on Build GRCh38
rs771989540 Chr.5: 123588138-123588138 on Build GRCh38
rs1028147929 Chr.5: 123588185-123588185 on Build GRCh38
rs373005811 Chr.5: 123588408-123588408 on Build GRCh38
rs578209661 Chr.5: 123588209-123588209 on Build GRCh38
rs747900819 Chr.5: 123588130-123588130 on Build GRCh38
rs960098746 Chr.5: 123588363-123588363 on Build GRCh38
rs1037221530 Chr.5: 123588125-123588125 on Build GRCh38
rs888111493 Chr.5: 123588087-123588087 on Build GRCh38
rs889594791 Chr.5: 123588235-123588235 on Build GRCh38
rs552932716 Chr.5: 123588182-123588182 on Build GRCh38
rs760276369 Chr.5: 123588034-123588034 on Build GRCh38
rs778586415 Chr.5: 123588094-123588094 on Build GRCh38
rs528554930 Chr.5: 123588411-123588411 on Build GRCh38
rs376153848 Chr.5: 123588393-123588393 on Build GRCh38
rs761799001 Chr.5: 123588027-123588027 on Build GRCh38
rs768926367 Chr.5: 123588120-123588120 on Build GRCh38
rs542764296 Chr.5: 123588379-123588379 on Build GRCh38
rs370024701 Chr.5: 123588032-123588032 on Build GRCh38
rs778766220 Chr.5: 123588121-123588121 on Build GRCh38
rs894869485 Chr.5: 123588284-123588284 on Build GRCh38
rs370721657 Chr.5: 123588396-123588396 on Build GRCh38

View Full Product Details