Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 126541888-126542386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH7A1
Entrez Gene ID
501
Gene Name
aldehyde dehydrogenase 7 family member A1
Gene Aliases
ATQ1, EPD, PDE
UniGene
Hs.483239
Gene Chromosome Location
Chr.5: 126541841-126595390 on Build GRCh38
Transcripts
NM_001182.4 NM_001201377.1 NM_001202404.1

SNP Detail

SNP ID Chromosome Location
rs930888047 Chr.5: 126541937-126541937 on Build GRCh38
rs948225017 Chr.5: 126541998-126542000 on Build GRCh38
rs905207304 Chr.5: 126542130-126542130 on Build GRCh38
rs780063692 Chr.5: 126542146-126542147 on Build GRCh38
rs1007418987 Chr.5: 126542147-126542148 on Build GRCh38
rs962266172 Chr.5: 126541926-126541926 on Build GRCh38
rs886059832 Chr.5: 126542144-126542145 on Build GRCh38
rs77525381 Chr.5: 126542149-126542149 on Build GRCh38
rs910764989 Chr.5: 126542298-126542298 on Build GRCh38
rs148309568 Chr.5: 126542182-126542182 on Build GRCh38
rs1019221520 Chr.5: 126542148-126542148 on Build GRCh38
rs189486979 Chr.5: 126542177-126542177 on Build GRCh38
rs989818285 Chr.5: 126542196-126542196 on Build GRCh38
rs184098590 Chr.5: 126542068-126542068 on Build GRCh38
rs756798087 Chr.5: 126541929-126541929 on Build GRCh38
rs550051176 Chr.5: 126541980-126541980 on Build GRCh38
rs749387811 Chr.5: 126542300-126542300 on Build GRCh38
rs5871217 Chr.5: 126541967-126541968 on Build GRCh38
rs920884186 Chr.5: 126541936-126541936 on Build GRCh38
rs886059835 Chr.5: 126542265-126542265 on Build GRCh38
rs11395324 Chr.5: 126542166-126542167 on Build GRCh38
rs1040134819 Chr.5: 126542058-126542058 on Build GRCh38
rs892942721 Chr.5: 126542073-126542073 on Build GRCh38
rs986311947 Chr.5: 126542016-126542016 on Build GRCh38
rs181028716 Chr.5: 126542201-126542201 on Build GRCh38
rs1054284940 Chr.5: 126542070-126542070 on Build GRCh38
rs1023281301 Chr.5: 126542099-126542099 on Build GRCh38
rs57369925 Chr.5: 126541961-126541961 on Build GRCh38
rs886059833 Chr.5: 126542147-126542147 on Build GRCh38
rs957043600 Chr.5: 126542194-126542194 on Build GRCh38
rs944414534 Chr.5: 126542305-126542305 on Build GRCh38
rs145424881 Chr.5: 126542123-126542123 on Build GRCh38
rs986362803 Chr.5: 126542249-126542249 on Build GRCh38
rs998808998 Chr.5: 126542146-126542146 on Build GRCh38
rs948193688 Chr.5: 126542288-126542288 on Build GRCh38
rs1045700495 Chr.5: 126542013-126542013 on Build GRCh38
rs201421709 Chr.5: 126542298-126542298 on Build GRCh38
rs939617301 Chr.5: 126542320-126542320 on Build GRCh38
rs976134399 Chr.5: 126542329-126542329 on Build GRCh38
rs944346904 Chr.5: 126542053-126542053 on Build GRCh38
rs775421792 Chr.5: 126542243-126542243 on Build GRCh38
rs368844061 Chr.5: 126542268-126542268 on Build GRCh38
rs59667289 Chr.5: 126542166-126542166 on Build GRCh38
rs1035768168 Chr.5: 126542171-126542171 on Build GRCh38
rs74415886 Chr.5: 126542362-126542362 on Build GRCh38
rs915375989 Chr.5: 126541987-126541987 on Build GRCh38
rs924420820 Chr.5: 126542350-126542350 on Build GRCh38
rs370866834 Chr.5: 126542036-126542036 on Build GRCh38
rs1035135579 Chr.5: 126542165-126542165 on Build GRCh38
rs370255749 Chr.5: 126542146-126542146 on Build GRCh38
rs192950824 Chr.5: 126542051-126542051 on Build GRCh38
rs1003940901 Chr.5: 126542147-126542147 on Build GRCh38
rs906796494 Chr.5: 126542021-126542021 on Build GRCh38
rs369855064 Chr.5: 126541979-126541979 on Build GRCh38
rs886059834 Chr.5: 126542184-126542184 on Build GRCh38
rs2035471 Chr.5: 126541943-126541943 on Build GRCh38
rs796191869 Chr.5: 126542069-126542069 on Build GRCh38
rs1012843515 Chr.5: 126542093-126542093 on Build GRCh38
rs1004231825 Chr.5: 126542037-126542037 on Build GRCh38
rs542701107 Chr.5: 126542080-126542081 on Build GRCh38
rs915678766 Chr.5: 126542233-126542233 on Build GRCh38
rs746329980 Chr.5: 126542059-126542059 on Build GRCh38
rs935747593 Chr.5: 126541941-126541941 on Build GRCh38
rs547600980 Chr.5: 126542074-126542074 on Build GRCh38

View Full Product Details