Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 126544351-126544860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH7A1
Entrez Gene ID
501
Gene Name
aldehyde dehydrogenase 7 family member A1
Gene Aliases
ATQ1, EPD, PDE
UniGene
Hs.483239
Gene Chromosome Location
Chr.5: 126541841-126595390 on Build GRCh38
Transcripts
NM_001182.4 NM_001201377.1 NM_001202404.1 XM_011543417.2 XM_017009493.1

SNP Detail

SNP ID Chromosome Location
rs946529644 Chr.5: 126544555-126544555 on Build GRCh38
rs775192146 Chr.5: 126544663-126544663 on Build GRCh38
rs1014306235 Chr.5: 126544757-126544758 on Build GRCh38
rs1001553962 Chr.5: 126544483-126544483 on Build GRCh38
rs1043290963 Chr.5: 126544468-126544468 on Build GRCh38
rs951201480 Chr.5: 126544547-126544547 on Build GRCh38
rs1022196214 Chr.5: 126544834-126544834 on Build GRCh38
rs542748598 Chr.5: 126544712-126544712 on Build GRCh38
rs930237249 Chr.5: 126544608-126544608 on Build GRCh38
rs781309579 Chr.5: 126544570-126544570 on Build GRCh38
rs904553449 Chr.5: 126544836-126544836 on Build GRCh38
rs982958181 Chr.5: 126544638-126544638 on Build GRCh38
rs1009488187 Chr.5: 126544797-126544797 on Build GRCh38
rs933755477 Chr.5: 126544589-126544589 on Build GRCh38
rs978883844 Chr.5: 126544506-126544506 on Build GRCh38
rs191020661 Chr.5: 126544505-126544505 on Build GRCh38
rs993404398 Chr.5: 126544582-126544582 on Build GRCh38
rs143317618 Chr.5: 126544753-126544753 on Build GRCh38
rs998414347 Chr.5: 126544380-126544380 on Build GRCh38
rs549472131 Chr.5: 126544765-126544765 on Build GRCh38
rs899562657 Chr.5: 126544684-126544684 on Build GRCh38
rs983999596 Chr.5: 126544555-126544556 on Build GRCh38
rs917533518 Chr.5: 126544594-126544594 on Build GRCh38
rs182676227 Chr.5: 126544510-126544510 on Build GRCh38
rs187827993 Chr.5: 126544816-126544816 on Build GRCh38
rs1060856 Chr.5: 126544417-126544417 on Build GRCh38
rs374361465 Chr.5: 126544825-126544825 on Build GRCh38
rs780328831 Chr.5: 126544377-126544377 on Build GRCh38
rs1020303264 Chr.5: 126544498-126544498 on Build GRCh38
rs886917113 Chr.5: 126544787-126544787 on Build GRCh38
rs755220331 Chr.5: 126544469-126544469 on Build GRCh38
rs922286721 Chr.5: 126544565-126544565 on Build GRCh38
rs556732113 Chr.5: 126544556-126544556 on Build GRCh38
rs1029608883 Chr.5: 126544411-126544411 on Build GRCh38
rs1025474645 Chr.5: 126544509-126544509 on Build GRCh38
rs570223278 Chr.5: 126544646-126544646 on Build GRCh38
rs904769449 Chr.5: 126544678-126544678 on Build GRCh38
rs574931385 Chr.5: 126544548-126544548 on Build GRCh38
rs899677964 Chr.5: 126544378-126544378 on Build GRCh38
rs55923017 Chr.5: 126544640-126544640 on Build GRCh38
rs1052171857 Chr.5: 126544597-126544597 on Build GRCh38
rs1010274186 Chr.5: 126544479-126544479 on Build GRCh38
rs933878443 Chr.5: 126544703-126544703 on Build GRCh38
rs112493228 Chr.5: 126544562-126544562 on Build GRCh38
rs1055796141 Chr.5: 126544740-126544740 on Build GRCh38
rs779037755 Chr.5: 126544534-126544534 on Build GRCh38
rs946234634 Chr.5: 126544644-126544644 on Build GRCh38
rs533678440 Chr.5: 126544561-126544561 on Build GRCh38
rs1051922586 Chr.5: 126544410-126544410 on Build GRCh38

View Full Product Details