Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 131160216-131160719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HINT1
Entrez Gene ID
3094
Gene Name
histidine triad nucleotide binding protein 1
Gene Aliases
HINT, NMAN, PKCI-1, PRKCNH1
UniGene
Hs.483305
Gene Chromosome Location
Chr.5: 131159283-131165348 on Build GRCh38
Transcripts
NM_005340.6 NR_024610.2 NR_024611.2 NR_073488.1 NR_134494.1 NR_134495.1

SNP Detail

SNP ID Chromosome Location
rs962876836 Chr.5: 131160441-131160441 on Build GRCh38
rs761407747 Chr.5: 131160355-131160355 on Build GRCh38
rs893357392 Chr.5: 131160584-131160584 on Build GRCh38
rs906047490 Chr.5: 131160287-131160287 on Build GRCh38
rs1054632241 Chr.5: 131160554-131160554 on Build GRCh38
rs758870389 Chr.5: 131160598-131160598 on Build GRCh38
rs927618614 Chr.5: 131160592-131160593 on Build GRCh38
rs185894916 Chr.5: 131160605-131160605 on Build GRCh38
rs989144127 Chr.5: 131160366-131160366 on Build GRCh38
rs1054975333 Chr.5: 131160644-131160644 on Build GRCh38
rs765645458 Chr.5: 131160491-131160491 on Build GRCh38
rs916470346 Chr.5: 131160678-131160678 on Build GRCh38
rs985359018 Chr.5: 131160433-131160433 on Build GRCh38
rs2551038 Chr.5: 131160446-131160446 on Build GRCh38
rs148490729 Chr.5: 131160509-131160509 on Build GRCh38
rs760448194 Chr.5: 131160484-131160484 on Build GRCh38
rs1045829230 Chr.5: 131160506-131160506 on Build GRCh38
rs572448652 Chr.5: 131160409-131160409 on Build GRCh38
rs767196680 Chr.5: 131160370-131160370 on Build GRCh38
rs966824018 Chr.5: 131160376-131160376 on Build GRCh38
rs1014347113 Chr.5: 131160266-131160266 on Build GRCh38
rs79379477 Chr.5: 131160296-131160296 on Build GRCh38
rs976547932 Chr.5: 131160384-131160384 on Build GRCh38
rs773846649 Chr.5: 131160323-131160323 on Build GRCh38
rs941332856 Chr.5: 131160457-131160457 on Build GRCh38
rs183543804 Chr.5: 131160497-131160497 on Build GRCh38
rs966416537 Chr.5: 131160654-131160654 on Build GRCh38
rs953891742 Chr.5: 131160403-131160403 on Build GRCh38
rs998332625 Chr.5: 131160686-131160686 on Build GRCh38
rs1024852623 Chr.5: 131160282-131160282 on Build GRCh38
rs922399722 Chr.5: 131160387-131160387 on Build GRCh38
rs554655155 Chr.5: 131160634-131160634 on Build GRCh38
rs1022351906 Chr.5: 131160274-131160274 on Build GRCh38
rs541826909 Chr.5: 131160247-131160247 on Build GRCh38
rs985705843 Chr.5: 131160417-131160417 on Build GRCh38
rs753012719 Chr.5: 131160508-131160508 on Build GRCh38
rs3852209 Chr.5: 131160322-131160322 on Build GRCh38

View Full Product Details