Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 131164725-131165218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HINT1
Entrez Gene ID
3094
Gene Name
histidine triad nucleotide binding protein 1
Gene Aliases
HINT, NMAN, PKCI-1, PRKCNH1
UniGene
Hs.483305
Gene Chromosome Location
Chr.5: 131159283-131165348 on Build GRCh38
Transcripts
NM_005340.6 NR_024610.2 NR_024611.2 NR_073488.1 NR_134494.1 NR_134495.1 XM_017009410.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6535682 c.40G>C p.G14R Chr.5: 131165166-131165166 on Build GRCh38
3993944 c.54C>A p.I18I Chr.5: 131165152-131165152 on Build GRCh38
5161864 c.99_100insT p.E34fs*1 Chr.5: 131165106-131165107 on Build GRCh38
5030460 c.91A>G p.I31V Chr.5: 131165115-131165115 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752131357 Chr.5: 131165067-131165067 on Build GRCh38
rs2278059 Chr.5: 131164867-131164867 on Build GRCh38
rs982282016 Chr.5: 131164934-131164934 on Build GRCh38
rs4696 Chr.5: 131165149-131165149 on Build GRCh38
rs940454651 Chr.5: 131164989-131164989 on Build GRCh38
rs1007779916 Chr.5: 131165084-131165084 on Build GRCh38
rs774181936 Chr.5: 131165120-131165120 on Build GRCh38
rs760249490 Chr.5: 131165148-131165148 on Build GRCh38
rs778441433 Chr.5: 131165015-131165015 on Build GRCh38
rs982339258 Chr.5: 131165171-131165171 on Build GRCh38
rs766117344 Chr.5: 131165059-131165059 on Build GRCh38
rs903136480 Chr.5: 131165045-131165045 on Build GRCh38
rs896368616 Chr.5: 131164781-131164781 on Build GRCh38
rs774274004 Chr.5: 131165048-131165048 on Build GRCh38
rs757554338 Chr.5: 131165184-131165184 on Build GRCh38
rs10070831 Chr.5: 131164779-131164779 on Build GRCh38
rs575806332 Chr.5: 131165025-131165025 on Build GRCh38
rs539829608 Chr.5: 131165064-131165064 on Build GRCh38
rs768380188 Chr.5: 131165043-131165043 on Build GRCh38
rs151094514 Chr.5: 131165172-131165172 on Build GRCh38
rs1019794186 Chr.5: 131164872-131164872 on Build GRCh38
rs745910594 Chr.5: 131165031-131165031 on Build GRCh38
rs770851222 Chr.5: 131165088-131165089 on Build GRCh38
rs551061023 Chr.5: 131164822-131164822 on Build GRCh38
rs887598461 Chr.5: 131164760-131164760 on Build GRCh38
rs558876472 Chr.5: 131165021-131165021 on Build GRCh38
rs1013476145 Chr.5: 131164782-131164782 on Build GRCh38
rs867670289 Chr.5: 131164921-131164921 on Build GRCh38
rs536666027 Chr.5: 131164869-131164869 on Build GRCh38
rs185252898 Chr.5: 131164827-131164827 on Build GRCh38
rs780266590 Chr.5: 131165097-131165097 on Build GRCh38
rs1030167603 Chr.5: 131164852-131164852 on Build GRCh38
rs773847241 Chr.5: 131164864-131164864 on Build GRCh38
rs1009671023 Chr.5: 131165099-131165099 on Build GRCh38
rs926794278 Chr.5: 131164941-131164941 on Build GRCh38
rs374378188 Chr.5: 131165051-131165051 on Build GRCh38
rs142857717 Chr.5: 131165053-131165053 on Build GRCh38
rs201262077 Chr.5: 131165135-131165135 on Build GRCh38
rs770361796 Chr.5: 131164815-131164815 on Build GRCh38
rs773013836 Chr.5: 131165147-131165147 on Build GRCh38
rs2278060 Chr.5: 131165058-131165058 on Build GRCh38
rs770090831 Chr.5: 131165036-131165036 on Build GRCh38
rs775684365 Chr.5: 131165037-131165037 on Build GRCh38
rs776457658 Chr.5: 131165165-131165165 on Build GRCh38
rs752008656 Chr.5: 131165173-131165173 on Build GRCh38
rs757310477 Chr.5: 131165022-131165022 on Build GRCh38
rs56697771 Chr.5: 131165081-131165081 on Build GRCh38
rs569078573 Chr.5: 131164807-131164807 on Build GRCh38
rs746787127 Chr.5: 131165065-131165066 on Build GRCh38
rs537137770 Chr.5: 131164820-131164820 on Build GRCh38
rs534053986 Chr.5: 131165119-131165119 on Build GRCh38
rs1055909797 Chr.5: 131164777-131164777 on Build GRCh38
rs1020229767 Chr.5: 131164794-131164794 on Build GRCh38
rs566118854 Chr.5: 131165040-131165040 on Build GRCh38
rs749444327 Chr.5: 131165113-131165113 on Build GRCh38
rs149782619 Chr.5: 131165096-131165096 on Build GRCh38
rs758826704 Chr.5: 131165012-131165012 on Build GRCh38
rs143635723 Chr.5: 131164887-131164887 on Build GRCh38
rs781517064 Chr.5: 131165080-131165080 on Build GRCh38
rs758477648 Chr.5: 131164980-131164980 on Build GRCh38
rs757828690 Chr.5: 131165069-131165069 on Build GRCh38
rs970665087 Chr.5: 131165169-131165169 on Build GRCh38
rs949632711 Chr.5: 131164925-131164925 on Build GRCh38
rs771734437 Chr.5: 131164890-131164890 on Build GRCh38
rs747865723 Chr.5: 131165133-131165133 on Build GRCh38
rs1032354755 Chr.5: 131165094-131165094 on Build GRCh38
rs768463956 Chr.5: 131165114-131165114 on Build GRCh38
rs2278058 Chr.5: 131164755-131164755 on Build GRCh38
rs748808451 Chr.5: 131164790-131164790 on Build GRCh38
rs781250694 Chr.5: 131165165-131165165 on Build GRCh38
rs921584461 Chr.5: 131164900-131164900 on Build GRCh38
rs777567225 Chr.5: 131165012-131165026 on Build GRCh38
rs768127548 Chr.5: 131165187-131165187 on Build GRCh38
rs532303774 Chr.5: 131164776-131164776 on Build GRCh38
rs140627344 Chr.5: 131164974-131164974 on Build GRCh38
rs758841138 Chr.5: 131165062-131165062 on Build GRCh38
rs746027226 Chr.5: 131165088-131165088 on Build GRCh38
rs112487594 Chr.5: 131164888-131164888 on Build GRCh38
rs969391375 Chr.5: 131164792-131164792 on Build GRCh38
rs972458319 Chr.5: 131164919-131164919 on Build GRCh38
rs959015488 Chr.5: 131164839-131164839 on Build GRCh38
rs984554551 Chr.5: 131164970-131164970 on Build GRCh38

View Full Product Details