Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132694991-132695529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF3A
Entrez Gene ID
11127
Gene Name
kinesin family member 3A
Gene Aliases
FLA10, KLP-20
UniGene
Hs.43670
Gene Chromosome Location
Chr.5: 132688808-132737633 on Build GRCh38
Transcripts
NM_001300791.1 NM_001300792.1 NM_007054.6 XM_006714526.3 XM_017008995.1 XM_017008996.1

SNP Detail

SNP ID Chromosome Location
rs115401962 Chr.5: 132695481-132695481 on Build GRCh38
rs765240503 Chr.5: 132695104-132695104 on Build GRCh38
rs919912827 Chr.5: 132695111-132695111 on Build GRCh38
rs547994884 Chr.5: 132695361-132695361 on Build GRCh38
rs989441945 Chr.5: 132695298-132695298 on Build GRCh38
rs755171856 Chr.5: 132695101-132695101 on Build GRCh38
rs983027725 Chr.5: 132695136-132695136 on Build GRCh38
rs914529642 Chr.5: 132695319-132695319 on Build GRCh38
rs534403024 Chr.5: 132695360-132695360 on Build GRCh38
rs915623882 Chr.5: 132695208-132695208 on Build GRCh38
rs926518316 Chr.5: 132695246-132695246 on Build GRCh38
rs149591075 Chr.5: 132695242-132695242 on Build GRCh38
rs952964188 Chr.5: 132695135-132695135 on Build GRCh38
rs546420155 Chr.5: 132695314-132695314 on Build GRCh38
rs752769773 Chr.5: 132695142-132695142 on Build GRCh38
rs942260919 Chr.5: 132695379-132695379 on Build GRCh38
rs377552306 Chr.5: 132695320-132695320 on Build GRCh38
rs191765622 Chr.5: 132695452-132695452 on Build GRCh38
rs548672342 Chr.5: 132695321-132695321 on Build GRCh38
rs144315164 Chr.5: 132695267-132695267 on Build GRCh38
rs1056883357 Chr.5: 132695154-132695154 on Build GRCh38
rs1050665744 Chr.5: 132695065-132695065 on Build GRCh38
rs868438169 Chr.5: 132695151-132695151 on Build GRCh38
rs923358306 Chr.5: 132695346-132695346 on Build GRCh38
rs897471018 Chr.5: 132695392-132695392 on Build GRCh38
rs757026054 Chr.5: 132695498-132695498 on Build GRCh38
rs970503155 Chr.5: 132695230-132695230 on Build GRCh38
rs765135626 Chr.5: 132695237-132695238 on Build GRCh38
rs1051029145 Chr.5: 132695353-132695353 on Build GRCh38
rs1024684865 Chr.5: 132695224-132695224 on Build GRCh38
rs754942602 Chr.5: 132695241-132695241 on Build GRCh38
rs993176597 Chr.5: 132695223-132695223 on Build GRCh38
rs778019855 Chr.5: 132695468-132695468 on Build GRCh38
rs993134201 Chr.5: 132695412-132695412 on Build GRCh38
rs1008346661 Chr.5: 132695040-132695040 on Build GRCh38
rs954542341 Chr.5: 132695036-132695036 on Build GRCh38
rs1024634028 Chr.5: 132695416-132695416 on Build GRCh38
rs889357788 Chr.5: 132695089-132695089 on Build GRCh38
rs1001935350 Chr.5: 132695437-132695437 on Build GRCh38
rs778059342 Chr.5: 132695247-132695247 on Build GRCh38
rs183965193 Chr.5: 132695461-132695461 on Build GRCh38
rs957911819 Chr.5: 132695457-132695457 on Build GRCh38
rs1037967569 Chr.5: 132695391-132695391 on Build GRCh38
rs980565879 Chr.5: 132695233-132695233 on Build GRCh38
rs889311732 Chr.5: 132695370-132695370 on Build GRCh38
rs933424397 Chr.5: 132695350-132695350 on Build GRCh38
rs958176788 Chr.5: 132695289-132695289 on Build GRCh38
rs1019708610 Chr.5: 132695449-132695449 on Build GRCh38
rs565780411 Chr.5: 132695352-132695352 on Build GRCh38
rs906290991 Chr.5: 132695417-132695417 on Build GRCh38
rs975789788 Chr.5: 132695095-132695095 on Build GRCh38
rs977463499 Chr.5: 132695331-132695331 on Build GRCh38
rs997687127 Chr.5: 132695046-132695046 on Build GRCh38
rs539607053 Chr.5: 132695463-132695463 on Build GRCh38
rs188614957 Chr.5: 132695143-132695143 on Build GRCh38

View Full Product Details