Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134969547-134970059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER3
Entrez Gene ID
347732
Gene Name
cation channel sperm associated 3
Gene Aliases
CACRC
UniGene
Hs.631804
Gene Chromosome Location
Chr.5: 134967906-135011707 on Build GRCh38
Transcripts
NM_178019.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
279588 c.161T>G p.I54R Chr.5: 134970001-134970001 on Build GRCh38
3850214 c.192G>A p.A64A Chr.5: 134970032-134970032 on Build GRCh38
6027122 c.204C>A p.A68A Chr.5: 134970044-134970044 on Build GRCh38
4534680 c.210G>A p.W70* Chr.5: 134970050-134970050 on Build GRCh38
3138306 c.186G>A p.S62S Chr.5: 134970026-134970026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145333202 Chr.5: 134969989-134969989 on Build GRCh38
rs778951845 Chr.5: 134969738-134969738 on Build GRCh38
rs528719821 Chr.5: 134969806-134969806 on Build GRCh38
rs574831794 Chr.5: 134969946-134969946 on Build GRCh38
rs992349976 Chr.5: 134969744-134969744 on Build GRCh38
rs368208576 Chr.5: 134969927-134969927 on Build GRCh38
rs374256296 Chr.5: 134970006-134970006 on Build GRCh38
rs989540573 Chr.5: 134969577-134969577 on Build GRCh38
rs747061105 Chr.5: 134969934-134969934 on Build GRCh38
rs980427122 Chr.5: 134969781-134969781 on Build GRCh38
rs12518563 Chr.5: 134969903-134969903 on Build GRCh38
rs115072996 Chr.5: 134969626-134969626 on Build GRCh38
rs766971508 Chr.5: 134969988-134969988 on Build GRCh38
rs765754154 Chr.5: 134969998-134969998 on Build GRCh38
rs371015408 Chr.5: 134969767-134969767 on Build GRCh38
rs756061764 Chr.5: 134970030-134970030 on Build GRCh38
rs1041530337 Chr.5: 134969959-134969959 on Build GRCh38
rs773873187 Chr.5: 134969974-134969974 on Build GRCh38
rs1047562498 Chr.5: 134969692-134969692 on Build GRCh38
rs751605598 Chr.5: 134970015-134970015 on Build GRCh38
rs536922872 Chr.5: 134970026-134970026 on Build GRCh38
rs531680169 Chr.5: 134969905-134969905 on Build GRCh38
rs758393074 Chr.5: 134969921-134969921 on Build GRCh38
rs116119095 Chr.5: 134969956-134969956 on Build GRCh38
rs936598219 Chr.5: 134969899-134969899 on Build GRCh38
rs529069942 Chr.5: 134969613-134969613 on Build GRCh38
rs960220087 Chr.5: 134969741-134969741 on Build GRCh38
rs750206641 Chr.5: 134969929-134969929 on Build GRCh38
rs149386608 Chr.5: 134969737-134969740 on Build GRCh38
rs767208333 Chr.5: 134969896-134969896 on Build GRCh38
rs200787247 Chr.5: 134969961-134969961 on Build GRCh38
rs150251956 Chr.5: 134970025-134970025 on Build GRCh38
rs1052340755 Chr.5: 134969904-134969904 on Build GRCh38
rs774996033 Chr.5: 134969958-134969958 on Build GRCh38
rs1031103396 Chr.5: 134969817-134969817 on Build GRCh38
rs1019688684 Chr.5: 134969621-134969621 on Build GRCh38
rs748556391 Chr.5: 134969730-134969730 on Build GRCh38
rs899871515 Chr.5: 134969622-134969622 on Build GRCh38
rs190448178 Chr.5: 134969637-134969637 on Build GRCh38
rs375185027 Chr.5: 134970032-134970032 on Build GRCh38
rs181082627 Chr.5: 134969720-134969720 on Build GRCh38
rs757144500 Chr.5: 134969939-134969939 on Build GRCh38
rs958086028 Chr.5: 134969572-134969572 on Build GRCh38
rs977189458 Chr.5: 134969636-134969638 on Build GRCh38
rs933229601 Chr.5: 134969652-134969652 on Build GRCh38
rs755443631 Chr.5: 134969894-134969894 on Build GRCh38
rs776913359 Chr.5: 134969657-134969657 on Build GRCh38
rs753014864 Chr.5: 134970000-134970000 on Build GRCh38
rs906750475 Chr.5: 134969769-134969769 on Build GRCh38
rs1011056961 Chr.5: 134969874-134969874 on Build GRCh38
rs761253604 Chr.5: 134969976-134969976 on Build GRCh38
rs764251114 Chr.5: 134970013-134970013 on Build GRCh38
rs982930570 Chr.5: 134969708-134969708 on Build GRCh38
rs1046489613 Chr.5: 134969752-134969752 on Build GRCh38
rs140980470 Chr.5: 134969963-134969963 on Build GRCh38
rs779835422 Chr.5: 134970031-134970031 on Build GRCh38
rs765524134 Chr.5: 134969915-134969915 on Build GRCh38
rs139264127 Chr.5: 134970033-134970033 on Build GRCh38
rs757373052 Chr.5: 134970023-134970023 on Build GRCh38
rs941738120 Chr.5: 134969709-134969709 on Build GRCh38
rs773884693 Chr.5: 134969981-134969981 on Build GRCh38
rs745636557 Chr.5: 134969950-134969950 on Build GRCh38
rs74567041 Chr.5: 134969745-134969745 on Build GRCh38
rs755971687 Chr.5: 134970032-134970033 on Build GRCh38

View Full Product Details