Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140440883-140441341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKHD1
Entrez Gene ID
54882
Gene Name
ankyrin repeat and KH domain containing 1
Gene Aliases
MASK, MASK1, PP2500, VBARP
UniGene
Hs.594084
Gene Chromosome Location
Chr.5: 140401814-140539856 on Build GRCh38
Transcripts
NM_001197030.1 NM_017747.2 NM_017978.2 NM_024668.3 NM_020690.5

Gene Symbol
ANKHD1-EIF4EBP3
Entrez Gene ID
404734
Gene Name
ANKHD1-EIF4EBP3 readthrough
Gene Aliases
MASK-BP3, MASK-BP3ARF
UniGene
Hs.594084
Gene Chromosome Location
Chr.5: 140401814-140549578 on Build GRCh38
Transcripts
NM_001197030.1 NM_017747.2 NM_017978.2 NM_024668.3 NM_020690.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6101819 c.856G>A p.D286N Chr.5: 140441085-140441085 on Build GRCh38
5645028 c.766-2A>T p.? Chr.5: 140440993-140440993 on Build GRCh38
4510020 c.824C>T p.P275L Chr.5: 140441053-140441053 on Build GRCh38
6209418 c.824C>G p.P275R Chr.5: 140441053-140441053 on Build GRCh38
1061636 c.781C>T p.H261Y Chr.5: 140441010-140441010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777563311 Chr.5: 140440956-140440956 on Build GRCh38
rs539909768 Chr.5: 140440933-140440933 on Build GRCh38
rs749057875 Chr.5: 140440957-140440957 on Build GRCh38
rs750076888 Chr.5: 140440974-140440975 on Build GRCh38
rs747081570 Chr.5: 140441142-140441142 on Build GRCh38
rs534783218 Chr.5: 140441111-140441111 on Build GRCh38
rs763489987 Chr.5: 140441163-140441163 on Build GRCh38
rs755792611 Chr.5: 140440953-140440953 on Build GRCh38
rs546989506 Chr.5: 140441125-140441125 on Build GRCh38
rs763148290 Chr.5: 140440946-140440946 on Build GRCh38
rs367661304 Chr.5: 140441140-140441140 on Build GRCh38
rs200350497 Chr.5: 140441090-140441090 on Build GRCh38
rs1052247322 Chr.5: 140440916-140440916 on Build GRCh38
rs750355964 Chr.5: 140441126-140441126 on Build GRCh38
rs557340931 Chr.5: 140441265-140441265 on Build GRCh38
rs777006053 Chr.5: 140441028-140441028 on Build GRCh38
rs763682115 Chr.5: 140441105-140441105 on Build GRCh38
rs752158402 Chr.5: 140441096-140441096 on Build GRCh38
rs560097963 Chr.5: 140441264-140441264 on Build GRCh38
rs896334235 Chr.5: 140440954-140440954 on Build GRCh38
rs768775888 Chr.5: 140440988-140440988 on Build GRCh38
rs757027493 Chr.5: 140441018-140441018 on Build GRCh38
rs1057411738 Chr.5: 140441185-140441185 on Build GRCh38
rs572879866 Chr.5: 140441287-140441287 on Build GRCh38
rs1019641143 Chr.5: 140440996-140440996 on Build GRCh38
rs762300623 Chr.5: 140441040-140441040 on Build GRCh38
rs1027482350 Chr.5: 140441231-140441231 on Build GRCh38
rs371550206 Chr.5: 140440977-140440977 on Build GRCh38
rs140694523 Chr.5: 140441087-140441087 on Build GRCh38
rs772123651 Chr.5: 140440973-140440973 on Build GRCh38
rs937301602 Chr.5: 140441167-140441167 on Build GRCh38
rs755808129 Chr.5: 140441172-140441173 on Build GRCh38
rs375851435 Chr.5: 140441169-140441169 on Build GRCh38
rs757022151 Chr.5: 140441119-140441119 on Build GRCh38
rs758272012 Chr.5: 140441133-140441133 on Build GRCh38
rs761318568 Chr.5: 140440962-140440964 on Build GRCh38
rs972883548 Chr.5: 140441045-140441045 on Build GRCh38
rs867698876 Chr.5: 140441187-140441187 on Build GRCh38
rs199675809 Chr.5: 140441149-140441149 on Build GRCh38
rs983882539 Chr.5: 140441137-140441137 on Build GRCh38
rs768885772 Chr.5: 140441147-140441147 on Build GRCh38
rs945695807 Chr.5: 140441269-140441269 on Build GRCh38
rs918665998 Chr.5: 140441132-140441132 on Build GRCh38
rs760321270 Chr.5: 140441098-140441098 on Build GRCh38
rs905157791 Chr.5: 140441131-140441131 on Build GRCh38
rs1042093237 Chr.5: 140440936-140440936 on Build GRCh38
rs766127894 Chr.5: 140441179-140441179 on Build GRCh38
rs757142587 Chr.5: 140440964-140440964 on Build GRCh38
rs951465467 Chr.5: 140441255-140441255 on Build GRCh38
rs935149932 Chr.5: 140440912-140440912 on Build GRCh38
rs1000999631 Chr.5: 140440909-140440909 on Build GRCh38
rs904232882 Chr.5: 140441299-140441299 on Build GRCh38
rs774126235 Chr.5: 140440980-140440980 on Build GRCh38
rs552482309 Chr.5: 140441072-140441072 on Build GRCh38
rs760070473 Chr.5: 140441184-140441184 on Build GRCh38
rs78779489 Chr.5: 140441084-140441084 on Build GRCh38
rs778787618 Chr.5: 140440966-140440966 on Build GRCh38
rs781241037 Chr.5: 140441148-140441148 on Build GRCh38
rs773784133 Chr.5: 140441158-140441158 on Build GRCh38
rs770271469 Chr.5: 140441154-140441154 on Build GRCh38
rs372916599 Chr.5: 140441168-140441168 on Build GRCh38
rs766989209 Chr.5: 140440945-140440945 on Build GRCh38
rs767001442 Chr.5: 140440969-140440972 on Build GRCh38
rs535753269 Chr.5: 140441205-140441205 on Build GRCh38
rs1032596693 Chr.5: 140440922-140440922 on Build GRCh38
rs775020190 Chr.5: 140441175-140441175 on Build GRCh38
rs373818155 Chr.5: 140440982-140440982 on Build GRCh38
rs745854947 Chr.5: 140440972-140440972 on Build GRCh38
rs770326358 Chr.5: 140441046-140441046 on Build GRCh38
rs545639063 Chr.5: 140441305-140441305 on Build GRCh38

View Full Product Details