Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140458543-140459054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKHD1
Entrez Gene ID
54882
Gene Name
ankyrin repeat and KH domain containing 1
Gene Aliases
MASK, MASK1, PP2500, VBARP
UniGene
Hs.594084
Gene Chromosome Location
Chr.5: 140401814-140539856 on Build GRCh38
Transcripts
NM_001197030.1 NM_017747.2 NM_017978.2 NM_024668.3 NM_020690.5

Gene Symbol
ANKHD1-EIF4EBP3
Entrez Gene ID
404734
Gene Name
ANKHD1-EIF4EBP3 readthrough
Gene Aliases
MASK-BP3, MASK-BP3ARF
UniGene
Hs.594084
Gene Chromosome Location
Chr.5: 140401814-140549578 on Build GRCh38
Transcripts
NM_001197030.1 NM_017747.2 NM_017978.2 NM_024668.3 NM_020690.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7280322 c.1328C>T p.T443M Chr.5: 140458710-140458710 on Build GRCh38
7280321 c.1328C>T p.T443M Chr.5: 140458710-140458710 on Build GRCh38
4604344 c.1442A>G p.E481G Chr.5: 140458824-140458824 on Build GRCh38
4819394 c.1470C>T p.L490L Chr.5: 140458852-140458852 on Build GRCh38
5934938 c.1439G>A p.R480Q Chr.5: 140458821-140458821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576200187 Chr.5: 140458576-140458576 on Build GRCh38
rs867506390 Chr.5: 140458958-140458958 on Build GRCh38
rs780917334 Chr.5: 140458907-140458907 on Build GRCh38
rs12522423 Chr.5: 140458996-140458996 on Build GRCh38
rs59992464 Chr.5: 140458980-140459013 on Build GRCh38
rs964969989 Chr.5: 140458621-140458621 on Build GRCh38
rs750756450 Chr.5: 140458646-140458646 on Build GRCh38
rs865972608 Chr.5: 140459019-140459019 on Build GRCh38
rs867393409 Chr.5: 140458967-140458967 on Build GRCh38
rs778602550 Chr.5: 140458599-140458599 on Build GRCh38
rs565002604 Chr.5: 140458705-140458705 on Build GRCh38
rs868241952 Chr.5: 140458743-140458743 on Build GRCh38
rs755432417 Chr.5: 140458703-140458703 on Build GRCh38
rs760630306 Chr.5: 140458612-140458612 on Build GRCh38
rs983157298 Chr.5: 140458706-140458706 on Build GRCh38
rs768660815 Chr.5: 140458620-140458620 on Build GRCh38
rs749974307 Chr.5: 140458881-140458881 on Build GRCh38
rs745570630 Chr.5: 140458603-140458603 on Build GRCh38
rs746175836 Chr.5: 140458603-140458607 on Build GRCh38
rs867517487 Chr.5: 140459001-140459001 on Build GRCh38
rs774661962 Chr.5: 140458822-140458822 on Build GRCh38
rs67470460 Chr.5: 140459021-140459022 on Build GRCh38
rs13181293 Chr.5: 140458980-140458980 on Build GRCh38
rs367846132 Chr.5: 140458839-140458839 on Build GRCh38
rs940140530 Chr.5: 140458720-140458720 on Build GRCh38
rs1030405187 Chr.5: 140458672-140458672 on Build GRCh38
rs137981861 Chr.5: 140458785-140458785 on Build GRCh38
rs766666489 Chr.5: 140458686-140458686 on Build GRCh38
rs372989365 Chr.5: 140458884-140458884 on Build GRCh38
rs371416734 Chr.5: 140458869-140458869 on Build GRCh38
rs527992081 Chr.5: 140458957-140458957 on Build GRCh38
rs529452262 Chr.5: 140458998-140458999 on Build GRCh38
rs756580738 Chr.5: 140458871-140458871 on Build GRCh38
rs765628006 Chr.5: 140458591-140458591 on Build GRCh38
rs13182482 Chr.5: 140458981-140458981 on Build GRCh38
rs373990442 Chr.5: 140458792-140458792 on Build GRCh38
rs748094929 Chr.5: 140458795-140458795 on Build GRCh38
rs531815286 Chr.5: 140459028-140459028 on Build GRCh38
rs376959794 Chr.5: 140458645-140458645 on Build GRCh38
rs773369317 Chr.5: 140458966-140458981 on Build GRCh38
rs777026290 Chr.5: 140458599-140458600 on Build GRCh38
rs748053504 Chr.5: 140458913-140458913 on Build GRCh38
rs974747283 Chr.5: 140458726-140458726 on Build GRCh38
rs866504423 Chr.5: 140459008-140459008 on Build GRCh38
rs543629262 Chr.5: 140458596-140458596 on Build GRCh38
rs540544442 Chr.5: 140458945-140458946 on Build GRCh38
rs745444334 Chr.5: 140458754-140458754 on Build GRCh38
rs747212907 Chr.5: 140458582-140458582 on Build GRCh38
rs867910523 Chr.5: 140458710-140458710 on Build GRCh38
rs756775082 Chr.5: 140458722-140458722 on Build GRCh38
rs751937989 Chr.5: 140458699-140458699 on Build GRCh38
rs763107019 Chr.5: 140458805-140458805 on Build GRCh38
rs748856776 Chr.5: 140458595-140458595 on Build GRCh38
rs370555001 Chr.5: 140458776-140458776 on Build GRCh38
rs12517445 Chr.5: 140458997-140458997 on Build GRCh38
rs907195127 Chr.5: 140458771-140458771 on Build GRCh38
rs559047271 Chr.5: 140458949-140458949 on Build GRCh38
rs763230445 Chr.5: 140458678-140458678 on Build GRCh38
rs764744392 Chr.5: 140458880-140458880 on Build GRCh38
rs60792257 Chr.5: 140458968-140458968 on Build GRCh38
rs867280525 Chr.5: 140458966-140458966 on Build GRCh38
rs773286779 Chr.5: 140458802-140458802 on Build GRCh38
rs746708176 Chr.5: 140458894-140458894 on Build GRCh38
rs561610861 Chr.5: 140459007-140459007 on Build GRCh38
rs771986177 Chr.5: 140458607-140458607 on Build GRCh38
rs778350646 Chr.5: 140458738-140458738 on Build GRCh38
rs57733337 Chr.5: 140458969-140458969 on Build GRCh38
rs868418492 Chr.5: 140458959-140458959 on Build GRCh38
rs758134675 Chr.5: 140458755-140458755 on Build GRCh38
rs866560069 Chr.5: 140459009-140459009 on Build GRCh38
rs753240242 Chr.5: 140458870-140458870 on Build GRCh38
rs549451384 Chr.5: 140458965-140458965 on Build GRCh38
rs771319947 Chr.5: 140458596-140458598 on Build GRCh38
rs754790793 Chr.5: 140458905-140458905 on Build GRCh38
rs541045034 Chr.5: 140458927-140458927 on Build GRCh38
rs868495414 Chr.5: 140459018-140459018 on Build GRCh38
rs576986791 Chr.5: 140458711-140458711 on Build GRCh38
rs574439746 Chr.5: 140458619-140458619 on Build GRCh38
rs59283405 Chr.5: 140458979-140458980 on Build GRCh38
rs746830674 Chr.5: 140458772-140458772 on Build GRCh38
rs779822603 Chr.5: 140458885-140458885 on Build GRCh38
rs371377581 Chr.5: 140458666-140458666 on Build GRCh38
rs374354738 Chr.5: 140458762-140458762 on Build GRCh38
rs776532697 Chr.5: 140458635-140458635 on Build GRCh38
rs765283258 Chr.5: 140458658-140458658 on Build GRCh38
rs948972316 Chr.5: 140458745-140458745 on Build GRCh38
rs766613690 Chr.5: 140458819-140458819 on Build GRCh38

View Full Product Details