Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141655270-141655764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARAP3
Entrez Gene ID
64411
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3
Gene Aliases
CENTD3, DRAG1
UniGene
Hs.726187
Gene Chromosome Location
Chr.5: 141653401-141682233 on Build GRCh38
Transcripts
NM_022481.5 XM_005268497.1 XM_005268498.1 XM_005268499.1 XM_006714792.1 XM_011537677.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1434755 c.3992T>C p.V1331A Chr.5: 141655739-141655739 on Build GRCh38
3994074 c.3985C>G p.Q1329E Chr.5: 141655746-141655746 on Build GRCh38
7356090 c.4111C>T p.R1371W Chr.5: 141655400-141655400 on Build GRCh38
4851661 c.4030C>T p.Q1344* Chr.5: 141655701-141655701 on Build GRCh38
4599659 c.3978T>G p.D1326E Chr.5: 141655753-141655753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995392164 Chr.5: 141655442-141655442 on Build GRCh38
rs746250184 Chr.5: 141655614-141655614 on Build GRCh38
rs764752528 Chr.5: 141655723-141655723 on Build GRCh38
rs375439028 Chr.5: 141655400-141655400 on Build GRCh38
rs148152980 Chr.5: 141655639-141655639 on Build GRCh38
rs369467470 Chr.5: 141655404-141655404 on Build GRCh38
rs574237739 Chr.5: 141655295-141655295 on Build GRCh38
rs2306340 Chr.5: 141655681-141655681 on Build GRCh38
rs768062603 Chr.5: 141655338-141655338 on Build GRCh38
rs771830541 Chr.5: 141655576-141655576 on Build GRCh38
rs375543507 Chr.5: 141655593-141655593 on Build GRCh38
rs905977392 Chr.5: 141655451-141655451 on Build GRCh38
rs746807026 Chr.5: 141655731-141655731 on Build GRCh38
rs766718819 Chr.5: 141655605-141655605 on Build GRCh38
rs759842878 Chr.5: 141655354-141655354 on Build GRCh38
rs749277998 Chr.5: 141655426-141655426 on Build GRCh38
rs777855990 Chr.5: 141655315-141655315 on Build GRCh38
rs376037371 Chr.5: 141655305-141655305 on Build GRCh38
rs72792351 Chr.5: 141655383-141655383 on Build GRCh38
rs898315456 Chr.5: 141655430-141655430 on Build GRCh38
rs755797274 Chr.5: 141655608-141655608 on Build GRCh38
rs774834869 Chr.5: 141655686-141655686 on Build GRCh38
rs778825308 Chr.5: 141655443-141655443 on Build GRCh38
rs34237538 Chr.5: 141655299-141655299 on Build GRCh38
rs1002975678 Chr.5: 141655462-141655462 on Build GRCh38
rs200830343 Chr.5: 141655410-141655410 on Build GRCh38
rs1021915849 Chr.5: 141655563-141655563 on Build GRCh38
rs377203403 Chr.5: 141655524-141655524 on Build GRCh38
rs770750790 Chr.5: 141655438-141655438 on Build GRCh38
rs201780469 Chr.5: 141655667-141655667 on Build GRCh38
rs557663444 Chr.5: 141655361-141655361 on Build GRCh38
rs1044857712 Chr.5: 141655448-141655448 on Build GRCh38
rs1010897051 Chr.5: 141655498-141655498 on Build GRCh38
rs778718658 Chr.5: 141655634-141655634 on Build GRCh38
rs764097681 Chr.5: 141655384-141655384 on Build GRCh38
rs374804842 Chr.5: 141655418-141655418 on Build GRCh38
rs758147299 Chr.5: 141655642-141655642 on Build GRCh38
rs141682206 Chr.5: 141655385-141655385 on Build GRCh38
rs750355617 Chr.5: 141655727-141655727 on Build GRCh38
rs776332564 Chr.5: 141655353-141655353 on Build GRCh38
rs141039995 Chr.5: 141655646-141655646 on Build GRCh38
rs763242488 Chr.5: 141655604-141655604 on Build GRCh38
rs912249880 Chr.5: 141655414-141655414 on Build GRCh38
rs542882607 Chr.5: 141655371-141655371 on Build GRCh38
rs369505822 Chr.5: 141655668-141655668 on Build GRCh38
rs748649591 Chr.5: 141655356-141655359 on Build GRCh38
rs753462754 Chr.5: 141655707-141655707 on Build GRCh38
rs775700507 Chr.5: 141655577-141655577 on Build GRCh38
rs972458607 Chr.5: 141655580-141655580 on Build GRCh38
rs779869242 Chr.5: 141655730-141655730 on Build GRCh38
rs754729678 Chr.5: 141655399-141655399 on Build GRCh38
rs142211119 Chr.5: 141655671-141655671 on Build GRCh38
rs758768178 Chr.5: 141655658-141655658 on Build GRCh38
rs540037842 Chr.5: 141655541-141655541 on Build GRCh38
rs772397508 Chr.5: 141655422-141655422 on Build GRCh38
rs114921133 Chr.5: 141655583-141655583 on Build GRCh38
rs763833196 Chr.5: 141655704-141655704 on Build GRCh38
rs2112261 Chr.5: 141655461-141655461 on Build GRCh38
rs376503540 Chr.5: 141655514-141655514 on Build GRCh38
rs368868045 Chr.5: 141655423-141655423 on Build GRCh38
rs765773102 Chr.5: 141655416-141655416 on Build GRCh38
rs768856824 Chr.5: 141655655-141655655 on Build GRCh38
rs775699105 Chr.5: 141655374-141655374 on Build GRCh38
rs529225343 Chr.5: 141655589-141655589 on Build GRCh38
rs753385527 Chr.5: 141655621-141655621 on Build GRCh38
rs749196255 Chr.5: 141655329-141655329 on Build GRCh38
rs762262959 Chr.5: 141655386-141655386 on Build GRCh38
rs750878671 Chr.5: 141655600-141655600 on Build GRCh38
rs768750584 Chr.5: 141655586-141655586 on Build GRCh38
rs1034442017 Chr.5: 141655473-141655473 on Build GRCh38
rs138955685 Chr.5: 141655663-141655663 on Build GRCh38
rs1050687690 Chr.5: 141655411-141655411 on Build GRCh38
rs138515824 Chr.5: 141655703-141655703 on Build GRCh38
rs145534243 Chr.5: 141655631-141655631 on Build GRCh38
rs777293917 Chr.5: 141655324-141655324 on Build GRCh38
rs776656679 Chr.5: 141655588-141655588 on Build GRCh38
rs55977140 Chr.5: 141655682-141655682 on Build GRCh38
rs147712909 Chr.5: 141655649-141655649 on Build GRCh38
rs79464052 Chr.5: 141655607-141655607 on Build GRCh38
rs529010789 Chr.5: 141655584-141655584 on Build GRCh38
rs763746634 Chr.5: 141655610-141655610 on Build GRCh38
rs773769179 Chr.5: 141655450-141655453 on Build GRCh38
rs745644933 Chr.5: 141655449-141655449 on Build GRCh38
rs1035650779 Chr.5: 141655481-141655481 on Build GRCh38
rs756729270 Chr.5: 141655711-141655711 on Build GRCh38
rs770781745 Chr.5: 141655332-141655332 on Build GRCh38
rs369710090 Chr.5: 141655728-141655728 on Build GRCh38
rs143698821 Chr.5: 141655396-141655396 on Build GRCh38
rs986518079 Chr.5: 141655633-141655633 on Build GRCh38
rs368395538 Chr.5: 141655339-141655339 on Build GRCh38
rs766303865 Chr.5: 141655445-141655445 on Build GRCh38
rs372420144 Chr.5: 141655397-141655397 on Build GRCh38
rs368935621 Chr.5: 141655594-141655594 on Build GRCh38
rs758929341 Chr.5: 141655594-141655609 on Build GRCh38

View Full Product Details