Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148481461-148481965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR4
Entrez Gene ID
3360
Gene Name
5-hydroxytryptamine receptor 4
Gene Aliases
5-HT4, 5-HT4R
UniGene
Hs.483773
Gene Chromosome Location
Chr.5: 148451032-148654527 on Build GRCh38
Transcripts
NM_000870.6 NM_001040169.2 NM_001040172.2 NM_001040173.2 NM_001286410.1 NM_199453.3 NR_104445.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5459170 c.1180T>C p.S394P Chr.5: 148481526-148481526 on Build GRCh38
6860311 c.1170_1171insA p.A391fs*6 Chr.5: 148481535-148481536 on Build GRCh38
5484749 c.1212G>A p.K404K Chr.5: 148481494-148481494 on Build GRCh38
6211921 c.1170delA p.A391fs*8 Chr.5: 148481536-148481536 on Build GRCh38
5512166 c.1161A>G p.R387R Chr.5: 148481545-148481545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910483874 Chr.5: 148481507-148481507 on Build GRCh38
rs946098214 Chr.5: 148481887-148481887 on Build GRCh38
rs559656368 Chr.5: 148481789-148481789 on Build GRCh38
rs201864275 Chr.5: 148481774-148481774 on Build GRCh38
rs545169183 Chr.5: 148481535-148481535 on Build GRCh38
rs192960935 Chr.5: 148481622-148481622 on Build GRCh38
rs1040591624 Chr.5: 148481780-148481780 on Build GRCh38
rs779333667 Chr.5: 148481702-148481702 on Build GRCh38
rs201822624 Chr.5: 148481917-148481917 on Build GRCh38
rs988709775 Chr.5: 148481570-148481570 on Build GRCh38
rs770443161 Chr.5: 148481535-148481536 on Build GRCh38
rs926080244 Chr.5: 148481929-148481929 on Build GRCh38
rs764839152 Chr.5: 148481496-148481496 on Build GRCh38
rs201079723 Chr.5: 148481515-148481515 on Build GRCh38
rs971025626 Chr.5: 148481616-148481616 on Build GRCh38
rs1034421888 Chr.5: 148481933-148481933 on Build GRCh38
rs200338220 Chr.5: 148481631-148481631 on Build GRCh38
rs199569491 Chr.5: 148481700-148481700 on Build GRCh38
rs1010172835 Chr.5: 148481832-148481832 on Build GRCh38
rs904593318 Chr.5: 148481593-148481593 on Build GRCh38
rs767603592 Chr.5: 148481869-148481869 on Build GRCh38
rs201442133 Chr.5: 148481717-148481717 on Build GRCh38
rs961774850 Chr.5: 148481669-148481669 on Build GRCh38
rs1014427984 Chr.5: 148481645-148481645 on Build GRCh38
rs201394278 Chr.5: 148481689-148481689 on Build GRCh38
rs200658163 Chr.5: 148481793-148481793 on Build GRCh38
rs201254223 Chr.5: 148481599-148481599 on Build GRCh38
rs569743741 Chr.5: 148481892-148481892 on Build GRCh38
rs199867556 Chr.5: 148481830-148481830 on Build GRCh38
rs200048367 Chr.5: 148481788-148481788 on Build GRCh38
rs1032870961 Chr.5: 148481800-148481800 on Build GRCh38
rs201015815 Chr.5: 148481886-148481886 on Build GRCh38
rs951855979 Chr.5: 148481740-148481740 on Build GRCh38
rs200445396 Chr.5: 148481539-148481539 on Build GRCh38
rs533266619 Chr.5: 148481818-148481818 on Build GRCh38
rs200826327 Chr.5: 148481712-148481712 on Build GRCh38
rs868393780 Chr.5: 148481587-148481587 on Build GRCh38
rs1000000263 Chr.5: 148481530-148481530 on Build GRCh38
rs184374791 Chr.5: 148481522-148481522 on Build GRCh38
rs770420966 Chr.5: 148481723-148481723 on Build GRCh38
rs866093557 Chr.5: 148481546-148481546 on Build GRCh38
rs758793667 Chr.5: 148481540-148481540 on Build GRCh38
rs377191925 Chr.5: 148481650-148481650 on Build GRCh38
rs976036602 Chr.5: 148481853-148481853 on Build GRCh38
rs996244809 Chr.5: 148481509-148481509 on Build GRCh38
rs549189995 Chr.5: 148481936-148481936 on Build GRCh38
rs184169540 Chr.5: 148481771-148481771 on Build GRCh38
rs755494029 Chr.5: 148481648-148481648 on Build GRCh38
rs186845398 Chr.5: 148481562-148481562 on Build GRCh38
rs901864548 Chr.5: 148481784-148481784 on Build GRCh38
rs201970020 Chr.5: 148481810-148481810 on Build GRCh38
rs199630659 Chr.5: 148481755-148481755 on Build GRCh38
rs1049261760 Chr.5: 148481752-148481752 on Build GRCh38
rs1024501751 Chr.5: 148481653-148481655 on Build GRCh38
rs1042083745 Chr.5: 148481904-148481904 on Build GRCh38
rs200219058 Chr.5: 148481598-148481598 on Build GRCh38
rs58336229 Chr.5: 148481541-148481541 on Build GRCh38
rs537301570 Chr.5: 148481905-148481905 on Build GRCh38
rs1001356687 Chr.5: 148481797-148481797 on Build GRCh38
rs746300490 Chr.5: 148481536-148481536 on Build GRCh38
rs913790769 Chr.5: 148481873-148481873 on Build GRCh38
rs201300949 Chr.5: 148481680-148481680 on Build GRCh38
rs199937443 Chr.5: 148481508-148481508 on Build GRCh38
rs145729392 Chr.5: 148481614-148481614 on Build GRCh38

View Full Product Details