Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148482249-148482756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR4
Entrez Gene ID
3360
Gene Name
5-hydroxytryptamine receptor 4
Gene Aliases
5-HT4, 5-HT4R
UniGene
Hs.483773
Gene Chromosome Location
Chr.5: 148451032-148654527 on Build GRCh38
Transcripts
NM_000870.6 NM_001040169.2 NM_001040172.2 NM_001040173.2 NM_001286410.1 NM_199453.3 NR_104445.1

SNP Detail

SNP ID Chromosome Location
rs878914756 Chr.5: 148482502-148482502 on Build GRCh38
rs577451505 Chr.5: 148482375-148482375 on Build GRCh38
rs200466318 Chr.5: 148482395-148482395 on Build GRCh38
rs932260494 Chr.5: 148482390-148482390 on Build GRCh38
rs773747421 Chr.5: 148482436-148482436 on Build GRCh38
rs201495367 Chr.5: 148482439-148482439 on Build GRCh38
rs140347566 Chr.5: 148482596-148482596 on Build GRCh38
rs201131114 Chr.5: 148482565-148482565 on Build GRCh38
rs979794980 Chr.5: 148482393-148482393 on Build GRCh38
rs200242169 Chr.5: 148482420-148482420 on Build GRCh38
rs146751166 Chr.5: 148482442-148482442 on Build GRCh38
rs1038698319 Chr.5: 148482509-148482509 on Build GRCh38
rs182014157 Chr.5: 148482609-148482609 on Build GRCh38
rs772601828 Chr.5: 148482417-148482417 on Build GRCh38
rs200019895 Chr.5: 148482519-148482519 on Build GRCh38
rs1012887435 Chr.5: 148482613-148482613 on Build GRCh38
rs951788489 Chr.5: 148482343-148482343 on Build GRCh38
rs950978510 Chr.5: 148482669-148482669 on Build GRCh38
rs1001486923 Chr.5: 148482451-148482451 on Build GRCh38
rs755898174 Chr.5: 148482549-148482551 on Build GRCh38
rs201736104 Chr.5: 148482422-148482422 on Build GRCh38
rs199724780 Chr.5: 148482427-148482427 on Build GRCh38
rs200945291 Chr.5: 148482522-148482522 on Build GRCh38
rs200529858 Chr.5: 148482430-148482430 on Build GRCh38
rs925490214 Chr.5: 148482699-148482699 on Build GRCh38
rs1022881136 Chr.5: 148482634-148482634 on Build GRCh38
rs907015867 Chr.5: 148482446-148482446 on Build GRCh38
rs1000684051 Chr.5: 148482589-148482589 on Build GRCh38
rs953644142 Chr.5: 148482635-148482635 on Build GRCh38
rs201244832 Chr.5: 148482367-148482367 on Build GRCh38
rs202080867 Chr.5: 148482614-148482614 on Build GRCh38
rs909484189 Chr.5: 148482693-148482694 on Build GRCh38
rs997378640 Chr.5: 148482362-148482362 on Build GRCh38
rs201865311 Chr.5: 148482535-148482535 on Build GRCh38
rs202082115 Chr.5: 148482338-148482338 on Build GRCh38
rs1030487707 Chr.5: 148482627-148482627 on Build GRCh38
rs1014603867 Chr.5: 148482358-148482358 on Build GRCh38
rs1007214511 Chr.5: 148482288-148482288 on Build GRCh38
rs919346992 Chr.5: 148482662-148482662 on Build GRCh38
rs201186530 Chr.5: 148482293-148482293 on Build GRCh38
rs866863737 Chr.5: 148482344-148482344 on Build GRCh38
rs79836777 Chr.5: 148482691-148482691 on Build GRCh38
rs762384256 Chr.5: 148482399-148482399 on Build GRCh38
rs549178112 Chr.5: 148482716-148482716 on Build GRCh38
rs887469381 Chr.5: 148482473-148482473 on Build GRCh38
rs201358674 Chr.5: 148482303-148482303 on Build GRCh38
rs373299287 Chr.5: 148482429-148482429 on Build GRCh38
rs55969025 Chr.5: 148482586-148482586 on Build GRCh38
rs988579257 Chr.5: 148482723-148482723 on Build GRCh38
rs761054632 Chr.5: 148482569-148482569 on Build GRCh38
rs35939085 Chr.5: 148482305-148482306 on Build GRCh38
rs943563462 Chr.5: 148482701-148482701 on Build GRCh38
rs943030030 Chr.5: 148482508-148482508 on Build GRCh38
rs1034041523 Chr.5: 148482309-148482309 on Build GRCh38
rs200387050 Chr.5: 148482657-148482657 on Build GRCh38
rs984569536 Chr.5: 148482346-148482346 on Build GRCh38
rs966661898 Chr.5: 148482385-148482385 on Build GRCh38
rs199861666 Chr.5: 148482300-148482300 on Build GRCh38
rs997643772 Chr.5: 148482570-148482570 on Build GRCh38
rs754164727 Chr.5: 148482588-148482588 on Build GRCh38
rs200987028 Chr.5: 148482444-148482444 on Build GRCh38
rs1032254723 Chr.5: 148482394-148482394 on Build GRCh38
rs79096841 Chr.5: 148482404-148482404 on Build GRCh38
rs201305067 Chr.5: 148482681-148482681 on Build GRCh38
rs202200971 Chr.5: 148482295-148482295 on Build GRCh38
rs201806754 Chr.5: 148482454-148482454 on Build GRCh38
rs929482175 Chr.5: 148482433-148482433 on Build GRCh38
rs887476350 Chr.5: 148482278-148482278 on Build GRCh38
rs976256314 Chr.5: 148482371-148482371 on Build GRCh38
rs200266253 Chr.5: 148482350-148482350 on Build GRCh38
rs199965643 Chr.5: 148482544-148482544 on Build GRCh38
rs199623862 Chr.5: 148482416-148482416 on Build GRCh38
rs971462038 Chr.5: 148482649-148482649 on Build GRCh38
rs1020696799 Chr.5: 148482584-148482584 on Build GRCh38
rs202135200 Chr.5: 148482384-148482384 on Build GRCh38
rs143534451 Chr.5: 148482724-148482724 on Build GRCh38

View Full Product Details