Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148523141-148523647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR4
Entrez Gene ID
3360
Gene Name
5-hydroxytryptamine receptor 4
Gene Aliases
5-HT4, 5-HT4R
UniGene
Hs.483773
Gene Chromosome Location
Chr.5: 148451032-148654527 on Build GRCh38
Transcripts
NM_000870.6 NM_001040169.2 NM_001040172.2 NM_001040173.2 NM_001286410.1 NM_199453.3 NR_104445.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6576203 c.405T>A p.P135P Chr.5: 148523295-148523295 on Build GRCh38
5479374 c.450G>A p.T150T Chr.5: 148523250-148523250 on Build GRCh38
736444 c.401C>A p.T134N Chr.5: 148523299-148523299 on Build GRCh38
736443 c.401C>A p.T134N Chr.5: 148523299-148523299 on Build GRCh38
1737000 c.403C>G p.P135A Chr.5: 148523297-148523297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777321602 Chr.5: 148523263-148523263 on Build GRCh38
rs776059787 Chr.5: 148523385-148523385 on Build GRCh38
rs544150856 Chr.5: 148523479-148523479 on Build GRCh38
rs1030053702 Chr.5: 148523257-148523257 on Build GRCh38
rs1051849767 Chr.5: 148523559-148523559 on Build GRCh38
rs748010908 Chr.5: 148523259-148523259 on Build GRCh38
rs535426797 Chr.5: 148523553-148523553 on Build GRCh38
rs201029786 Chr.5: 148523324-148523324 on Build GRCh38
rs757324943 Chr.5: 148523501-148523501 on Build GRCh38
rs748739412 Chr.5: 148523272-148523272 on Build GRCh38
rs565545791 Chr.5: 148523568-148523568 on Build GRCh38
rs2277049 Chr.5: 148523525-148523525 on Build GRCh38
rs764165547 Chr.5: 148523408-148523408 on Build GRCh38
rs764685473 Chr.5: 148523296-148523296 on Build GRCh38
rs1032512599 Chr.5: 148523570-148523570 on Build GRCh38
rs755634253 Chr.5: 148523261-148523261 on Build GRCh38
rs770653018 Chr.5: 148523281-148523281 on Build GRCh38
rs527342626 Chr.5: 148523402-148523402 on Build GRCh38
rs781359980 Chr.5: 148523183-148523183 on Build GRCh38
rs200816055 Chr.5: 148523255-148523255 on Build GRCh38
rs112373372 Chr.5: 148523520-148523520 on Build GRCh38
rs756670124 Chr.5: 148523373-148523373 on Build GRCh38
rs201964147 Chr.5: 148523290-148523290 on Build GRCh38
rs199857988 Chr.5: 148523307-148523307 on Build GRCh38
rs376809075 Chr.5: 148523339-148523339 on Build GRCh38
rs199980452 Chr.5: 148523366-148523366 on Build GRCh38
rs534758839 Chr.5: 148523521-148523524 on Build GRCh38
rs1003947007 Chr.5: 148523231-148523231 on Build GRCh38
rs756106837 Chr.5: 148523353-148523353 on Build GRCh38
rs955180998 Chr.5: 148523521-148523525 on Build GRCh38
rs888893908 Chr.5: 148523560-148523560 on Build GRCh38
rs748875247 Chr.5: 148523370-148523370 on Build GRCh38
rs146910658 Chr.5: 148523313-148523313 on Build GRCh38
rs201815307 Chr.5: 148523340-148523340 on Build GRCh38
rs941947786 Chr.5: 148523497-148523497 on Build GRCh38
rs2277050 Chr.5: 148523409-148523409 on Build GRCh38
rs909046235 Chr.5: 148523457-148523457 on Build GRCh38
rs997168795 Chr.5: 148523548-148523548 on Build GRCh38
rs988407598 Chr.5: 148523363-148523363 on Build GRCh38
rs760871391 Chr.5: 148523184-148523184 on Build GRCh38
rs750006766 Chr.5: 148523590-148523590 on Build GRCh38
rs988126264 Chr.5: 148523576-148523576 on Build GRCh38
rs746263123 Chr.5: 148523286-148523286 on Build GRCh38
rs986699119 Chr.5: 148523350-148523350 on Build GRCh38
rs768115367 Chr.5: 148523164-148523164 on Build GRCh38
rs199886610 Chr.5: 148523185-148523185 on Build GRCh38
rs200046865 Chr.5: 148523251-148523251 on Build GRCh38
rs747374233 Chr.5: 148523386-148523386 on Build GRCh38
rs79129596 Chr.5: 148523375-148523375 on Build GRCh38
rs200634941 Chr.5: 148523388-148523388 on Build GRCh38
rs754130194 Chr.5: 148523227-148523227 on Build GRCh38
rs1055672273 Chr.5: 148523421-148523421 on Build GRCh38
rs568325522 Chr.5: 148523535-148523535 on Build GRCh38
rs958314950 Chr.5: 148523299-148523299 on Build GRCh38
rs750569562 Chr.5: 148523331-148523331 on Build GRCh38
rs201866413 Chr.5: 148523382-148523382 on Build GRCh38
rs201950460 Chr.5: 148523285-148523285 on Build GRCh38
rs1003877760 Chr.5: 148523460-148523460 on Build GRCh38
rs570436308 Chr.5: 148523438-148523438 on Build GRCh38
rs745449224 Chr.5: 148523378-148523378 on Build GRCh38
rs573733073 Chr.5: 148523441-148523441 on Build GRCh38
rs201869932 Chr.5: 148523241-148523241 on Build GRCh38
rs750827426 Chr.5: 148523382-148523383 on Build GRCh38
rs781137239 Chr.5: 148523258-148523258 on Build GRCh38
rs201353257 Chr.5: 148523338-148523338 on Build GRCh38
rs776982804 Chr.5: 148523390-148523390 on Build GRCh38
rs191977381 Chr.5: 148523489-148523489 on Build GRCh38
rs759348473 Chr.5: 148523174-148523174 on Build GRCh38
rs897497128 Chr.5: 148523529-148523529 on Build GRCh38
rs761768921 Chr.5: 148523310-148523310 on Build GRCh38
rs368907191 Chr.5: 148523349-148523349 on Build GRCh38
rs200514272 Chr.5: 148523176-148523176 on Build GRCh38
rs186099216 Chr.5: 148523291-148523291 on Build GRCh38
rs553538259 Chr.5: 148523564-148523564 on Build GRCh38
rs200667420 Chr.5: 148523250-148523250 on Build GRCh38
rs886808721 Chr.5: 148523447-148523447 on Build GRCh38
rs141026046 Chr.5: 148523220-148523220 on Build GRCh38
rs1003773525 Chr.5: 148523430-148523430 on Build GRCh38
rs539499410 Chr.5: 148523574-148523574 on Build GRCh38

View Full Product Details