Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149271647-149272179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1L1
Entrez Gene ID
134265
Gene Name
actin filament associated protein 1 like 1
Gene Aliases
-
UniGene
Hs.483793
Gene Chromosome Location
Chr.5: 149271838-149343637 on Build GRCh38
Transcripts
NM_001146337.2 NM_001323062.1 NM_001323063.1 NM_152406.3

SNP Detail

SNP ID Chromosome Location
rs909796940 Chr.5: 149271928-149271928 on Build GRCh38
rs559565437 Chr.5: 149271925-149271925 on Build GRCh38
rs923912134 Chr.5: 149271718-149271718 on Build GRCh38
rs941229533 Chr.5: 149272122-149272122 on Build GRCh38
rs933709137 Chr.5: 149271972-149271972 on Build GRCh38
rs570797446 Chr.5: 149271684-149271684 on Build GRCh38
rs958117241 Chr.5: 149271721-149271721 on Build GRCh38
rs975093723 Chr.5: 149272124-149272124 on Build GRCh38
rs1051937614 Chr.5: 149271983-149271983 on Build GRCh38
rs375581729 Chr.5: 149271864-149271864 on Build GRCh38
rs929914146 Chr.5: 149271922-149271922 on Build GRCh38
rs996342035 Chr.5: 149271705-149271705 on Build GRCh38
rs374424011 Chr.5: 149272114-149272114 on Build GRCh38
rs1031006946 Chr.5: 149271841-149271841 on Build GRCh38
rs879263531 Chr.5: 149271955-149271955 on Build GRCh38
rs1010068986 Chr.5: 149272044-149272044 on Build GRCh38
rs537867429 Chr.5: 149271813-149271813 on Build GRCh38
rs947930851 Chr.5: 149271730-149271730 on Build GRCh38
rs554331403 Chr.5: 149271945-149271945 on Build GRCh38
rs927764110 Chr.5: 149271899-149271899 on Build GRCh38
rs1031254821 Chr.5: 149272009-149272009 on Build GRCh38
rs938079624 Chr.5: 149271683-149271683 on Build GRCh38
rs1010893371 Chr.5: 149272039-149272039 on Build GRCh38
rs901415434 Chr.5: 149271714-149271714 on Build GRCh38
rs909829359 Chr.5: 149272120-149272120 on Build GRCh38
rs867020798 Chr.5: 149271754-149271754 on Build GRCh38
rs751294554 Chr.5: 149271707-149271707 on Build GRCh38
rs959462652 Chr.5: 149272154-149272154 on Build GRCh38
rs371634293 Chr.5: 149272107-149272107 on Build GRCh38
rs927934544 Chr.5: 149271752-149271752 on Build GRCh38
rs1046642945 Chr.5: 149271751-149271751 on Build GRCh38
rs1048564769 Chr.5: 149271695-149271695 on Build GRCh38
rs559967087 Chr.5: 149271792-149271792 on Build GRCh38
rs899842223 Chr.5: 149272089-149272089 on Build GRCh38
rs1023701249 Chr.5: 149271870-149271870 on Build GRCh38
rs887017127 Chr.5: 149271849-149271849 on Build GRCh38
rs1005315054 Chr.5: 149271760-149271760 on Build GRCh38
rs1034869284 Chr.5: 149272078-149272078 on Build GRCh38
rs533817120 Chr.5: 149272034-149272034 on Build GRCh38
rs969433924 Chr.5: 149271876-149271876 on Build GRCh38
rs750476998 Chr.5: 149271726-149271726 on Build GRCh38
rs576742016 Chr.5: 149272061-149272061 on Build GRCh38
rs771857107 Chr.5: 149272058-149272059 on Build GRCh38
rs999916803 Chr.5: 149271994-149271994 on Build GRCh38
rs1041029609 Chr.5: 149272084-149272084 on Build GRCh38
rs543874551 Chr.5: 149272100-149272100 on Build GRCh38
rs910025873 Chr.5: 149271697-149271697 on Build GRCh38
rs933762226 Chr.5: 149272158-149272158 on Build GRCh38
rs958066781 Chr.5: 149271857-149271857 on Build GRCh38
rs982545257 Chr.5: 149272138-149272138 on Build GRCh38
rs890728979 Chr.5: 149271840-149271840 on Build GRCh38
rs996387231 Chr.5: 149271831-149271831 on Build GRCh38
rs1039512013 Chr.5: 149271809-149271809 on Build GRCh38
rs1049403027 Chr.5: 149271729-149271729 on Build GRCh38
rs558603165 Chr.5: 149272055-149272055 on Build GRCh38
rs12054749 Chr.5: 149272148-149272148 on Build GRCh38
rs62378114 Chr.5: 149271832-149271832 on Build GRCh38
rs941028149 Chr.5: 149271931-149271931 on Build GRCh38
rs1039373058 Chr.5: 149271937-149271937 on Build GRCh38
rs989488370 Chr.5: 149271722-149271722 on Build GRCh38
rs940295928 Chr.5: 149271699-149271699 on Build GRCh38
rs1037652017 Chr.5: 149271704-149271704 on Build GRCh38
rs982616486 Chr.5: 149272111-149272111 on Build GRCh38
rs981980556 Chr.5: 149271893-149271893 on Build GRCh38
rs971326935 Chr.5: 149272133-149272133 on Build GRCh38
rs989439650 Chr.5: 149271868-149271868 on Build GRCh38
rs955115419 Chr.5: 149271710-149271710 on Build GRCh38
rs999689400 Chr.5: 149272002-149272002 on Build GRCh38
rs141004478 Chr.5: 149271947-149271947 on Build GRCh38
rs1032853449 Chr.5: 149272022-149272022 on Build GRCh38

View Full Product Details