Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149306257-149306762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1L1
Entrez Gene ID
134265
Gene Name
actin filament associated protein 1 like 1
Gene Aliases
-
UniGene
Hs.483793
Gene Chromosome Location
Chr.5: 149271838-149343637 on Build GRCh38
Transcripts
NM_001146337.2 NM_001323062.1 NM_001323063.1 NM_152406.3 XM_011537558.1 XM_017009036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
482326 c.491G>T p.S164I Chr.5: 149306360-149306360 on Build GRCh38
1182136 c.510G>T p.R170S Chr.5: 149306379-149306379 on Build GRCh38
5466337 c.464T>C p.V155A Chr.5: 149306333-149306333 on Build GRCh38
3941181 c.483C>T p.D161D Chr.5: 149306352-149306352 on Build GRCh38
3852643 c.498C>T p.Y166Y Chr.5: 149306367-149306367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375197456 Chr.5: 149306352-149306352 on Build GRCh38
rs375381815 Chr.5: 149306577-149306577 on Build GRCh38
rs369988842 Chr.5: 149306578-149306578 on Build GRCh38
rs199519861 Chr.5: 149306309-149306309 on Build GRCh38
rs371770954 Chr.5: 149306345-149306345 on Build GRCh38
rs770906538 Chr.5: 149306342-149306342 on Build GRCh38
rs953685670 Chr.5: 149306671-149306671 on Build GRCh38
rs550069903 Chr.5: 149306389-149306389 on Build GRCh38
rs577136952 Chr.5: 149306336-149306336 on Build GRCh38
rs772080451 Chr.5: 149306324-149306324 on Build GRCh38
rs570732464 Chr.5: 149306323-149306323 on Build GRCh38
rs373926033 Chr.5: 149306509-149306509 on Build GRCh38
rs772116959 Chr.5: 149306430-149306430 on Build GRCh38
rs184818004 Chr.5: 149306353-149306353 on Build GRCh38
rs552457940 Chr.5: 149306281-149306281 on Build GRCh38
rs781206348 Chr.5: 149306393-149306393 on Build GRCh38
rs780146879 Chr.5: 149306380-149306380 on Build GRCh38
rs779281062 Chr.5: 149306318-149306318 on Build GRCh38
rs749033706 Chr.5: 149306340-149306340 on Build GRCh38
rs760254303 Chr.5: 149306447-149306447 on Build GRCh38
rs774464815 Chr.5: 149306421-149306421 on Build GRCh38
rs1027040374 Chr.5: 149306538-149306538 on Build GRCh38
rs750878464 Chr.5: 149306354-149306354 on Build GRCh38
rs750782012 Chr.5: 149306464-149306464 on Build GRCh38
rs768069570 Chr.5: 149306463-149306463 on Build GRCh38
rs895773891 Chr.5: 149306456-149306456 on Build GRCh38
rs976444671 Chr.5: 149306374-149306374 on Build GRCh38
rs965690106 Chr.5: 149306601-149306601 on Build GRCh38
rs141207215 Chr.5: 149306392-149306392 on Build GRCh38
rs189292207 Chr.5: 149306561-149306561 on Build GRCh38
rs932406099 Chr.5: 149306361-149306361 on Build GRCh38
rs978387350 Chr.5: 149306711-149306711 on Build GRCh38
rs756581727 Chr.5: 149306283-149306283 on Build GRCh38
rs888519342 Chr.5: 149306541-149306541 on Build GRCh38
rs905152778 Chr.5: 149306487-149306487 on Build GRCh38
rs757837319 Chr.5: 149306310-149306310 on Build GRCh38
rs1016174060 Chr.5: 149306581-149306581 on Build GRCh38
rs368770665 Chr.5: 149306288-149306288 on Build GRCh38
rs1002305924 Chr.5: 149306502-149306502 on Build GRCh38
rs906590055 Chr.5: 149306552-149306552 on Build GRCh38
rs925731016 Chr.5: 149306720-149306720 on Build GRCh38
rs749123492 Chr.5: 149306395-149306395 on Build GRCh38
rs148231801 Chr.5: 149306313-149306313 on Build GRCh38
rs370534234 Chr.5: 149306386-149306386 on Build GRCh38
rs952830343 Chr.5: 149306285-149306285 on Build GRCh38
rs763803154 Chr.5: 149306449-149306449 on Build GRCh38
rs772404581 Chr.5: 149306514-149306514 on Build GRCh38
rs1053346017 Chr.5: 149306420-149306420 on Build GRCh38
rs537332788 Chr.5: 149306422-149306422 on Build GRCh38
rs780835501 Chr.5: 149306721-149306721 on Build GRCh38
rs754125774 Chr.5: 149306303-149306303 on Build GRCh38
rs954926206 Chr.5: 149306385-149306385 on Build GRCh38
rs747190987 Chr.5: 149306325-149306325 on Build GRCh38
rs758504414 Chr.5: 149306370-149306370 on Build GRCh38
rs775194668 Chr.5: 149306439-149306439 on Build GRCh38
rs554174125 Chr.5: 149306483-149306483 on Build GRCh38
rs770586787 Chr.5: 149306413-149306413 on Build GRCh38
rs535435718 Chr.5: 149306468-149306468 on Build GRCh38
rs764848425 Chr.5: 149306658-149306658 on Build GRCh38
rs773116721 Chr.5: 149306328-149306328 on Build GRCh38
rs200939762 Chr.5: 149306327-149306327 on Build GRCh38
rs760448524 Chr.5: 149306349-149306349 on Build GRCh38
rs759142458 Chr.5: 149306343-149306343 on Build GRCh38
rs558422522 Chr.5: 149306616-149306616 on Build GRCh38

View Full Product Details