Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149374094-149374634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL17B
Entrez Gene ID
27190
Gene Name
interleukin 17B
Gene Aliases
IL-17B, IL-20, NIRF, ZCYTO7
UniGene
Hs.156979
Gene Chromosome Location
Chr.5: 149374267-149404202 on Build GRCh38
Transcripts
NM_001317987.1 NM_014443.2 XM_011537622.2 XM_017009346.1 XM_017009347.1 XM_017009348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4550760 c.508G>A p.E170K Chr.5: 149374404-149374404 on Build GRCh38
1435085 c.498C>T p.R166R Chr.5: 149374414-149374414 on Build GRCh38
3211554 c.527G>A p.C176Y Chr.5: 149374385-149374385 on Build GRCh38
4692182 c.340G>A p.V114M Chr.5: 149374572-149374572 on Build GRCh38
3211557 c.475C>T p.R159C Chr.5: 149374437-149374437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147582010 Chr.5: 149374443-149374443 on Build GRCh38
rs150719306 Chr.5: 149374414-149374414 on Build GRCh38
rs774894146 Chr.5: 149374377-149374377 on Build GRCh38
rs765240048 Chr.5: 149374463-149374463 on Build GRCh38
rs777144026 Chr.5: 149374349-149374349 on Build GRCh38
rs770682054 Chr.5: 149374369-149374369 on Build GRCh38
rs752743282 Chr.5: 149374437-149374437 on Build GRCh38
rs775703909 Chr.5: 149374450-149374450 on Build GRCh38
rs933815623 Chr.5: 149374424-149374424 on Build GRCh38
rs773424096 Chr.5: 149374464-149374464 on Build GRCh38
rs540819324 Chr.5: 149374266-149374266 on Build GRCh38
rs954053525 Chr.5: 149374594-149374594 on Build GRCh38
rs373011468 Chr.5: 149374580-149374580 on Build GRCh38
rs777015384 Chr.5: 149374521-149374521 on Build GRCh38
rs374213956 Chr.5: 149374505-149374505 on Build GRCh38
rs2227461 Chr.5: 149374444-149374444 on Build GRCh38
rs778562390 Chr.5: 149374494-149374494 on Build GRCh38
rs757977589 Chr.5: 149374436-149374437 on Build GRCh38
rs896724282 Chr.5: 149374560-149374560 on Build GRCh38
rs371293844 Chr.5: 149374544-149374544 on Build GRCh38
rs773319996 Chr.5: 149374591-149374591 on Build GRCh38
rs770259524 Chr.5: 149374590-149374590 on Build GRCh38
rs778537657 Chr.5: 149374353-149374365 on Build GRCh38
rs772510012 Chr.5: 149374390-149374390 on Build GRCh38
rs777391829 Chr.5: 149374441-149374441 on Build GRCh38
rs753773022 Chr.5: 149374490-149374490 on Build GRCh38
rs758105855 Chr.5: 149374497-149374497 on Build GRCh38
rs562575563 Chr.5: 149374422-149374422 on Build GRCh38
rs1015717341 Chr.5: 149374445-149374445 on Build GRCh38
rs770992357 Chr.5: 149374605-149374605 on Build GRCh38
rs560682874 Chr.5: 149374306-149374306 on Build GRCh38
rs550871130 Chr.5: 149374461-149374461 on Build GRCh38
rs201298520 Chr.5: 149374385-149374385 on Build GRCh38
rs763153907 Chr.5: 149374473-149374473 on Build GRCh38
rs745574528 Chr.5: 149374496-149374496 on Build GRCh38
rs144395727 Chr.5: 149374458-149374458 on Build GRCh38
rs763211165 Chr.5: 149374545-149374545 on Build GRCh38
rs1052840455 Chr.5: 149374415-149374415 on Build GRCh38
rs139906316 Chr.5: 149374474-149374474 on Build GRCh38
rs140917979 Chr.5: 149374554-149374554 on Build GRCh38
rs1037161396 Chr.5: 149374389-149374389 on Build GRCh38
rs528081511 Chr.5: 149374413-149374413 on Build GRCh38
rs766478963 Chr.5: 149374551-149374551 on Build GRCh38
rs761086057 Chr.5: 149374395-149374395 on Build GRCh38
rs777552982 Chr.5: 149374528-149374529 on Build GRCh38
rs755299932 Chr.5: 149374578-149374578 on Build GRCh38
rs149908263 Chr.5: 149374442-149374442 on Build GRCh38
rs757888351 Chr.5: 149374572-149374572 on Build GRCh38
rs754779395 Chr.5: 149374427-149374427 on Build GRCh38
rs781128792 Chr.5: 149374337-149374337 on Build GRCh38
rs897120821 Chr.5: 149374575-149374575 on Build GRCh38
rs756453286 Chr.5: 149374421-149374421 on Build GRCh38
rs748708206 Chr.5: 149374581-149374581 on Build GRCh38
rs201150206 Chr.5: 149374522-149374522 on Build GRCh38
rs557148061 Chr.5: 149374239-149374239 on Build GRCh38
rs992685382 Chr.5: 149374258-149374258 on Build GRCh38
rs932580756 Chr.5: 149374324-149374324 on Build GRCh38
rs749925422 Chr.5: 149374569-149374569 on Build GRCh38
rs769924513 Chr.5: 149374534-149374534 on Build GRCh38
rs757247536 Chr.5: 149374561-149374561 on Build GRCh38
rs2227462 Chr.5: 149374203-149374203 on Build GRCh38
rs751326386 Chr.5: 149374321-149374321 on Build GRCh38
rs41291955 Chr.5: 149374486-149374486 on Build GRCh38
rs776072998 Chr.5: 149374391-149374391 on Build GRCh38
rs748585346 Chr.5: 149374528-149374528 on Build GRCh38
rs926877779 Chr.5: 149374297-149374297 on Build GRCh38
rs367765361 Chr.5: 149374487-149374487 on Build GRCh38
rs943710020 Chr.5: 149374408-149374408 on Build GRCh38
rs201386909 Chr.5: 149374457-149374457 on Build GRCh38
rs762063982 Chr.5: 149374416-149374416 on Build GRCh38
rs904115900 Chr.5: 149374201-149374201 on Build GRCh38
rs760004479 Chr.5: 149374384-149374384 on Build GRCh38
rs753894362 Chr.5: 149374558-149374558 on Build GRCh38
rs202136839 Chr.5: 149374460-149374460 on Build GRCh38
rs143579457 Chr.5: 149374516-149374516 on Build GRCh38
rs1006706587 Chr.5: 149374599-149374600 on Build GRCh38
rs746616674 Chr.5: 149374574-149374574 on Build GRCh38
rs115498474 Chr.5: 149374240-149374240 on Build GRCh38
rs774447320 Chr.5: 149374608-149374608 on Build GRCh38
rs745448406 Chr.5: 149374447-149374447 on Build GRCh38
rs377701351 Chr.5: 149374506-149374506 on Build GRCh38
rs769350081 Chr.5: 149374344-149374344 on Build GRCh38
rs749401276 Chr.5: 149374600-149374600 on Build GRCh38
rs369715279 Chr.5: 149374492-149374492 on Build GRCh38
rs369120658 Chr.5: 149374339-149374339 on Build GRCh38
rs749024161 Chr.5: 149374365-149374365 on Build GRCh38
rs754730200 Chr.5: 149374336-149374336 on Build GRCh38
rs973891845 Chr.5: 149374300-149374300 on Build GRCh38
rs1022531480 Chr.5: 149374139-149374139 on Build GRCh38
rs774590961 Chr.5: 149374553-149374553 on Build GRCh38
rs778496736 Chr.5: 149374412-149374412 on Build GRCh38
rs2227460 Chr.5: 149374573-149374573 on Build GRCh38
rs764889728 Chr.5: 149374562-149374562 on Build GRCh38
rs138991563 Chr.5: 149374436-149374436 on Build GRCh38
rs755991452 Chr.5: 149374438-149374438 on Build GRCh38
rs752411869 Chr.5: 149374485-149374485 on Build GRCh38
rs532107768 Chr.5: 149374448-149374448 on Build GRCh38

View Full Product Details