Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151512463-151512963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT2
Entrez Gene ID
2196
Gene Name
FAT atypical cadherin 2
Gene Aliases
CDHF8, CDHR9, HFAT2, MEGF1
UniGene
Hs.591255
Gene Chromosome Location
Chr.5: 151504092-151594822 on Build GRCh38
Transcripts
NM_001447.2 XM_006714761.3 XM_011537600.2 XM_011537603.2 XM_017009224.1 XM_017009225.1 XM_011537595.2 XM_017009217.1 XM_017009219.1

Gene Symbol
SLC36A1
Entrez Gene ID
206358
Gene Name
solute carrier family 36 member 1
Gene Aliases
Dct1, LYAAT1, PAT1, TRAMD3
UniGene
Hs.269004
Gene Chromosome Location
Chr.5: 151411344-151556318 on Build GRCh38
Transcripts
NM_001447.2 XM_006714761.3 XM_011537600.2 XM_011537603.2 XM_017009224.1 XM_017009225.1 XM_011537595.2 XM_017009217.1 XM_017009219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5638384 c.11565C>T p.I3855I Chr.5: 151512505-151512505 on Build GRCh38
5495003 c.11570_11581del12 p.V3857_M3860delVEEM Chr.5: 151512489-151512500 on Build GRCh38
1206579 c.11557C>T p.H3853Y Chr.5: 151512513-151512513 on Build GRCh38
1754137 c.11482C>T p.Q3828* Chr.5: 151512588-151512588 on Build GRCh38
3994125 c.11582A>G p.D3861G Chr.5: 151512488-151512488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114246360 Chr.5: 151512619-151512619 on Build GRCh38
rs781458716 Chr.5: 151512488-151512488 on Build GRCh38
rs774191244 Chr.5: 151512505-151512505 on Build GRCh38
rs768355542 Chr.5: 151512657-151512657 on Build GRCh38
rs577692335 Chr.5: 151512866-151512866 on Build GRCh38
rs763565008 Chr.5: 151512526-151512526 on Build GRCh38
rs773459282 Chr.5: 151512934-151512934 on Build GRCh38
rs779603748 Chr.5: 151512865-151512865 on Build GRCh38
rs10476987 Chr.5: 151512928-151512928 on Build GRCh38
rs762295424 Chr.5: 151512537-151512537 on Build GRCh38
rs148932476 Chr.5: 151512594-151512594 on Build GRCh38
rs867387324 Chr.5: 151512582-151512582 on Build GRCh38
rs766794625 Chr.5: 151512565-151512565 on Build GRCh38
rs932573858 Chr.5: 151512575-151512575 on Build GRCh38
rs771727323 Chr.5: 151512631-151512631 on Build GRCh38
rs758635718 Chr.5: 151512559-151512559 on Build GRCh38
rs76034849 Chr.5: 151512839-151512839 on Build GRCh38
rs113205558 Chr.5: 151512497-151512497 on Build GRCh38
rs147044570 Chr.5: 151512829-151512829 on Build GRCh38
rs756222270 Chr.5: 151512614-151512614 on Build GRCh38
rs1054814328 Chr.5: 151512714-151512714 on Build GRCh38
rs1010993206 Chr.5: 151512825-151512825 on Build GRCh38
rs750948303 Chr.5: 151512558-151512558 on Build GRCh38
rs1015349204 Chr.5: 151512881-151512881 on Build GRCh38
rs962460594 Chr.5: 151512890-151512890 on Build GRCh38
rs770828078 Chr.5: 151512498-151512498 on Build GRCh38
rs745837864 Chr.5: 151512630-151512630 on Build GRCh38
rs913717225 Chr.5: 151512563-151512563 on Build GRCh38
rs201257346 Chr.5: 151512556-151512556 on Build GRCh38
rs569664435 Chr.5: 151512520-151512520 on Build GRCh38
rs937631616 Chr.5: 151512713-151512713 on Build GRCh38
rs755296412 Chr.5: 151512589-151512589 on Build GRCh38
rs1036815351 Chr.5: 151512830-151512830 on Build GRCh38
rs193123632 Chr.5: 151512924-151512924 on Build GRCh38
rs368333541 Chr.5: 151512861-151512861 on Build GRCh38
rs1019560516 Chr.5: 151512902-151512902 on Build GRCh38
rs749627752 Chr.5: 151512674-151512674 on Build GRCh38
rs781403525 Chr.5: 151512595-151512595 on Build GRCh38
rs534833542 Chr.5: 151512835-151512835 on Build GRCh38
rs925231118 Chr.5: 151512712-151512712 on Build GRCh38
rs779865392 Chr.5: 151512640-151512640 on Build GRCh38
rs867049412 Chr.5: 151512557-151512557 on Build GRCh38
rs143929309 Chr.5: 151512572-151512572 on Build GRCh38
rs896174677 Chr.5: 151512819-151512819 on Build GRCh38
rs551397930 Chr.5: 151512517-151512517 on Build GRCh38
rs1003917130 Chr.5: 151512867-151512867 on Build GRCh38
rs745581891 Chr.5: 151512511-151512511 on Build GRCh38
rs755067306 Chr.5: 151512487-151512487 on Build GRCh38
rs769051538 Chr.5: 151512842-151512842 on Build GRCh38
rs149409918 Chr.5: 151512519-151512519 on Build GRCh38
rs749212402 Chr.5: 151512491-151512491 on Build GRCh38
rs542812365 Chr.5: 151512536-151512536 on Build GRCh38
rs752891593 Chr.5: 151512609-151512609 on Build GRCh38
rs747251482 Chr.5: 151512486-151512486 on Build GRCh38

View Full Product Details