Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160253470-160253991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNJL
Entrez Gene ID
79616
Gene Name
cyclin J like
Gene Aliases
-
UniGene
Hs.14070
Gene Chromosome Location
Chr.5: 160251652-160339592 on Build GRCh38
Transcripts
NM_001308173.1 NM_024565.6 NR_131769.1 XM_006714917.3 XM_011534646.2 XM_011534647.2 XM_017009846.1 XM_017009847.1 XM_017009848.1 XM_017009849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623052 c.1006C>A p.L336I Chr.5: 160253680-160253680 on Build GRCh38
6623054 c.901G>A p.V301I Chr.5: 160253785-160253785 on Build GRCh38
136509 c.1186C>T p.P396S Chr.5: 160253500-160253500 on Build GRCh38
384674 c.999C>T p.Y333Y Chr.5: 160253687-160253687 on Build GRCh38
1435678 c.1123G>T p.G375C Chr.5: 160253563-160253563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968459583 Chr.5: 160253687-160253687 on Build GRCh38
rs764300195 Chr.5: 160253600-160253600 on Build GRCh38
rs779624773 Chr.5: 160253627-160253627 on Build GRCh38
rs753664479 Chr.5: 160253510-160253510 on Build GRCh38
rs556666531 Chr.5: 160253787-160253787 on Build GRCh38
rs370421409 Chr.5: 160253593-160253593 on Build GRCh38
rs199879462 Chr.5: 160253509-160253509 on Build GRCh38
rs778551885 Chr.5: 160253561-160253561 on Build GRCh38
rs925631458 Chr.5: 160253952-160253952 on Build GRCh38
rs960837107 Chr.5: 160253937-160253937 on Build GRCh38
rs765219491 Chr.5: 160253578-160253578 on Build GRCh38
rs200717032 Chr.5: 160253544-160253544 on Build GRCh38
rs774961375 Chr.5: 160253572-160253572 on Build GRCh38
rs74564582 Chr.5: 160253682-160253682 on Build GRCh38
rs755752513 Chr.5: 160253834-160253834 on Build GRCh38
rs757655932 Chr.5: 160253667-160253667 on Build GRCh38
rs375125722 Chr.5: 160253822-160253822 on Build GRCh38
rs778870108 Chr.5: 160253770-160253770 on Build GRCh38
rs779422643 Chr.5: 160253681-160253681 on Build GRCh38
rs200539258 Chr.5: 160253702-160253702 on Build GRCh38
rs373648473 Chr.5: 160253534-160253534 on Build GRCh38
rs777541658 Chr.5: 160253755-160253755 on Build GRCh38
rs759656265 Chr.5: 160253585-160253585 on Build GRCh38
rs754310300 Chr.5: 160253603-160253603 on Build GRCh38
rs760394370 Chr.5: 160253573-160253573 on Build GRCh38
rs545974239 Chr.5: 160253786-160253786 on Build GRCh38
rs145884784 Chr.5: 160253805-160253805 on Build GRCh38
rs779603501 Chr.5: 160253613-160253613 on Build GRCh38
rs113060117 Chr.5: 160253735-160253735 on Build GRCh38
rs377363287 Chr.5: 160253592-160253592 on Build GRCh38
rs375236790 Chr.5: 160253677-160253677 on Build GRCh38
rs377179668 Chr.5: 160253636-160253636 on Build GRCh38
rs199743869 Chr.5: 160253652-160253652 on Build GRCh38
rs773766538 Chr.5: 160253851-160253852 on Build GRCh38
rs760851064 Chr.5: 160253786-160253788 on Build GRCh38
rs371574226 Chr.5: 160253507-160253507 on Build GRCh38
rs939530328 Chr.5: 160253753-160253753 on Build GRCh38
rs751707240 Chr.5: 160253519-160253519 on Build GRCh38
rs769691898 Chr.5: 160253639-160253639 on Build GRCh38
rs751951751 Chr.5: 160253748-160253748 on Build GRCh38
rs934422783 Chr.5: 160253800-160253800 on Build GRCh38
rs374113107 Chr.5: 160253579-160253579 on Build GRCh38
rs746572826 Chr.5: 160253632-160253632 on Build GRCh38
rs755182751 Chr.5: 160253521-160253521 on Build GRCh38
rs1028218083 Chr.5: 160253858-160253858 on Build GRCh38
rs568430292 Chr.5: 160253728-160253728 on Build GRCh38
rs775331235 Chr.5: 160253506-160253506 on Build GRCh38
rs375241736 Chr.5: 160253609-160253609 on Build GRCh38
rs766689556 Chr.5: 160253812-160253812 on Build GRCh38
rs747988573 Chr.5: 160253638-160253638 on Build GRCh38
rs1009786255 Chr.5: 160253933-160253933 on Build GRCh38
rs760847342 Chr.5: 160253723-160253723 on Build GRCh38
rs765627148 Chr.5: 160253729-160253729 on Build GRCh38
rs563335624 Chr.5: 160253865-160253865 on Build GRCh38
rs781527268 Chr.5: 160253722-160253723 on Build GRCh38
rs980129629 Chr.5: 160253951-160253951 on Build GRCh38
rs750898193 Chr.5: 160253678-160253678 on Build GRCh38
rs942537903 Chr.5: 160253556-160253556 on Build GRCh38
rs549608310 Chr.5: 160253950-160253950 on Build GRCh38
rs541363127 Chr.5: 160253934-160253934 on Build GRCh38
rs149766851 Chr.5: 160253764-160253764 on Build GRCh38
rs771632303 Chr.5: 160253555-160253555 on Build GRCh38
rs759256343 Chr.5: 160253504-160253504 on Build GRCh38
rs1053447706 Chr.5: 160253722-160253722 on Build GRCh38
rs368571956 Chr.5: 160253684-160253684 on Build GRCh38
rs757688602 Chr.5: 160253723-160253723 on Build GRCh38
rs141268151 Chr.5: 160253543-160253543 on Build GRCh38
rs187903121 Chr.5: 160253574-160253574 on Build GRCh38
rs370674819 Chr.5: 160253666-160253666 on Build GRCh38
rs747381182 Chr.5: 160253568-160253578 on Build GRCh38
rs780706066 Chr.5: 160253742-160253742 on Build GRCh38
rs761737257 Chr.5: 160253577-160253577 on Build GRCh38
rs199835106 Chr.5: 160253503-160253503 on Build GRCh38
rs770516651 Chr.5: 160253492-160253492 on Build GRCh38
rs757014993 Chr.5: 160253901-160253901 on Build GRCh38
rs986191659 Chr.5: 160253720-160253720 on Build GRCh38
rs373108109 Chr.5: 160253564-160253564 on Build GRCh38
rs535480366 Chr.5: 160253715-160253715 on Build GRCh38
rs745814652 Chr.5: 160253771-160253771 on Build GRCh38
rs533035690 Chr.5: 160253614-160253614 on Build GRCh38
rs972808297 Chr.5: 160253914-160253914 on Build GRCh38
rs758945577 Chr.5: 160253679-160253679 on Build GRCh38
rs752290089 Chr.5: 160253743-160253743 on Build GRCh38
rs533202225 Chr.5: 160253947-160253947 on Build GRCh38
rs916775809 Chr.5: 160253945-160253945 on Build GRCh38
rs940157997 Chr.5: 160253518-160253518 on Build GRCh38
rs771724000 Chr.5: 160253568-160253568 on Build GRCh38
rs749114542 Chr.5: 160253713-160253713 on Build GRCh38
rs753076404 Chr.5: 160253536-160253536 on Build GRCh38
rs149011672 Chr.5: 160253836-160253836 on Build GRCh38
rs776850524 Chr.5: 160253785-160253785 on Build GRCh38
rs752273604 Chr.5: 160253831-160253831 on Build GRCh38
rs757793736 Chr.5: 160253624-160253624 on Build GRCh38
rs941027125 Chr.5: 160253809-160253809 on Build GRCh38
rs756373783 Chr.5: 160253597-160253597 on Build GRCh38
rs767440638 Chr.5: 160253659-160253659 on Build GRCh38
rs574039512 Chr.5: 160253765-160253765 on Build GRCh38
rs143801848 Chr.5: 160253938-160253938 on Build GRCh38
rs760050652 Chr.5: 160253820-160253820 on Build GRCh38
rs901110449 Chr.5: 160253565-160253565 on Build GRCh38
rs780867242 Chr.5: 160253685-160253685 on Build GRCh38
rs201558292 Chr.5: 160253750-160253750 on Build GRCh38
rs114822899 Chr.5: 160253902-160253902 on Build GRCh38
rs973268801 Chr.5: 160253738-160253738 on Build GRCh38
rs369592831 Chr.5: 160253512-160253512 on Build GRCh38
rs754474861 Chr.5: 160253741-160253741 on Build GRCh38
rs769531066 Chr.5: 160253706-160253706 on Build GRCh38
rs554607246 Chr.5: 160253791-160253791 on Build GRCh38
rs768794808 Chr.5: 160253779-160253779 on Build GRCh38
rs770305386 Chr.5: 160253851-160253854 on Build GRCh38
rs1016934604 Chr.5: 160253879-160253879 on Build GRCh38
rs939603948 Chr.5: 160253559-160253559 on Build GRCh38
rs753462505 Chr.5: 160253839-160253839 on Build GRCh38
rs763418490 Chr.5: 160253806-160253806 on Build GRCh38
rs773004663 Chr.5: 160253708-160253708 on Build GRCh38
rs1005907684 Chr.5: 160253878-160253878 on Build GRCh38
rs756394538 Chr.5: 160253537-160253537 on Build GRCh38
rs554916887 Chr.5: 160253727-160253727 on Build GRCh38
rs773096728 Chr.5: 160253645-160253645 on Build GRCh38
rs377751987 Chr.5: 160253814-160253814 on Build GRCh38
rs201691454 Chr.5: 160253734-160253734 on Build GRCh38

View Full Product Details