Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160280447-160280941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNJL
Entrez Gene ID
79616
Gene Name
cyclin J like
Gene Aliases
-
UniGene
Hs.14070
Gene Chromosome Location
Chr.5: 160251652-160339592 on Build GRCh38
Transcripts
NM_001308173.1 NM_024565.6 NR_131769.1 XM_006714917.3 XM_011534646.2 XM_017009846.1 XM_017009847.1 XM_017009848.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
127571 c.87C>T p.F29F Chr.5: 160280718-160280718 on Build GRCh38
5356527 c.218G>A p.R73H Chr.5: 160280587-160280587 on Build GRCh38
1496065 c.247T>A p.Y83N Chr.5: 160280558-160280558 on Build GRCh38
1435680 c.179G>A p.R60Q Chr.5: 160280626-160280626 on Build GRCh38
3853328 c.392C>T p.A131V Chr.5: 160280548-160280548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748396775 Chr.5: 160280690-160280690 on Build GRCh38
rs760994374 Chr.5: 160280494-160280494 on Build GRCh38
rs763372251 Chr.5: 160280572-160280572 on Build GRCh38
rs758438587 Chr.5: 160280502-160280502 on Build GRCh38
rs1036103176 Chr.5: 160280752-160280752 on Build GRCh38
rs78941682 Chr.5: 160280839-160280839 on Build GRCh38
rs960413113 Chr.5: 160280471-160280471 on Build GRCh38
rs752338883 Chr.5: 160280645-160280645 on Build GRCh38
rs758476138 Chr.5: 160280785-160280785 on Build GRCh38
rs748330346 Chr.5: 160280621-160280621 on Build GRCh38
rs745821860 Chr.5: 160280682-160280682 on Build GRCh38
rs762197142 Chr.5: 160280552-160280552 on Build GRCh38
rs373884688 Chr.5: 160280772-160280772 on Build GRCh38
rs760052950 Chr.5: 160280747-160280747 on Build GRCh38
rs906026584 Chr.5: 160280716-160280716 on Build GRCh38
rs145916838 Chr.5: 160280817-160280817 on Build GRCh38
rs184397934 Chr.5: 160280672-160280672 on Build GRCh38
rs757197305 Chr.5: 160280599-160280599 on Build GRCh38
rs372269963 Chr.5: 160280547-160280547 on Build GRCh38
rs760349678 Chr.5: 160280757-160280757 on Build GRCh38
rs369138047 Chr.5: 160280642-160280642 on Build GRCh38
rs749186098 Chr.5: 160280594-160280594 on Build GRCh38
rs750924245 Chr.5: 160280506-160280506 on Build GRCh38
rs775777510 Chr.5: 160280535-160280535 on Build GRCh38
rs757875700 Chr.5: 160280508-160280508 on Build GRCh38
rs751837920 Chr.5: 160280666-160280695 on Build GRCh38
rs538634248 Chr.5: 160280585-160280585 on Build GRCh38
rs774273324 Chr.5: 160280485-160280485 on Build GRCh38
rs775221000 Chr.5: 160280478-160280478 on Build GRCh38
rs987800964 Chr.5: 160280768-160280768 on Build GRCh38
rs190541591 Chr.5: 160280756-160280756 on Build GRCh38
rs968755286 Chr.5: 160280781-160280781 on Build GRCh38
rs540298937 Chr.5: 160280771-160280771 on Build GRCh38
rs772079141 Chr.5: 160280489-160280489 on Build GRCh38
rs373054799 Chr.5: 160280712-160280712 on Build GRCh38
rs755050977 Chr.5: 160280686-160280686 on Build GRCh38
rs910306118 Chr.5: 160280803-160280803 on Build GRCh38
rs755500612 Chr.5: 160280851-160280851 on Build GRCh38
rs759706153 Chr.5: 160280486-160280486 on Build GRCh38
rs754309994 Chr.5: 160280497-160280497 on Build GRCh38
rs920488694 Chr.5: 160280828-160280828 on Build GRCh38
rs769591969 Chr.5: 160280524-160280524 on Build GRCh38
rs775032139 Chr.5: 160280903-160280903 on Build GRCh38
rs549614911 Chr.5: 160280891-160280891 on Build GRCh38
rs954430476 Chr.5: 160280788-160280788 on Build GRCh38
rs752382255 Chr.5: 160280587-160280587 on Build GRCh38
rs374745062 Chr.5: 160280519-160280519 on Build GRCh38
rs367839311 Chr.5: 160280499-160280499 on Build GRCh38
rs374697453 Chr.5: 160280476-160280476 on Build GRCh38
rs368348160 Chr.5: 160280579-160280579 on Build GRCh38
rs753554517 Chr.5: 160280767-160280767 on Build GRCh38
rs775808580 Chr.5: 160280492-160280492 on Build GRCh38
rs374868101 Chr.5: 160280678-160280678 on Build GRCh38
rs765870739 Chr.5: 160280577-160280577 on Build GRCh38
rs950120929 Chr.5: 160280867-160280867 on Build GRCh38
rs561110535 Chr.5: 160280892-160280892 on Build GRCh38
rs201843807 Chr.5: 160280581-160280581 on Build GRCh38
rs764546553 Chr.5: 160280496-160280496 on Build GRCh38
rs780953085 Chr.5: 160280518-160280518 on Build GRCh38
rs76732875 Chr.5: 160280724-160280724 on Build GRCh38
rs766921931 Chr.5: 160280627-160280627 on Build GRCh38
rs780256615 Chr.5: 160280600-160280600 on Build GRCh38
rs899095117 Chr.5: 160280613-160280613 on Build GRCh38
rs751148381 Chr.5: 160280513-160280513 on Build GRCh38
rs368002668 Chr.5: 160280546-160280546 on Build GRCh38
rs752771342 Chr.5: 160280794-160280794 on Build GRCh38
rs562548157 Chr.5: 160280838-160280838 on Build GRCh38
rs979252487 Chr.5: 160280796-160280796 on Build GRCh38
rs773517273 Chr.5: 160280626-160280626 on Build GRCh38
rs771316396 Chr.5: 160280728-160280728 on Build GRCh38
rs895799079 Chr.5: 160280866-160280866 on Build GRCh38
rs759010486 Chr.5: 160280641-160280641 on Build GRCh38
rs182372982 Chr.5: 160280910-160280910 on Build GRCh38
rs185171463 Chr.5: 160280911-160280911 on Build GRCh38
rs757225101 Chr.5: 160280674-160280674 on Build GRCh38
rs73817375 Chr.5: 160280751-160280751 on Build GRCh38
rs777728909 Chr.5: 160280526-160280526 on Build GRCh38
rs369287405 Chr.5: 160280687-160280687 on Build GRCh38
rs774794553 Chr.5: 160280742-160280742 on Build GRCh38
rs140468741 Chr.5: 160280616-160280616 on Build GRCh38
rs773605574 Chr.5: 160280717-160280717 on Build GRCh38
rs781320174 Chr.5: 160280793-160280793 on Build GRCh38
rs1038909624 Chr.5: 160280824-160280824 on Build GRCh38
rs114120913 Chr.5: 160280498-160280498 on Build GRCh38
rs536303839 Chr.5: 160280509-160280509 on Build GRCh38
rs760654439 Chr.5: 160280488-160280488 on Build GRCh38
rs200208965 Chr.5: 160280477-160280477 on Build GRCh38
rs777446144 Chr.5: 160280588-160280588 on Build GRCh38
rs560313062 Chr.5: 160280805-160280805 on Build GRCh38
rs377115435 Chr.5: 160280553-160280553 on Build GRCh38
rs764077101 Chr.5: 160280602-160280603 on Build GRCh38
rs146670939 Chr.5: 160280468-160280468 on Build GRCh38
rs755729272 Chr.5: 160280647-160280647 on Build GRCh38
rs765517663 Chr.5: 160280670-160280670 on Build GRCh38
rs993463749 Chr.5: 160280467-160280467 on Build GRCh38
rs777317480 Chr.5: 160280881-160280881 on Build GRCh38
rs546703146 Chr.5: 160280534-160280534 on Build GRCh38
rs770127604 Chr.5: 160280624-160280624 on Build GRCh38
rs773820498 Chr.5: 160280637-160280637 on Build GRCh38
rs545469809 Chr.5: 160280819-160280819 on Build GRCh38
rs749209105 Chr.5: 160280532-160280532 on Build GRCh38
rs747122783 Chr.5: 160280615-160280615 on Build GRCh38
rs777818404 Chr.5: 160280809-160280809 on Build GRCh38
rs925110614 Chr.5: 160280798-160280798 on Build GRCh38
rs146011937 Chr.5: 160280797-160280798 on Build GRCh38
rs754693446 Chr.5: 160280517-160280517 on Build GRCh38
rs763691965 Chr.5: 160280758-160280758 on Build GRCh38
rs750882960 Chr.5: 160280571-160280571 on Build GRCh38

View Full Product Details