Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 161685939-161686434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA6
Entrez Gene ID
2559
Gene Name
gamma-aminobutyric acid type A receptor alpha6 subunit
Gene Aliases
-
UniGene
Hs.90791
Gene Chromosome Location
Chr.5: 161685652-161702592 on Build GRCh38
Transcripts
NM_000811.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1065301 c.4G>A p.A2T Chr.5: 161685993-161685993 on Build GRCh38
269216 c.49G>A p.A17T Chr.5: 161686240-161686240 on Build GRCh38
5934983 c.155G>A p.G52E Chr.5: 161686346-161686346 on Build GRCh38
3613548 c.136C>T p.R46W Chr.5: 161686327-161686327 on Build GRCh38
3827611 c.111C>T p.D37D Chr.5: 161686302-161686302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377669113 Chr.5: 161686255-161686255 on Build GRCh38
rs905119195 Chr.5: 161686141-161686141 on Build GRCh38
rs200373173 Chr.5: 161686285-161686285 on Build GRCh38
rs144795512 Chr.5: 161686342-161686342 on Build GRCh38
rs781685109 Chr.5: 161686361-161686361 on Build GRCh38
rs914745007 Chr.5: 161686197-161686197 on Build GRCh38
rs983522820 Chr.5: 161686337-161686337 on Build GRCh38
rs999163444 Chr.5: 161686088-161686088 on Build GRCh38
rs370888800 Chr.5: 161686284-161686284 on Build GRCh38
rs148791978 Chr.5: 161686133-161686133 on Build GRCh38
rs996020649 Chr.5: 161686001-161686001 on Build GRCh38
rs757447117 Chr.5: 161686328-161686328 on Build GRCh38
rs746198834 Chr.5: 161686250-161686250 on Build GRCh38
rs549588611 Chr.5: 161686137-161686137 on Build GRCh38
rs747533030 Chr.5: 161686267-161686267 on Build GRCh38
rs768135154 Chr.5: 161686221-161686221 on Build GRCh38
rs373363000 Chr.5: 161686327-161686327 on Build GRCh38
rs773960748 Chr.5: 161686394-161686394 on Build GRCh38
rs757498238 Chr.5: 161686239-161686239 on Build GRCh38
rs989670780 Chr.5: 161686171-161686171 on Build GRCh38
rs754101422 Chr.5: 161686234-161686234 on Build GRCh38
rs373712601 Chr.5: 161686051-161686051 on Build GRCh38
rs760111344 Chr.5: 161685969-161685969 on Build GRCh38
rs1002505258 Chr.5: 161686184-161686184 on Build GRCh38
rs747606411 Chr.5: 161686364-161686364 on Build GRCh38
rs761508240 Chr.5: 161686075-161686075 on Build GRCh38
rs112717732 Chr.5: 161686251-161686251 on Build GRCh38
rs779283943 Chr.5: 161686245-161686245 on Build GRCh38
rs750343780 Chr.5: 161685995-161685995 on Build GRCh38
rs140035570 Chr.5: 161686229-161686229 on Build GRCh38
rs866538329 Chr.5: 161685963-161685963 on Build GRCh38
rs772922854 Chr.5: 161686182-161686182 on Build GRCh38
rs755371389 Chr.5: 161686354-161686354 on Build GRCh38
rs59181402 Chr.5: 161686397-161686397 on Build GRCh38
rs762766703 Chr.5: 161686194-161686194 on Build GRCh38
rs766289625 Chr.5: 161686215-161686215 on Build GRCh38
rs764507806 Chr.5: 161686368-161686368 on Build GRCh38
rs763766120 Chr.5: 161685970-161685970 on Build GRCh38
rs756187679 Chr.5: 161686225-161686225 on Build GRCh38
rs372211814 Chr.5: 161686379-161686379 on Build GRCh38
rs771866728 Chr.5: 161686398-161686398 on Build GRCh38
rs187766196 Chr.5: 161686338-161686338 on Build GRCh38
rs775009633 Chr.5: 161685967-161685967 on Build GRCh38
rs776338909 Chr.5: 161686056-161686056 on Build GRCh38
rs758271057 Chr.5: 161685997-161685997 on Build GRCh38
rs769394469 Chr.5: 161686291-161686291 on Build GRCh38
rs999425239 Chr.5: 161686389-161686389 on Build GRCh38
rs954386810 Chr.5: 161686261-161686261 on Build GRCh38
rs770747148 Chr.5: 161686380-161686380 on Build GRCh38
rs766834585 Chr.5: 161686385-161686386 on Build GRCh38
rs749514674 Chr.5: 161686023-161686023 on Build GRCh38
rs943997003 Chr.5: 161686248-161686248 on Build GRCh38
rs202174476 Chr.5: 161686054-161686054 on Build GRCh38
rs371494767 Chr.5: 161686095-161686095 on Build GRCh38
rs556539275 Chr.5: 161686323-161686323 on Build GRCh38
rs774275315 Chr.5: 161686093-161686093 on Build GRCh38
rs895010288 Chr.5: 161686049-161686049 on Build GRCh38
rs374513435 Chr.5: 161686373-161686373 on Build GRCh38
rs771697716 Chr.5: 161685962-161685962 on Build GRCh38
rs764881079 Chr.5: 161686181-161686181 on Build GRCh38
rs13188991 Chr.5: 161685998-161685998 on Build GRCh38
rs370922712 Chr.5: 161686004-161686004 on Build GRCh38
rs772644589 Chr.5: 161686039-161686039 on Build GRCh38
rs367675471 Chr.5: 161686377-161686377 on Build GRCh38
rs766905192 Chr.5: 161686292-161686292 on Build GRCh38
rs368424653 Chr.5: 161686316-161686316 on Build GRCh38
rs779480814 Chr.5: 161686033-161686033 on Build GRCh38
rs760188021 Chr.5: 161686053-161686053 on Build GRCh38
rs746250382 Chr.5: 161686034-161686034 on Build GRCh38
rs572588976 Chr.5: 161686346-161686346 on Build GRCh38
rs751098162 Chr.5: 161686334-161686334 on Build GRCh38
rs1008555446 Chr.5: 161686280-161686280 on Build GRCh38
rs562907722 Chr.5: 161685988-161685988 on Build GRCh38
rs759451237 Chr.5: 161686325-161686325 on Build GRCh38
rs947903028 Chr.5: 161686349-161686349 on Build GRCh38
rs761561088 Chr.5: 161685994-161685994 on Build GRCh38
rs762713704 Chr.5: 161686301-161686301 on Build GRCh38
rs1047795083 Chr.5: 161686089-161686089 on Build GRCh38
rs778034380 Chr.5: 161686006-161686006 on Build GRCh38

View Full Product Details