Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178978192-178978699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRM6
Entrez Gene ID
2916
Gene Name
glutamate metabotropic receptor 6
Gene Aliases
CSNB1B, GPRC1F, MGLUR6, mGlu6
UniGene
Hs.248131
Gene Chromosome Location
Chr.5: 178978327-178995123 on Build GRCh38
Transcripts
NM_000843.3

SNP Detail

SNP ID Chromosome Location
rs557926279 Chr.5: 178978547-178978547 on Build GRCh38
rs953671176 Chr.5: 178978613-178978613 on Build GRCh38
rs946124701 Chr.5: 178978220-178978220 on Build GRCh38
rs539994152 Chr.5: 178978531-178978531 on Build GRCh38
rs764285993 Chr.5: 178978538-178978538 on Build GRCh38
rs754789483 Chr.5: 178978576-178978576 on Build GRCh38
rs759574528 Chr.5: 178978340-178978340 on Build GRCh38
rs1028386379 Chr.5: 178978606-178978606 on Build GRCh38
rs1030721509 Chr.5: 178978416-178978416 on Build GRCh38
rs4700765 Chr.5: 178978250-178978250 on Build GRCh38
rs760870186 Chr.5: 178978499-178978499 on Build GRCh38
rs113856620 Chr.5: 178978307-178978307 on Build GRCh38
rs527845857 Chr.5: 178978523-178978523 on Build GRCh38
rs192684059 Chr.5: 178978398-178978398 on Build GRCh38
rs949984136 Chr.5: 178978321-178978321 on Build GRCh38
rs898245671 Chr.5: 178978519-178978519 on Build GRCh38
rs78242013 Chr.5: 178978420-178978420 on Build GRCh38
rs1056190380 Chr.5: 178978308-178978308 on Build GRCh38
rs894415689 Chr.5: 178978310-178978310 on Build GRCh38
rs967498687 Chr.5: 178978668-178978668 on Build GRCh38
rs778638375 Chr.5: 178978673-178978673 on Build GRCh38
rs577277103 Chr.5: 178978484-178978484 on Build GRCh38
rs542539788 Chr.5: 178978368-178978368 on Build GRCh38
rs750005391 Chr.5: 178978231-178978231 on Build GRCh38
rs1016680684 Chr.5: 178978505-178978505 on Build GRCh38
rs576264179 Chr.5: 178978652-178978652 on Build GRCh38
rs559624691 Chr.5: 178978412-178978416 on Build GRCh38
rs551558381 Chr.5: 178978285-178978285 on Build GRCh38
rs967703291 Chr.5: 178978403-178978403 on Build GRCh38
rs925945660 Chr.5: 178978290-178978290 on Build GRCh38
rs549290725 Chr.5: 178978347-178978347 on Build GRCh38
rs1047117887 Chr.5: 178978274-178978274 on Build GRCh38
rs552708683 Chr.5: 178978439-178978439 on Build GRCh38
rs537336447 Chr.5: 178978309-178978309 on Build GRCh38
rs974311930 Chr.5: 178978586-178978586 on Build GRCh38
rs886282965 Chr.5: 178978278-178978278 on Build GRCh38
rs977583974 Chr.5: 178978670-178978670 on Build GRCh38
rs988097618 Chr.5: 178978640-178978640 on Build GRCh38
rs1003922543 Chr.5: 178978467-178978467 on Build GRCh38

View Full Product Details