Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178981749-178982258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRM6
Entrez Gene ID
2916
Gene Name
glutamate metabotropic receptor 6
Gene Aliases
CSNB1B, GPRC1F, MGLUR6, mGlu6
UniGene
Hs.248131
Gene Chromosome Location
Chr.5: 178978327-178995123 on Build GRCh38
Transcripts
NM_000843.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6487915 c.2523C>A p.Y841* Chr.5: 178981768-178981768 on Build GRCh38
4419378 c.2484C>T p.A828A Chr.5: 178981807-178981807 on Build GRCh38
1253829 c.2510T>A p.V837E Chr.5: 178981781-178981781 on Build GRCh38
4003466 c.2437-6G>A p.? Chr.5: 178981860-178981860 on Build GRCh38
4453032 c.2437A>G p.I813V Chr.5: 178981854-178981854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984692161 Chr.5: 178982090-178982090 on Build GRCh38
rs746263697 Chr.5: 178981863-178981863 on Build GRCh38
rs779172935 Chr.5: 178981784-178981784 on Build GRCh38
rs184219010 Chr.5: 178982110-178982110 on Build GRCh38
rs933033215 Chr.5: 178982127-178982127 on Build GRCh38
rs372122761 Chr.5: 178981891-178981891 on Build GRCh38
rs762952668 Chr.5: 178981811-178981811 on Build GRCh38
rs202157072 Chr.5: 178981804-178981804 on Build GRCh38
rs1034169915 Chr.5: 178982016-178982016 on Build GRCh38
rs2071249 Chr.5: 178981834-178981834 on Build GRCh38
rs1037843562 Chr.5: 178982208-178982208 on Build GRCh38
rs576201208 Chr.5: 178982102-178982102 on Build GRCh38
rs1041409829 Chr.5: 178981894-178981894 on Build GRCh38
rs923045815 Chr.5: 178982123-178982123 on Build GRCh38
rs747793174 Chr.5: 178982144-178982144 on Build GRCh38
rs942246425 Chr.5: 178982199-178982199 on Build GRCh38
rs17078857 Chr.5: 178981842-178981842 on Build GRCh38
rs751758654 Chr.5: 178981802-178981802 on Build GRCh38
rs756676915 Chr.5: 178981885-178981885 on Build GRCh38
rs904093016 Chr.5: 178982198-178982198 on Build GRCh38
rs558406753 Chr.5: 178982010-178982010 on Build GRCh38
rs1013566198 Chr.5: 178982020-178982020 on Build GRCh38
rs778450628 Chr.5: 178982141-178982141 on Build GRCh38
rs191922065 Chr.5: 178982103-178982103 on Build GRCh38
rs527990284 Chr.5: 178982185-178982185 on Build GRCh38
rs754475767 Chr.5: 178981828-178981828 on Build GRCh38
rs945620310 Chr.5: 178982160-178982160 on Build GRCh38
rs993729370 Chr.5: 178982226-178982226 on Build GRCh38
rs560377837 Chr.5: 178982133-178982133 on Build GRCh38
rs1044291445 Chr.5: 178981956-178981956 on Build GRCh38
rs993677108 Chr.5: 178981990-178981990 on Build GRCh38
rs780609247 Chr.5: 178981916-178981916 on Build GRCh38
rs751097143 Chr.5: 178981827-178981827 on Build GRCh38
rs1052752901 Chr.5: 178981908-178981908 on Build GRCh38
rs1044020063 Chr.5: 178982166-178982166 on Build GRCh38
rs754672046 Chr.5: 178981779-178981779 on Build GRCh38
rs938219931 Chr.5: 178982205-178982205 on Build GRCh38
rs758906054 Chr.5: 178981872-178981872 on Build GRCh38
rs557977290 Chr.5: 178982064-178982064 on Build GRCh38
rs61731185 Chr.5: 178981782-178981782 on Build GRCh38
rs912866770 Chr.5: 178982150-178982150 on Build GRCh38
rs188372378 Chr.5: 178982212-178982212 on Build GRCh38
rs116585300 Chr.5: 178982109-178982109 on Build GRCh38
rs562563029 Chr.5: 178982227-178982227 on Build GRCh38
rs777098868 Chr.5: 178981795-178981795 on Build GRCh38
rs545781361 Chr.5: 178981964-178981964 on Build GRCh38
rs768658323 Chr.5: 178981890-178981890 on Build GRCh38
rs766952342 Chr.5: 178981985-178981985 on Build GRCh38
rs557789793 Chr.5: 178982009-178982009 on Build GRCh38
rs958590893 Chr.5: 178982050-178982050 on Build GRCh38
rs746890644 Chr.5: 178981904-178981904 on Build GRCh38
rs898073982 Chr.5: 178981965-178981965 on Build GRCh38
rs114970739 Chr.5: 178981860-178981860 on Build GRCh38
rs373757775 Chr.5: 178981794-178981794 on Build GRCh38
rs771257943 Chr.5: 178981783-178981783 on Build GRCh38
rs571315792 Chr.5: 178982140-178982140 on Build GRCh38
rs1055205142 Chr.5: 178982209-178982209 on Build GRCh38
rs150784083 Chr.5: 178981954-178981954 on Build GRCh38
rs1002779658 Chr.5: 178982001-178982001 on Build GRCh38
rs896559603 Chr.5: 178982012-178982012 on Build GRCh38
rs115297481 Chr.5: 178981907-178981907 on Build GRCh38
rs780597433 Chr.5: 178981873-178981873 on Build GRCh38
rs746340451 Chr.5: 178981789-178981789 on Build GRCh38
rs945619424 Chr.5: 178981955-178981955 on Build GRCh38
rs747528585 Chr.5: 178981875-178981875 on Build GRCh38
rs1050207852 Chr.5: 178982152-178982152 on Build GRCh38
rs865820370 Chr.5: 178981805-178981805 on Build GRCh38
rs562923528 Chr.5: 178982108-178982108 on Build GRCh38
rs1026252513 Chr.5: 178982034-178982034 on Build GRCh38
rs145930962 Chr.5: 178981806-178981806 on Build GRCh38
rs906565024 Chr.5: 178982230-178982230 on Build GRCh38
rs768852907 Chr.5: 178981798-178981798 on Build GRCh38
rs980894877 Chr.5: 178982011-178982011 on Build GRCh38
rs889318283 Chr.5: 178981914-178981914 on Build GRCh38
rs766938162 Chr.5: 178981829-178981829 on Build GRCh38
rs755747636 Chr.5: 178981835-178981835 on Build GRCh38
rs766490379 Chr.5: 178981819-178981819 on Build GRCh38

View Full Product Details