Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179127741-179128194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS2
Entrez Gene ID
9509
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 2
Gene Aliases
ADAM-TS2, ADAMTS-2, ADAMTS-3, NPI, PC I-NP, PCI-NP, PCINP, PCPNI, PNPI
UniGene
Hs.23871
Gene Chromosome Location
Chr.5: 179110851-179345430 on Build GRCh38
Transcripts
NM_014244.4 XM_017010063.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3735276 c.2617G>T p.G873W Chr.5: 179127959-179127959 on Build GRCh38
5908469 c.2569G>A p.E857K Chr.5: 179128007-179128007 on Build GRCh38
6170929 c.2617G>A p.G873R Chr.5: 179127959-179127959 on Build GRCh38
301017 c.2508C>A p.I836I Chr.5: 179128068-179128068 on Build GRCh38
5893023 c.2615G>A p.G872E Chr.5: 179127961-179127961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753127866 Chr.5: 179127945-179127945 on Build GRCh38
rs531522458 Chr.5: 179127816-179127816 on Build GRCh38
rs990127201 Chr.5: 179127921-179127921 on Build GRCh38
rs990350223 Chr.5: 179127780-179127780 on Build GRCh38
rs867353605 Chr.5: 179128106-179128106 on Build GRCh38
rs375502905 Chr.5: 179127920-179127920 on Build GRCh38
rs376995938 Chr.5: 179128097-179128097 on Build GRCh38
rs1011419172 Chr.5: 179128070-179128070 on Build GRCh38
rs750543911 Chr.5: 179128086-179128086 on Build GRCh38
rs761360859 Chr.5: 179127919-179127919 on Build GRCh38
rs978000210 Chr.5: 179128173-179128173 on Build GRCh38
rs753947828 Chr.5: 179127953-179127953 on Build GRCh38
rs142355768 Chr.5: 179127981-179127981 on Build GRCh38
rs547128868 Chr.5: 179127944-179127944 on Build GRCh38
rs943437047 Chr.5: 179127950-179127950 on Build GRCh38
rs867087162 Chr.5: 179127986-179127986 on Build GRCh38
rs545682014 Chr.5: 179127802-179127802 on Build GRCh38
rs1006572919 Chr.5: 179127817-179127817 on Build GRCh38
rs1057517277 Chr.5: 179128118-179128124 on Build GRCh38
rs776605625 Chr.5: 179127984-179127984 on Build GRCh38
rs373287507 Chr.5: 179128109-179128109 on Build GRCh38
rs1011496700 Chr.5: 179128152-179128152 on Build GRCh38
rs756651045 Chr.5: 179127946-179127946 on Build GRCh38
rs933767735 Chr.5: 179127788-179127788 on Build GRCh38
rs754467396 Chr.5: 179128047-179128047 on Build GRCh38
rs7705437 Chr.5: 179128084-179128084 on Build GRCh38
rs894562597 Chr.5: 179128149-179128149 on Build GRCh38
rs141293393 Chr.5: 179127807-179127807 on Build GRCh38
rs779277427 Chr.5: 179127962-179127962 on Build GRCh38
rs745450151 Chr.5: 179127949-179127950 on Build GRCh38
rs762463826 Chr.5: 179128019-179128019 on Build GRCh38
rs753157368 Chr.5: 179127940-179127940 on Build GRCh38
rs919478485 Chr.5: 179127893-179127893 on Build GRCh38
rs758515627 Chr.5: 179128088-179128088 on Build GRCh38
rs762670979 Chr.5: 179127937-179127937 on Build GRCh38
rs753997330 Chr.5: 179127851-179127851 on Build GRCh38
rs764687535 Chr.5: 179128046-179128046 on Build GRCh38
rs145016043 Chr.5: 179127963-179127963 on Build GRCh38
rs2303644 Chr.5: 179128044-179128044 on Build GRCh38
rs878880727 Chr.5: 179127861-179127861 on Build GRCh38
rs766088822 Chr.5: 179128034-179128034 on Build GRCh38
rs779695756 Chr.5: 179128041-179128043 on Build GRCh38
rs1035391110 Chr.5: 179128133-179128133 on Build GRCh38
rs201592583 Chr.5: 179128089-179128089 on Build GRCh38
rs971503610 Chr.5: 179127887-179127887 on Build GRCh38
rs928265597 Chr.5: 179127843-179127843 on Build GRCh38
rs76488852 Chr.5: 179128043-179128043 on Build GRCh38
rs1021612968 Chr.5: 179128154-179128154 on Build GRCh38
rs35445112 Chr.5: 179128096-179128096 on Build GRCh38
rs748892203 Chr.5: 179128128-179128128 on Build GRCh38
rs757848152 Chr.5: 179128080-179128080 on Build GRCh38
rs138983307 Chr.5: 179128069-179128069 on Build GRCh38
rs772792600 Chr.5: 179128017-179128017 on Build GRCh38
rs368959910 Chr.5: 179128136-179128136 on Build GRCh38
rs372549560 Chr.5: 179127982-179127982 on Build GRCh38
rs372242497 Chr.5: 179127977-179127977 on Build GRCh38
rs377085746 Chr.5: 179128110-179128110 on Build GRCh38
rs73344606 Chr.5: 179127831-179127831 on Build GRCh38
rs367706210 Chr.5: 179127932-179127932 on Build GRCh38
rs528937903 Chr.5: 179127911-179127911 on Build GRCh38
rs1003393247 Chr.5: 179127892-179127892 on Build GRCh38
rs750980406 Chr.5: 179127910-179127910 on Build GRCh38
rs755513065 Chr.5: 179128034-179128035 on Build GRCh38
rs762297315 Chr.5: 179127805-179127805 on Build GRCh38
rs766947359 Chr.5: 179127983-179127983 on Build GRCh38
rs182103023 Chr.5: 179128107-179128107 on Build GRCh38
rs1038610461 Chr.5: 179127823-179127823 on Build GRCh38
rs769226679 Chr.5: 179128008-179128008 on Build GRCh38
rs747653130 Chr.5: 179128007-179128007 on Build GRCh38
rs777107579 Chr.5: 179128111-179128111 on Build GRCh38
rs373433328 Chr.5: 179128169-179128169 on Build GRCh38
rs1034341135 Chr.5: 179127900-179127900 on Build GRCh38
rs74989438 Chr.5: 179127763-179127763 on Build GRCh38
rs78784112 Chr.5: 179128161-179128161 on Build GRCh38
rs189809035 Chr.5: 179127863-179127863 on Build GRCh38

View Full Product Details