Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150265715-150266222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APH1A
Entrez Gene ID
51107
Gene Name
aph-1 homolog A, gamma-secretase subunit
Gene Aliases
6530402N02Rik, APH-1, APH-1A, CGI-78
UniGene
Hs.108408
Gene Chromosome Location
Chr.1: 150265399-150282291 on Build GRCh38
Transcripts
NM_001077628.2 NM_001243771.1 NM_001243772.1 NM_016022.3 NR_045033.1 NR_045034.1 NR_045035.1 XM_017001417.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6051959 c.768T>C p.Y256Y Chr.1: 150266160-150266160 on Build GRCh38
6494261 c.735C>G p.C245W Chr.1: 150266193-150266193 on Build GRCh38
3976100 c.772G>A p.A258T Chr.1: 150266156-150266156 on Build GRCh38
2181837 c.739C>T p.R247W Chr.1: 150266189-150266189 on Build GRCh38
3473347 c.785C>T p.P262L Chr.1: 150266143-150266143 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs184760849 Chr.1: 150265761-150265761 on Build GRCh38
rs782714941 Chr.1: 150266119-150266119 on Build GRCh38
rs201035904 Chr.1: 150265957-150265957 on Build GRCh38
rs200654934 Chr.1: 150265839-150265839 on Build GRCh38
rs199961462 Chr.1: 150265790-150265790 on Build GRCh38
rs199926282 Chr.1: 150265879-150265879 on Build GRCh38
rs370719475 Chr.1: 150266150-150266150 on Build GRCh38
rs967793541 Chr.1: 150266076-150266076 on Build GRCh38
rs201818552 Chr.1: 150265849-150265849 on Build GRCh38
rs782398924 Chr.1: 150265846-150265846 on Build GRCh38
rs201754927 Chr.1: 150265873-150265873 on Build GRCh38
rs781990836 Chr.1: 150266099-150266099 on Build GRCh38
rs1038170929 Chr.1: 150265943-150265943 on Build GRCh38
rs200432613 Chr.1: 150265956-150265956 on Build GRCh38
rs587650711 Chr.1: 150266065-150266065 on Build GRCh38
rs74126634 Chr.1: 150266193-150266193 on Build GRCh38
rs200528248 Chr.1: 150266027-150266027 on Build GRCh38
rs202091565 Chr.1: 150265980-150265980 on Build GRCh38
rs587750560 Chr.1: 150265910-150265910 on Build GRCh38
rs782434457 Chr.1: 150266138-150266138 on Build GRCh38
rs964294762 Chr.1: 150265811-150265811 on Build GRCh38
rs369757504 Chr.1: 150266111-150266111 on Build GRCh38
rs782155978 Chr.1: 150266087-150266087 on Build GRCh38
rs71622685 Chr.1: 150266173-150266173 on Build GRCh38
rs1021139844 Chr.1: 150266066-150266066 on Build GRCh38
rs782780478 Chr.1: 150265905-150265905 on Build GRCh38
rs781834956 Chr.1: 150266123-150266123 on Build GRCh38
rs782123265 Chr.1: 150266110-150266110 on Build GRCh38
rs908397072 Chr.1: 150265867-150265867 on Build GRCh38
rs200779875 Chr.1: 150265766-150265766 on Build GRCh38
rs1033143052 Chr.1: 150266095-150266095 on Build GRCh38
rs587653801 Chr.1: 150265749-150265749 on Build GRCh38
rs376130182 Chr.1: 150266139-150266139 on Build GRCh38
rs1009295416 Chr.1: 150266051-150266051 on Build GRCh38
rs782641325 Chr.1: 150266188-150266188 on Build GRCh38
rs201553051 Chr.1: 150266171-150266171 on Build GRCh38
rs374950774 Chr.1: 150265927-150265927 on Build GRCh38
rs145520277 Chr.1: 150265760-150265760 on Build GRCh38
rs201261520 Chr.1: 150265853-150265853 on Build GRCh38
rs201855910 Chr.1: 150265882-150265882 on Build GRCh38
rs964686469 Chr.1: 150266113-150266113 on Build GRCh38
rs201411127 Chr.1: 150266080-150266080 on Build GRCh38
rs200240863 Chr.1: 150266115-150266115 on Build GRCh38
rs373144924 Chr.1: 150266118-150266118 on Build GRCh38
rs781939905 Chr.1: 150266081-150266081 on Build GRCh38
rs1056088933 Chr.1: 150265988-150265988 on Build GRCh38
rs934098020 Chr.1: 150265850-150265850 on Build GRCh38
rs199646767 Chr.1: 150266090-150266090 on Build GRCh38
rs201826227 Chr.1: 150265781-150265781 on Build GRCh38
rs201013519 Chr.1: 150265880-150265880 on Build GRCh38
rs975415056 Chr.1: 150265813-150265813 on Build GRCh38
rs189623935 Chr.1: 150265851-150265851 on Build GRCh38
rs1002071391 Chr.1: 150265979-150265979 on Build GRCh38
rs782355078 Chr.1: 150266192-150266192 on Build GRCh38

View Full Product Details