Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150267867-150268359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APH1A
Entrez Gene ID
51107
Gene Name
aph-1 homolog A, gamma-secretase subunit
Gene Aliases
6530402N02Rik, APH-1, APH-1A, CGI-78
UniGene
Hs.108408
Gene Chromosome Location
Chr.1: 150265399-150282291 on Build GRCh38
Transcripts
NM_001077628.2 NM_001243771.1 NM_001243772.1 NM_016022.3 NR_045033.1 NR_045034.1 NR_045035.1 XM_017001417.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6584101 c.149C>G p.S50C Chr.1: 150268092-150268092 on Build GRCh38
73695 c.236C>G p.S79C Chr.1: 150268005-150268005 on Build GRCh38
4142584 c.267C>A p.A89A Chr.1: 150267974-150267974 on Build GRCh38
4142583 c.267C>A p.A89A Chr.1: 150267974-150267974 on Build GRCh38
95763 c.156C>T p.V52V Chr.1: 150268085-150268085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782467630 Chr.1: 150267951-150267951 on Build GRCh38
rs782546378 Chr.1: 150267983-150267983 on Build GRCh38
rs931610213 Chr.1: 150268156-150268156 on Build GRCh38
rs370361277 Chr.1: 150267972-150267972 on Build GRCh38
rs199775906 Chr.1: 150268090-150268090 on Build GRCh38
rs1132746 Chr.1: 150268105-150268105 on Build GRCh38
rs895379864 Chr.1: 150268182-150268182 on Build GRCh38
rs782154484 Chr.1: 150267938-150267938 on Build GRCh38
rs200212776 Chr.1: 150267945-150267945 on Build GRCh38
rs958012352 Chr.1: 150267995-150267995 on Build GRCh38
rs201613749 Chr.1: 150267989-150267989 on Build GRCh38
rs201789562 Chr.1: 150267908-150267908 on Build GRCh38
rs758736013 Chr.1: 150268195-150268195 on Build GRCh38
rs781839898 Chr.1: 150268094-150268094 on Build GRCh38
rs587724165 Chr.1: 150267949-150267949 on Build GRCh38
rs920080806 Chr.1: 150268112-150268112 on Build GRCh38
rs180846860 Chr.1: 150268324-150268324 on Build GRCh38
rs199530341 Chr.1: 150268165-150268165 on Build GRCh38
rs782319679 Chr.1: 150268045-150268045 on Build GRCh38
rs1032202408 Chr.1: 150268283-150268283 on Build GRCh38
rs377210432 Chr.1: 150267935-150267935 on Build GRCh38
rs778308427 Chr.1: 150268309-150268309 on Build GRCh38
rs587614048 Chr.1: 150268291-150268291 on Build GRCh38
rs987105227 Chr.1: 150268043-150268043 on Build GRCh38
rs1046349418 Chr.1: 150268249-150268249 on Build GRCh38
rs1000017680 Chr.1: 150268281-150268281 on Build GRCh38
rs781881118 Chr.1: 150267921-150267921 on Build GRCh38
rs782617910 Chr.1: 150267965-150267965 on Build GRCh38
rs782715592 Chr.1: 150268139-150268139 on Build GRCh38
rs200730753 Chr.1: 150268099-150268099 on Build GRCh38
rs782301701 Chr.1: 150267974-150267974 on Build GRCh38
rs201522669 Chr.1: 150268153-150268153 on Build GRCh38
rs782361743 Chr.1: 150268228-150268229 on Build GRCh38
rs782390347 Chr.1: 150268009-150268009 on Build GRCh38
rs782057336 Chr.1: 150267911-150267911 on Build GRCh38
rs974642209 Chr.1: 150268017-150268017 on Build GRCh38
rs907623872 Chr.1: 150268068-150268068 on Build GRCh38
rs782009159 Chr.1: 150268335-150268335 on Build GRCh38
rs782672503 Chr.1: 150267927-150267927 on Build GRCh38
rs201414382 Chr.1: 150267918-150267918 on Build GRCh38
rs782136204 Chr.1: 150268056-150268056 on Build GRCh38
rs781979095 Chr.1: 150268064-150268064 on Build GRCh38
rs587693950 Chr.1: 150268293-150268293 on Build GRCh38
rs587628123 Chr.1: 150268034-150268034 on Build GRCh38
rs1037240845 Chr.1: 150268097-150268097 on Build GRCh38
rs782245600 Chr.1: 150268004-150268004 on Build GRCh38
rs902602747 Chr.1: 150268261-150268261 on Build GRCh38
rs373094296 Chr.1: 150268033-150268033 on Build GRCh38
rs1055285642 Chr.1: 150268177-150268177 on Build GRCh38
rs79224311 Chr.1: 150268230-150268230 on Build GRCh38
rs377458916 Chr.1: 150268060-150268060 on Build GRCh38
rs782050875 Chr.1: 150268123-150268123 on Build GRCh38
rs782675679 Chr.1: 150267988-150267988 on Build GRCh38
rs782125351 Chr.1: 150268073-150268073 on Build GRCh38
rs587764147 Chr.1: 150268248-150268248 on Build GRCh38

View Full Product Details