Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150622038-150622538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENSA
Entrez Gene ID
2029
Gene Name
endosulfine alpha
Gene Aliases
ARPP-19e
UniGene
Hs.632456
Gene Chromosome Location
Chr.1: 150622123-150629622 on Build GRCh38
Transcripts
NM_004436.2 NM_207042.1 NM_207045.1 NM_207046.1

SNP Detail

SNP ID Chromosome Location
rs1025256846 Chr.1: 150622387-150622387 on Build GRCh38
rs991953665 Chr.1: 150622156-150622156 on Build GRCh38
rs1049604678 Chr.1: 150622238-150622238 on Build GRCh38
rs1001396202 Chr.1: 150622307-150622307 on Build GRCh38
rs587630875 Chr.1: 150622174-150622174 on Build GRCh38
rs921530814 Chr.1: 150622487-150622487 on Build GRCh38
rs1015673879 Chr.1: 150622287-150622287 on Build GRCh38
rs752097607 Chr.1: 150622236-150622236 on Build GRCh38
rs141944696 Chr.1: 150622290-150622290 on Build GRCh38
rs748612084 Chr.1: 150622146-150622146 on Build GRCh38
rs974355501 Chr.1: 150622477-150622477 on Build GRCh38
rs146911121 Chr.1: 150622350-150622350 on Build GRCh38
rs773277109 Chr.1: 150622243-150622243 on Build GRCh38
rs11550414 Chr.1: 150622277-150622277 on Build GRCh38
rs587716873 Chr.1: 150622202-150622202 on Build GRCh38
rs978285214 Chr.1: 150622430-150622430 on Build GRCh38
rs1033586800 Chr.1: 150622148-150622148 on Build GRCh38
rs891282013 Chr.1: 150622249-150622249 on Build GRCh38
rs1008020922 Chr.1: 150622288-150622288 on Build GRCh38
rs587714097 Chr.1: 150622168-150622168 on Build GRCh38
rs966810009 Chr.1: 150622302-150622302 on Build GRCh38
rs1053701 Chr.1: 150622225-150622225 on Build GRCh38
rs962721786 Chr.1: 150622064-150622064 on Build GRCh38
rs925050134 Chr.1: 150622441-150622441 on Build GRCh38
rs112448510 Chr.1: 150622452-150622452 on Build GRCh38
rs925308471 Chr.1: 150622138-150622138 on Build GRCh38
rs977856075 Chr.1: 150622201-150622201 on Build GRCh38
rs970978168 Chr.1: 150622445-150622445 on Build GRCh38
rs587736327 Chr.1: 150622319-150622319 on Build GRCh38
rs1048365606 Chr.1: 150622205-150622205 on Build GRCh38
rs760667512 Chr.1: 150622297-150622297 on Build GRCh38
rs996707671 Chr.1: 150622233-150622233 on Build GRCh38
rs779288831 Chr.1: 150622089-150622089 on Build GRCh38
rs957702483 Chr.1: 150622460-150622460 on Build GRCh38
rs1004727250 Chr.1: 150622257-150622257 on Build GRCh38
rs959203096 Chr.1: 150622149-150622149 on Build GRCh38
rs966550936 Chr.1: 150622417-150622417 on Build GRCh38
rs879193802 Chr.1: 150622231-150622231 on Build GRCh38
rs936339345 Chr.1: 150622209-150622209 on Build GRCh38
rs992020483 Chr.1: 150622356-150622356 on Build GRCh38
rs369618778 Chr.1: 150622169-150622169 on Build GRCh38
rs186747174 Chr.1: 150622351-150622351 on Build GRCh38
rs1021012574 Chr.1: 150622060-150622060 on Build GRCh38
rs372867833 Chr.1: 150622210-150622210 on Build GRCh38
rs967198231 Chr.1: 150622065-150622065 on Build GRCh38
rs587729545 Chr.1: 150622215-150622215 on Build GRCh38
rs587620299 Chr.1: 150622391-150622391 on Build GRCh38
rs587690910 Chr.1: 150622211-150622211 on Build GRCh38
rs867317351 Chr.1: 150622312-150622312 on Build GRCh38
rs979420205 Chr.1: 150622122-150622122 on Build GRCh38
rs587610444 Chr.1: 150622327-150622327 on Build GRCh38
rs866558323 Chr.1: 150622368-150622368 on Build GRCh38
rs762095856 Chr.1: 150622300-150622300 on Build GRCh38
rs756164180 Chr.1: 150622493-150622495 on Build GRCh38
rs891058841 Chr.1: 150622253-150622253 on Build GRCh38

View Full Product Details