Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150624249-150624784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENSA
Entrez Gene ID
2029
Gene Name
endosulfine alpha
Gene Aliases
ARPP-19e
UniGene
Hs.632456
Gene Chromosome Location
Chr.1: 150622123-150629622 on Build GRCh38
Transcripts
NM_004436.2 NM_207042.1 NM_207043.1 NM_207044.1 NM_207045.1 NM_207046.1 NM_207047.1

SNP Detail

SNP ID Chromosome Location
rs767399394 Chr.1: 150624457-150624457 on Build GRCh38
rs781095753 Chr.1: 150624346-150624347 on Build GRCh38
rs889331836 Chr.1: 150624396-150624396 on Build GRCh38
rs1050388812 Chr.1: 150624369-150624369 on Build GRCh38
rs999443451 Chr.1: 150624490-150624490 on Build GRCh38
rs972325563 Chr.1: 150624607-150624608 on Build GRCh38
rs587648697 Chr.1: 150624614-150624614 on Build GRCh38
rs1058183 Chr.1: 150624291-150624291 on Build GRCh38
rs986086416 Chr.1: 150624650-150624650 on Build GRCh38
rs1040924428 Chr.1: 150624460-150624460 on Build GRCh38
rs930620433 Chr.1: 150624353-150624353 on Build GRCh38
rs142896422 Chr.1: 150624531-150624531 on Build GRCh38
rs587751720 Chr.1: 150624576-150624576 on Build GRCh38
rs1009143408 Chr.1: 150624441-150624441 on Build GRCh38
rs1023342558 Chr.1: 150624541-150624541 on Build GRCh38
rs182083095 Chr.1: 150624649-150624649 on Build GRCh38
rs1030729369 Chr.1: 150624526-150624526 on Build GRCh38
rs930736528 Chr.1: 150624647-150624647 on Build GRCh38
rs903381577 Chr.1: 150624467-150624467 on Build GRCh38
rs772075532 Chr.1: 150624572-150624575 on Build GRCh38
rs141928183 Chr.1: 150624578-150624578 on Build GRCh38
rs961874624 Chr.1: 150624593-150624593 on Build GRCh38
rs982072114 Chr.1: 150624575-150624575 on Build GRCh38
rs927773824 Chr.1: 150624578-150624578 on Build GRCh38
rs186736317 Chr.1: 150624686-150624686 on Build GRCh38
rs745633901 Chr.1: 150624542-150624542 on Build GRCh38
rs761693369 Chr.1: 150624347-150624347 on Build GRCh38
rs587706412 Chr.1: 150624661-150624661 on Build GRCh38
rs749196509 Chr.1: 150624690-150624690 on Build GRCh38
rs189503757 Chr.1: 150624420-150624420 on Build GRCh38
rs920691707 Chr.1: 150624300-150624300 on Build GRCh38
rs920639505 Chr.1: 150624615-150624615 on Build GRCh38
rs910577886 Chr.1: 150624751-150624751 on Build GRCh38
rs145047562 Chr.1: 150624538-150624538 on Build GRCh38
rs111962711 Chr.1: 150624550-150624550 on Build GRCh38

View Full Product Details