Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180947428-180947925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL8
Entrez Gene ID
79908
Gene Name
butyrophilin like 8
Gene Aliases
BTN9.2
UniGene
Hs.189109
Gene Chromosome Location
Chr.5: 180899077-180952166 on Build GRCh38
Transcripts
NM_001040462.2 NM_001159707.1 NM_001159708.1 NM_001159709.1 NM_001159710.1 NM_024850.2 XM_011534649.1 XM_011534650.2 XM_011534651.2 XM_011534652.2 XM_011534653.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3614999 c.783C>T p.F261F Chr.5: 180947621-180947621 on Build GRCh38
4631026 c.732C>T p.C244C Chr.5: 180947570-180947570 on Build GRCh38
3615000 c.783C>T p.F261F Chr.5: 180947621-180947621 on Build GRCh38
3614995 c.722G>A p.G241E Chr.5: 180947560-180947560 on Build GRCh38
4585797 c.793C>A p.R265R Chr.5: 180947631-180947631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746228253 Chr.5: 180947666-180947666 on Build GRCh38
rs758892520 Chr.5: 180947669-180947669 on Build GRCh38
rs752462116 Chr.5: 180947481-180947481 on Build GRCh38
rs749642802 Chr.5: 180947529-180947529 on Build GRCh38
rs750267407 Chr.5: 180947724-180947724 on Build GRCh38
rs778989265 Chr.5: 180947516-180947516 on Build GRCh38
rs932112687 Chr.5: 180947831-180947831 on Build GRCh38
rs772606674 Chr.5: 180947620-180947620 on Build GRCh38
rs774551782 Chr.5: 180947491-180947491 on Build GRCh38
rs768977071 Chr.5: 180947685-180947685 on Build GRCh38
rs754492652 Chr.5: 180947652-180947652 on Build GRCh38
rs778162308 Chr.5: 180947528-180947528 on Build GRCh38
rs763666240 Chr.5: 180947740-180947740 on Build GRCh38
rs990604203 Chr.5: 180947823-180947823 on Build GRCh38
rs572816228 Chr.5: 180947874-180947874 on Build GRCh38
rs755423339 Chr.5: 180947796-180947796 on Build GRCh38
rs772962690 Chr.5: 180947707-180947707 on Build GRCh38
rs751140395 Chr.5: 180947651-180947651 on Build GRCh38
rs747736610 Chr.5: 180947592-180947592 on Build GRCh38
rs3733755 Chr.5: 180947484-180947484 on Build GRCh38
rs752371948 Chr.5: 180947730-180947730 on Build GRCh38
rs780755505 Chr.5: 180947760-180947760 on Build GRCh38
rs748197859 Chr.5: 180947697-180947697 on Build GRCh38
rs758931146 Chr.5: 180947755-180947755 on Build GRCh38
rs867663404 Chr.5: 180947692-180947692 on Build GRCh38
rs750203638 Chr.5: 180947642-180947642 on Build GRCh38
rs747345842 Chr.5: 180947787-180947787 on Build GRCh38
rs74842309 Chr.5: 180947460-180947460 on Build GRCh38
rs760832363 Chr.5: 180947627-180947627 on Build GRCh38
rs752373981 Chr.5: 180947661-180947661 on Build GRCh38
rs1008964349 Chr.5: 180947600-180947600 on Build GRCh38
rs182695105 Chr.5: 180947812-180947812 on Build GRCh38
rs7724813 Chr.5: 180947523-180947523 on Build GRCh38
rs771303790 Chr.5: 180947598-180947598 on Build GRCh38
rs1050726694 Chr.5: 180947836-180947836 on Build GRCh38
rs532378819 Chr.5: 180947624-180947624 on Build GRCh38
rs140986948 Chr.5: 180947530-180947530 on Build GRCh38
rs983167176 Chr.5: 180947738-180947738 on Build GRCh38
rs753358025 Chr.5: 180947742-180947742 on Build GRCh38
rs907947224 Chr.5: 180947456-180947456 on Build GRCh38
rs777005903 Chr.5: 180947632-180947632 on Build GRCh38
rs3733756 Chr.5: 180947534-180947534 on Build GRCh38
rs762078103 Chr.5: 180947633-180947633 on Build GRCh38
rs775606854 Chr.5: 180947556-180947556 on Build GRCh38
rs144756085 Chr.5: 180947584-180947584 on Build GRCh38
rs908000757 Chr.5: 180947802-180947802 on Build GRCh38
rs7711990 Chr.5: 180947898-180947898 on Build GRCh38
rs777716686 Chr.5: 180947739-180947744 on Build GRCh38
rs746398929 Chr.5: 180947615-180947615 on Build GRCh38
rs766330247 Chr.5: 180947648-180947648 on Build GRCh38
rs200633883 Chr.5: 180947631-180947631 on Build GRCh38
rs765024377 Chr.5: 180947576-180947576 on Build GRCh38
rs764973233 Chr.5: 180947634-180947634 on Build GRCh38
rs778744847 Chr.5: 180947497-180947497 on Build GRCh38
rs773528281 Chr.5: 180947462-180947462 on Build GRCh38
rs767571247 Chr.5: 180947501-180947501 on Build GRCh38
rs774114636 Chr.5: 180947722-180947722 on Build GRCh38
rs764018913 Chr.5: 180947509-180947509 on Build GRCh38
rs780627997 Chr.5: 180947655-180947655 on Build GRCh38
rs946571952 Chr.5: 180947832-180947832 on Build GRCh38
rs759534901 Chr.5: 180947496-180947496 on Build GRCh38
rs753752482 Chr.5: 180947572-180947572 on Build GRCh38
rs777032458 Chr.5: 180947693-180947693 on Build GRCh38
rs372311297 Chr.5: 180947732-180947732 on Build GRCh38
rs1007047589 Chr.5: 180947647-180947647 on Build GRCh38
rs768267851 Chr.5: 180947526-180947526 on Build GRCh38
rs373935289 Chr.5: 180947700-180947700 on Build GRCh38
rs552118493 Chr.5: 180947678-180947678 on Build GRCh38
rs920777889 Chr.5: 180947820-180947820 on Build GRCh38
rs766290501 Chr.5: 180947718-180947718 on Build GRCh38
rs779292643 Chr.5: 180947612-180947612 on Build GRCh38
rs865935015 Chr.5: 180947869-180947869 on Build GRCh38
rs774753065 Chr.5: 180947558-180947558 on Build GRCh38
rs752881764 Chr.5: 180947503-180947503 on Build GRCh38
rs771086335 Chr.5: 180947758-180947758 on Build GRCh38
rs781513863 Chr.5: 180947801-180947801 on Build GRCh38
rs754687088 Chr.5: 180947586-180947586 on Build GRCh38
rs753662487 Chr.5: 180947514-180947514 on Build GRCh38
rs750351235 Chr.5: 180947578-180947578 on Build GRCh38
rs755757367 Chr.5: 180947662-180947662 on Build GRCh38
rs756360979 Chr.5: 180947505-180947505 on Build GRCh38
rs923890678 Chr.5: 180947870-180947870 on Build GRCh38
rs779191429 Chr.5: 180947664-180947664 on Build GRCh38
rs368136851 Chr.5: 180947533-180947533 on Build GRCh38
rs757036689 Chr.5: 180947897-180947897 on Build GRCh38
rs1024614138 Chr.5: 180947737-180947737 on Build GRCh38
rs780822227 Chr.5: 180947591-180947591 on Build GRCh38
rs762849904 Chr.5: 180947646-180947646 on Build GRCh38
rs757860892 Chr.5: 180947522-180947522 on Build GRCh38
rs759370260 Chr.5: 180947726-180947726 on Build GRCh38
rs757320411 Chr.5: 180947515-180947515 on Build GRCh38
rs752424821 Chr.5: 180947714-180947716 on Build GRCh38
rs112294461 Chr.5: 180947566-180947566 on Build GRCh38
rs757282227 Chr.5: 180947811-180947811 on Build GRCh38
rs970782740 Chr.5: 180947758-180947758 on Build GRCh38
rs868291515 Chr.5: 180947776-180947776 on Build GRCh38
rs746162554 Chr.5: 180947786-180947786 on Build GRCh38
rs758346728 Chr.5: 180947736-180947737 on Build GRCh38
rs373702637 Chr.5: 180947520-180947520 on Build GRCh38
rs548535334 Chr.5: 180947808-180947808 on Build GRCh38
rs772072929 Chr.5: 180947540-180947540 on Build GRCh38
rs200185599 Chr.5: 180947728-180947728 on Build GRCh38
rs748413340 Chr.5: 180947804-180947804 on Build GRCh38
rs781113740 Chr.5: 180947774-180947784 on Build GRCh38
rs569031325 Chr.5: 180947716-180947716 on Build GRCh38
rs371555886 Chr.5: 180947575-180947575 on Build GRCh38
rs747089959 Chr.5: 180947750-180947750 on Build GRCh38
rs982504467 Chr.5: 180947770-180947770 on Build GRCh38

View Full Product Details