Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 17462961-17463501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAP2
Entrez Gene ID
10486
Gene Name
CAP, adenylate cyclase-associated protein, 2 (yeast)
Gene Aliases
-
UniGene
Hs.132902
Gene Chromosome Location
Chr.6: 17393216-17557792 on Build GRCh38
Transcripts
NM_006366.2 XM_011514233.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7359407 c.236A>G p.H79R Chr.6: 17463009-17463009 on Build GRCh38
3829710 c.280C>G p.Q94E Chr.6: 17463053-17463053 on Build GRCh38
6535868 c.256C>G p.R86G Chr.6: 17463029-17463029 on Build GRCh38
4911165 c.255G>A p.Q85Q Chr.6: 17463028-17463028 on Build GRCh38
2855760 c.275C>T p.A92V Chr.6: 17463048-17463048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953640093 Chr.6: 17463190-17463190 on Build GRCh38
rs763452857 Chr.6: 17463114-17463114 on Build GRCh38
rs997167108 Chr.6: 17463181-17463181 on Build GRCh38
rs576888236 Chr.6: 17463297-17463297 on Build GRCh38
rs139384783 Chr.6: 17463365-17463365 on Build GRCh38
rs373704137 Chr.6: 17463112-17463112 on Build GRCh38
rs937272147 Chr.6: 17463328-17463328 on Build GRCh38
rs536085484 Chr.6: 17463044-17463044 on Build GRCh38
rs757857111 Chr.6: 17463371-17463371 on Build GRCh38
rs778691556 Chr.6: 17463073-17463073 on Build GRCh38
rs76004196 Chr.6: 17463282-17463282 on Build GRCh38
rs746939378 Chr.6: 17463093-17463093 on Build GRCh38
rs73375206 Chr.6: 17463319-17463319 on Build GRCh38
rs745957013 Chr.6: 17463077-17463078 on Build GRCh38
rs149970340 Chr.6: 17463296-17463297 on Build GRCh38
rs1042988849 Chr.6: 17463297-17463300 on Build GRCh38
rs192171108 Chr.6: 17463409-17463409 on Build GRCh38
rs994247487 Chr.6: 17463456-17463456 on Build GRCh38
rs764719096 Chr.6: 17463247-17463247 on Build GRCh38
rs759778551 Chr.6: 17463007-17463007 on Build GRCh38
rs186017308 Chr.6: 17463396-17463396 on Build GRCh38
rs911115766 Chr.6: 17463182-17463183 on Build GRCh38
rs543412487 Chr.6: 17463085-17463085 on Build GRCh38
rs753057191 Chr.6: 17463029-17463029 on Build GRCh38
rs774742567 Chr.6: 17463116-17463116 on Build GRCh38
rs770962271 Chr.6: 17462991-17462991 on Build GRCh38
rs151328050 Chr.6: 17463250-17463250 on Build GRCh38
rs115281341 Chr.6: 17463362-17463362 on Build GRCh38
rs564685741 Chr.6: 17463269-17463269 on Build GRCh38
rs888816346 Chr.6: 17463437-17463437 on Build GRCh38
rs376763640 Chr.6: 17463101-17463101 on Build GRCh38
rs918426020 Chr.6: 17463143-17463143 on Build GRCh38
rs199714097 Chr.6: 17463291-17463291 on Build GRCh38
rs73721574 Chr.6: 17463472-17463472 on Build GRCh38
rs181026597 Chr.6: 17463375-17463375 on Build GRCh38
rs781381526 Chr.6: 17463058-17463059 on Build GRCh38
rs146348579 Chr.6: 17463032-17463032 on Build GRCh38
rs755630870 Chr.6: 17463062-17463062 on Build GRCh38
rs982122742 Chr.6: 17463252-17463252 on Build GRCh38
rs764499873 Chr.6: 17463115-17463115 on Build GRCh38
rs3777696 Chr.6: 17463435-17463435 on Build GRCh38
rs796872952 Chr.6: 17463152-17463152 on Build GRCh38
rs765221868 Chr.6: 17463396-17463396 on Build GRCh38
rs1030423124 Chr.6: 17463186-17463186 on Build GRCh38
rs945809515 Chr.6: 17463281-17463281 on Build GRCh38
rs1032518931 Chr.6: 17463460-17463460 on Build GRCh38
rs770201772 Chr.6: 17463108-17463108 on Build GRCh38
rs753287490 Chr.6: 17463123-17463123 on Build GRCh38
rs762342130 Chr.6: 17463117-17463117 on Build GRCh38
rs970800550 Chr.6: 17463251-17463251 on Build GRCh38
rs565212964 Chr.6: 17463427-17463427 on Build GRCh38
rs754558500 Chr.6: 17463071-17463071 on Build GRCh38
rs753521133 Chr.6: 17463070-17463070 on Build GRCh38
rs555940327 Chr.6: 17463086-17463086 on Build GRCh38
rs373541335 Chr.6: 17463080-17463080 on Build GRCh38
rs370999523 Chr.6: 17463355-17463355 on Build GRCh38
rs531859798 Chr.6: 17463397-17463397 on Build GRCh38
rs1025928856 Chr.6: 17463458-17463458 on Build GRCh38
rs61413697 Chr.6: 17463295-17463295 on Build GRCh38
rs984027530 Chr.6: 17463166-17463166 on Build GRCh38
rs759677585 Chr.6: 17463369-17463369 on Build GRCh38
rs575737149 Chr.6: 17463119-17463119 on Build GRCh38
rs779569230 Chr.6: 17463063-17463063 on Build GRCh38
rs868428982 Chr.6: 17463065-17463065 on Build GRCh38
rs1001101556 Chr.6: 17463351-17463351 on Build GRCh38
rs1007800180 Chr.6: 17463444-17463444 on Build GRCh38
rs1056151963 Chr.6: 17463350-17463350 on Build GRCh38
rs770934561 Chr.6: 17463098-17463098 on Build GRCh38
rs933092684 Chr.6: 17463377-17463377 on Build GRCh38
rs369966020 Chr.6: 17463061-17463061 on Build GRCh38
rs371347557 Chr.6: 17463104-17463104 on Build GRCh38
rs374033818 Chr.6: 17463197-17463197 on Build GRCh38
rs765705086 Chr.6: 17463011-17463011 on Build GRCh38
rs184340394 Chr.6: 17463293-17463293 on Build GRCh38
rs78293214 Chr.6: 17463280-17463280 on Build GRCh38
rs1008247134 Chr.6: 17463191-17463191 on Build GRCh38
rs958168079 Chr.6: 17463224-17463224 on Build GRCh38

View Full Product Details