Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 17542652-17543159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAP2
Entrez Gene ID
10486
Gene Name
CAP, adenylate cyclase-associated protein, 2 (yeast)
Gene Aliases
-
UniGene
Hs.132902
Gene Chromosome Location
Chr.6: 17393216-17557792 on Build GRCh38
Transcripts
NM_006366.2 XM_011514233.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1076261 c.1156G>T p.D386Y Chr.6: 17543090-17543090 on Build GRCh38
2855784 c.1206C>T p.I402I Chr.6: 17543140-17543140 on Build GRCh38
2855781 c.1058T>C p.V353A Chr.6: 17542892-17542892 on Build GRCh38
6325180 c.1071C>A p.F357L Chr.6: 17542905-17542905 on Build GRCh38
74005 c.1103G>T p.G368V Chr.6: 17542937-17542937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553490731 Chr.6: 17542763-17542763 on Build GRCh38
rs767372510 Chr.6: 17543045-17543045 on Build GRCh38
rs371070473 Chr.6: 17542982-17542982 on Build GRCh38
rs757206634 Chr.6: 17542807-17542807 on Build GRCh38
rs775228317 Chr.6: 17542911-17542911 on Build GRCh38
rs748794176 Chr.6: 17543123-17543123 on Build GRCh38
rs774116770 Chr.6: 17543039-17543039 on Build GRCh38
rs536207385 Chr.6: 17543044-17543045 on Build GRCh38
rs560388450 Chr.6: 17543095-17543095 on Build GRCh38
rs767603383 Chr.6: 17542954-17542954 on Build GRCh38
rs751099486 Chr.6: 17543079-17543079 on Build GRCh38
rs1038434182 Chr.6: 17542981-17542981 on Build GRCh38
rs776656785 Chr.6: 17542874-17542874 on Build GRCh38
rs370853277 Chr.6: 17543051-17543051 on Build GRCh38
rs752890966 Chr.6: 17543078-17543078 on Build GRCh38
rs28360582 Chr.6: 17542956-17542956 on Build GRCh38
rs960911253 Chr.6: 17543126-17543126 on Build GRCh38
rs376897856 Chr.6: 17542899-17542899 on Build GRCh38
rs748667060 Chr.6: 17542871-17542871 on Build GRCh38
rs756242516 Chr.6: 17542967-17542967 on Build GRCh38
rs957666194 Chr.6: 17543093-17543093 on Build GRCh38
rs762449193 Chr.6: 17542912-17542912 on Build GRCh38
rs771189477 Chr.6: 17542873-17542873 on Build GRCh38
rs779180614 Chr.6: 17542859-17542859 on Build GRCh38
rs781266775 Chr.6: 17542814-17542814 on Build GRCh38
rs762677454 Chr.6: 17542791-17542791 on Build GRCh38
rs747567074 Chr.6: 17543099-17543099 on Build GRCh38
rs539648488 Chr.6: 17542762-17542762 on Build GRCh38
rs994355286 Chr.6: 17542997-17542997 on Build GRCh38
rs755505207 Chr.6: 17542717-17542717 on Build GRCh38
rs757765135 Chr.6: 17543109-17543109 on Build GRCh38
rs540097863 Chr.6: 17543075-17543075 on Build GRCh38
rs773700042 Chr.6: 17542892-17542892 on Build GRCh38
rs780374161 Chr.6: 17542853-17542853 on Build GRCh38
rs769162149 Chr.6: 17542906-17542906 on Build GRCh38
rs762851731 Chr.6: 17542799-17542799 on Build GRCh38
rs745419382 Chr.6: 17542898-17542898 on Build GRCh38
rs1054845760 Chr.6: 17542941-17542941 on Build GRCh38
rs772625299 Chr.6: 17542889-17542889 on Build GRCh38
rs889377628 Chr.6: 17542948-17542948 on Build GRCh38
rs768259848 Chr.6: 17543129-17543129 on Build GRCh38
rs758120322 Chr.6: 17543041-17543042 on Build GRCh38
rs9256 Chr.6: 17543071-17543071 on Build GRCh38
rs768593490 Chr.6: 17542980-17542980 on Build GRCh38
rs544566542 Chr.6: 17542985-17542985 on Build GRCh38
rs748821889 Chr.6: 17543031-17543031 on Build GRCh38
rs376229266 Chr.6: 17543034-17543034 on Build GRCh38
rs758781435 Chr.6: 17543086-17543086 on Build GRCh38
rs546943293 Chr.6: 17542800-17542800 on Build GRCh38
rs374169218 Chr.6: 17542995-17542995 on Build GRCh38
rs190478527 Chr.6: 17542887-17542887 on Build GRCh38
rs756116731 Chr.6: 17542824-17542824 on Build GRCh38
rs2296249 Chr.6: 17542805-17542805 on Build GRCh38
rs774807895 Chr.6: 17543029-17543029 on Build GRCh38
rs763967309 Chr.6: 17542928-17542928 on Build GRCh38
rs139239123 Chr.6: 17542785-17542785 on Build GRCh38
rs992410319 Chr.6: 17542700-17542700 on Build GRCh38
rs769094419 Chr.6: 17542858-17542858 on Build GRCh38
rs778176298 Chr.6: 17542784-17542784 on Build GRCh38
rs1050673883 Chr.6: 17542936-17542936 on Build GRCh38
rs781721921 Chr.6: 17543121-17543121 on Build GRCh38
rs747422816 Chr.6: 17543016-17543016 on Build GRCh38
rs765479560 Chr.6: 17543077-17543077 on Build GRCh38
rs761750593 Chr.6: 17543042-17543042 on Build GRCh38
rs752454203 Chr.6: 17543011-17543012 on Build GRCh38
rs753922328 Chr.6: 17543074-17543074 on Build GRCh38
rs990553047 Chr.6: 17542693-17542693 on Build GRCh38
rs774350356 Chr.6: 17542929-17542929 on Build GRCh38
rs769374592 Chr.6: 17543019-17543019 on Build GRCh38
rs182301134 Chr.6: 17543058-17543058 on Build GRCh38
rs761701506 Chr.6: 17542951-17542951 on Build GRCh38
rs779093953 Chr.6: 17542988-17542988 on Build GRCh38
rs1051208897 Chr.6: 17543008-17543008 on Build GRCh38
rs778297246 Chr.6: 17543014-17543014 on Build GRCh38
rs750597851 Chr.6: 17542820-17542820 on Build GRCh38
rs752256619 Chr.6: 17542789-17542789 on Build GRCh38
rs1002629569 Chr.6: 17543080-17543080 on Build GRCh38
rs758728011 Chr.6: 17542998-17542998 on Build GRCh38

View Full Product Details