Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 18249562-18250059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEK
Entrez Gene ID
7913
Gene Name
DEK proto-oncogene
Gene Aliases
D6S231E
UniGene
Hs.484813
Gene Chromosome Location
Chr.6: 18224169-18264568 on Build GRCh38
Transcripts
NM_001134709.1 NM_003472.3 XM_017011273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6642223 c.745A>G p.K249E Chr.6: 18249668-18249668 on Build GRCh38
4918059 c.726G>A p.E242E Chr.6: 18249687-18249687 on Build GRCh38
5884340 c.670G>A p.E224K Chr.6: 18249743-18249743 on Build GRCh38
1076394 c.756A>C p.E252D Chr.6: 18249657-18249657 on Build GRCh38
6642225 c.613A>G p.K205E Chr.6: 18249800-18249800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021251992 Chr.6: 18249687-18249687 on Build GRCh38
rs760448785 Chr.6: 18249856-18249856 on Build GRCh38
rs768559077 Chr.6: 18249854-18249855 on Build GRCh38
rs775976286 Chr.6: 18249818-18249818 on Build GRCh38
rs767101165 Chr.6: 18249733-18249733 on Build GRCh38
rs528964669 Chr.6: 18249871-18249871 on Build GRCh38
rs753552037 Chr.6: 18249838-18249838 on Build GRCh38
rs1032191236 Chr.6: 18249888-18249888 on Build GRCh38
rs537340667 Chr.6: 18249970-18249970 on Build GRCh38
rs748859851 Chr.6: 18249666-18249668 on Build GRCh38
rs746085365 Chr.6: 18249810-18249810 on Build GRCh38
rs749636020 Chr.6: 18249881-18249881 on Build GRCh38
rs775732626 Chr.6: 18249676-18249676 on Build GRCh38
rs910022553 Chr.6: 18249665-18249665 on Build GRCh38
rs763357931 Chr.6: 18249731-18249731 on Build GRCh38
rs746143497 Chr.6: 18249875-18249875 on Build GRCh38
rs377602386 Chr.6: 18249611-18249611 on Build GRCh38
rs747299568 Chr.6: 18249697-18249697 on Build GRCh38
rs747085380 Chr.6: 18249791-18249791 on Build GRCh38
rs575836736 Chr.6: 18249627-18249627 on Build GRCh38
rs766934022 Chr.6: 18249614-18249614 on Build GRCh38
rs982522310 Chr.6: 18249962-18249962 on Build GRCh38
rs758166395 Chr.6: 18249865-18249865 on Build GRCh38
rs80090873 Chr.6: 18249987-18249987 on Build GRCh38
rs766175503 Chr.6: 18249897-18249897 on Build GRCh38
rs1051400994 Chr.6: 18249770-18249770 on Build GRCh38
rs372079467 Chr.6: 18249790-18249790 on Build GRCh38
rs773146547 Chr.6: 18249668-18249668 on Build GRCh38
rs766042113 Chr.6: 18249757-18249757 on Build GRCh38
rs759502911 Chr.6: 18249931-18249931 on Build GRCh38
rs753562715 Chr.6: 18249901-18249901 on Build GRCh38
rs533180195 Chr.6: 18249898-18249898 on Build GRCh38
rs994009625 Chr.6: 18249752-18249752 on Build GRCh38
rs769074388 Chr.6: 18249831-18249831 on Build GRCh38
rs958050966 Chr.6: 18249648-18249648 on Build GRCh38
rs1025619047 Chr.6: 18249973-18249973 on Build GRCh38
rs771080663 Chr.6: 18249799-18249799 on Build GRCh38
rs765871877 Chr.6: 18249605-18249605 on Build GRCh38
rs763553643 Chr.6: 18249829-18249829 on Build GRCh38
rs142284086 Chr.6: 18250034-18250034 on Build GRCh38
rs200257894 Chr.6: 18249726-18249726 on Build GRCh38
rs776207453 Chr.6: 18249723-18249723 on Build GRCh38
rs778671273 Chr.6: 18249772-18249772 on Build GRCh38
rs752469035 Chr.6: 18249863-18249863 on Build GRCh38
rs781569055 Chr.6: 18249801-18249801 on Build GRCh38
rs771093832 Chr.6: 18249700-18249700 on Build GRCh38
rs758239404 Chr.6: 18249785-18249785 on Build GRCh38
rs777589341 Chr.6: 18249867-18249867 on Build GRCh38
rs893803843 Chr.6: 18249952-18249952 on Build GRCh38
rs748009155 Chr.6: 18249649-18249649 on Build GRCh38
rs984154431 Chr.6: 18250025-18250025 on Build GRCh38
rs1012197571 Chr.6: 18249902-18249902 on Build GRCh38
rs756795891 Chr.6: 18249612-18249612 on Build GRCh38
rs543058394 Chr.6: 18249640-18249640 on Build GRCh38
rs368379351 Chr.6: 18249734-18249734 on Build GRCh38
rs769379509 Chr.6: 18249606-18249607 on Build GRCh38
rs574777664 Chr.6: 18249929-18249929 on Build GRCh38
rs564260336 Chr.6: 18249894-18249894 on Build GRCh38
rs371138981 Chr.6: 18249879-18249879 on Build GRCh38
rs1053901529 Chr.6: 18249944-18249944 on Build GRCh38
rs561297111 Chr.6: 18249852-18249852 on Build GRCh38
rs770107817 Chr.6: 18249878-18249878 on Build GRCh38
rs569975518 Chr.6: 18249957-18249957 on Build GRCh38
rs552228825 Chr.6: 18249784-18249784 on Build GRCh38
rs889648166 Chr.6: 18249932-18249932 on Build GRCh38
rs926824535 Chr.6: 18249746-18249746 on Build GRCh38
rs1039914491 Chr.6: 18249694-18249697 on Build GRCh38
rs140697635 Chr.6: 18249706-18249706 on Build GRCh38
rs186273654 Chr.6: 18249623-18249623 on Build GRCh38
rs116394771 Chr.6: 18249988-18249988 on Build GRCh38
rs760192042 Chr.6: 18249750-18249750 on Build GRCh38
rs942981432 Chr.6: 18249681-18249681 on Build GRCh38
rs765804831 Chr.6: 18249837-18249837 on Build GRCh38
rs754715706 Chr.6: 18249642-18249642 on Build GRCh38
rs201443004 Chr.6: 18249887-18249887 on Build GRCh38
rs770218067 Chr.6: 18249708-18249708 on Build GRCh38
rs768804503 Chr.6: 18249817-18249817 on Build GRCh38
rs779863414 Chr.6: 18249635-18249635 on Build GRCh38
rs759173674 Chr.6: 18249849-18249849 on Build GRCh38
rs774631500 Chr.6: 18249832-18249832 on Build GRCh38
rs754768955 Chr.6: 18249758-18249758 on Build GRCh38
rs752735347 Chr.6: 18249781-18249781 on Build GRCh38
rs772257561 Chr.6: 18249655-18249655 on Build GRCh38
rs201872731 Chr.6: 18249873-18249873 on Build GRCh38
rs1049999216 Chr.6: 18249909-18249909 on Build GRCh38
rs146067091 Chr.6: 18249943-18249943 on Build GRCh38
rs200071802 Chr.6: 18249645-18249645 on Build GRCh38
rs549402222 Chr.6: 18249980-18249980 on Build GRCh38
rs776948529 Chr.6: 18249701-18249701 on Build GRCh38
rs190799106 Chr.6: 18250029-18250029 on Build GRCh38
rs775257092 Chr.6: 18249661-18249664 on Build GRCh38

View Full Product Details