Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 20402527-20403031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F3
Entrez Gene ID
1871
Gene Name
E2F transcription factor 3
Gene Aliases
E2F-3
UniGene
Hs.269408
Gene Chromosome Location
Chr.6: 20353285-20493714 on Build GRCh38
Transcripts
NM_001949.4 XM_011514324.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6924796 c.340C>T p.Q114* Chr.6: 20402572-20402572 on Build GRCh38
5869852 c.326G>A p.R109K Chr.6: 20402558-20402558 on Build GRCh38
7293304 c.373G>A p.G125S Chr.6: 20402605-20402605 on Build GRCh38
6027194 c.306G>A p.T102T Chr.6: 20402538-20402538 on Build GRCh38
6941855 c.362_376del15 p.S124_G128delSGGGG Chr.6: 20402594-20402608 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs968915746 Chr.6: 20402890-20402890 on Build GRCh38
rs927292978 Chr.6: 20402926-20402926 on Build GRCh38
rs772648779 Chr.6: 20402555-20402555 on Build GRCh38
rs1021814753 Chr.6: 20402889-20402890 on Build GRCh38
rs923031760 Chr.6: 20402862-20402862 on Build GRCh38
rs1042549752 Chr.6: 20402892-20402892 on Build GRCh38
rs1004798735 Chr.6: 20402560-20402560 on Build GRCh38
rs1042497695 Chr.6: 20402722-20402722 on Build GRCh38
rs1050075345 Chr.6: 20402812-20402812 on Build GRCh38
rs563319150 Chr.6: 20402956-20402956 on Build GRCh38
rs774979715 Chr.6: 20402571-20402571 on Build GRCh38
rs746263274 Chr.6: 20402603-20402605 on Build GRCh38
rs920468183 Chr.6: 20403008-20403008 on Build GRCh38
rs781324605 Chr.6: 20402602-20402603 on Build GRCh38
rs970770531 Chr.6: 20402780-20402780 on Build GRCh38
rs192088136 Chr.6: 20402804-20402804 on Build GRCh38
rs940484727 Chr.6: 20402749-20402749 on Build GRCh38
rs762613834 Chr.6: 20402572-20402572 on Build GRCh38
rs573715776 Chr.6: 20402669-20402669 on Build GRCh38
rs956445555 Chr.6: 20402636-20402636 on Build GRCh38
rs987247639 Chr.6: 20402616-20402616 on Build GRCh38
rs779296551 Chr.6: 20402548-20402548 on Build GRCh38
rs1054445611 Chr.6: 20402930-20402930 on Build GRCh38
rs1028681244 Chr.6: 20402603-20402611 on Build GRCh38
rs866487583 Chr.6: 20402993-20402993 on Build GRCh38
rs933492725 Chr.6: 20402707-20402707 on Build GRCh38
rs886292940 Chr.6: 20402554-20402554 on Build GRCh38
rs895362853 Chr.6: 20402963-20402963 on Build GRCh38
rs980050751 Chr.6: 20402711-20402711 on Build GRCh38
rs902783733 Chr.6: 20402731-20402731 on Build GRCh38
rs189058327 Chr.6: 20402579-20402579 on Build GRCh38
rs994236349 Chr.6: 20403007-20403007 on Build GRCh38
rs890150444 Chr.6: 20402815-20402815 on Build GRCh38
rs1019776071 Chr.6: 20402630-20402630 on Build GRCh38
rs940804546 Chr.6: 20402999-20402999 on Build GRCh38
rs546158753 Chr.6: 20402751-20402751 on Build GRCh38
rs769609186 Chr.6: 20402567-20402567 on Build GRCh38
rs544998788 Chr.6: 20402916-20402916 on Build GRCh38
rs1057182343 Chr.6: 20402729-20402729 on Build GRCh38
rs1026129719 Chr.6: 20402808-20402808 on Build GRCh38
rs774125742 Chr.6: 20402585-20402585 on Build GRCh38
rs761717617 Chr.6: 20402610-20402610 on Build GRCh38
rs989070752 Chr.6: 20402668-20402668 on Build GRCh38
rs923480058 Chr.6: 20402704-20402704 on Build GRCh38
rs531223371 Chr.6: 20402982-20402982 on Build GRCh38
rs773195816 Chr.6: 20402728-20402728 on Build GRCh38
rs184141332 Chr.6: 20402843-20402843 on Build GRCh38
rs1012373846 Chr.6: 20402767-20402767 on Build GRCh38
rs907911043 Chr.6: 20402998-20402998 on Build GRCh38
rs748608331 Chr.6: 20402553-20402553 on Build GRCh38
rs760882201 Chr.6: 20402888-20402888 on Build GRCh38
rs895132746 Chr.6: 20402752-20402752 on Build GRCh38
rs947667423 Chr.6: 20402692-20402692 on Build GRCh38
rs956491973 Chr.6: 20402832-20402832 on Build GRCh38
rs746438847 Chr.6: 20402827-20402827 on Build GRCh38
rs933039650 Chr.6: 20402875-20402875 on Build GRCh38
rs960063221 Chr.6: 20402941-20402941 on Build GRCh38
rs974340575 Chr.6: 20402845-20402845 on Build GRCh38
rs1025068668 Chr.6: 20402778-20402778 on Build GRCh38
rs1037909678 Chr.6: 20403004-20403004 on Build GRCh38
rs1012671757 Chr.6: 20402978-20402978 on Build GRCh38
rs936966586 Chr.6: 20402895-20402895 on Build GRCh38
rs1001499314 Chr.6: 20402732-20402732 on Build GRCh38
rs767441139 Chr.6: 20402611-20402611 on Build GRCh38
rs776338397 Chr.6: 20402558-20402558 on Build GRCh38
rs924084184 Chr.6: 20402894-20402894 on Build GRCh38
rs908911186 Chr.6: 20402833-20402833 on Build GRCh38
rs950687594 Chr.6: 20402810-20402810 on Build GRCh38
rs745366763 Chr.6: 20402563-20402563 on Build GRCh38
rs985100408 Chr.6: 20402819-20402819 on Build GRCh38

View Full Product Details