Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 27807723-27808217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H2AI
Entrez Gene ID
8329
Gene Name
histone cluster 1, H2ai
Gene Aliases
H2A/c, H2AFC
UniGene
Hs.534035
Gene Chromosome Location
Chr.6: 27808199-27808667 on Build GRCh38
Transcripts
NM_003509.2 NM_003519.3

Gene Symbol
HIST1H2BL
Entrez Gene ID
8340
Gene Name
histone cluster 1, H2bl
Gene Aliases
H2B/c, H2BFC, dJ97D16.4
UniGene
Hs.137594
Gene Chromosome Location
Chr.6: 27807479-27807931 on Build GRCh38
Transcripts
NM_003509.2 NM_003519.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7077202 c.72G>A p.K24K Chr.6: 27807835-27807835 on Build GRCh38
320832 c.20C>T p.S7F Chr.6: 27807887-27807887 on Build GRCh38
1076955 c.20C>A p.S7Y Chr.6: 27807887-27807887 on Build GRCh38
4353587 c.90C>T p.R30R Chr.6: 27807817-27807817 on Build GRCh38
3829878 c.134T>G p.V45G Chr.6: 27807773-27807773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574118993 Chr.6: 27808003-27808003 on Build GRCh38
rs766364793 Chr.6: 27808162-27808162 on Build GRCh38
rs536051122 Chr.6: 27807829-27807829 on Build GRCh38
rs758565851 Chr.6: 27808172-27808172 on Build GRCh38
rs565241481 Chr.6: 27808163-27808163 on Build GRCh38
rs774046092 Chr.6: 27807947-27807947 on Build GRCh38
rs779323148 Chr.6: 27807790-27807790 on Build GRCh38
rs770097608 Chr.6: 27807787-27807787 on Build GRCh38
rs781712221 Chr.6: 27808178-27808178 on Build GRCh38
rs780283212 Chr.6: 27807789-27807789 on Build GRCh38
rs781524346 Chr.6: 27807783-27807783 on Build GRCh38
rs764759987 Chr.6: 27807745-27807745 on Build GRCh38
rs771058376 Chr.6: 27808188-27808188 on Build GRCh38
rs144303836 Chr.6: 27807830-27807830 on Build GRCh38
rs1019412235 Chr.6: 27807847-27807847 on Build GRCh38
rs774693408 Chr.6: 27807794-27807794 on Build GRCh38
rs111621043 Chr.6: 27808048-27808048 on Build GRCh38
rs145663173 Chr.6: 27807908-27807908 on Build GRCh38
rs769540388 Chr.6: 27808189-27808189 on Build GRCh38
rs199859119 Chr.6: 27808174-27808174 on Build GRCh38
rs372391364 Chr.6: 27807859-27807859 on Build GRCh38
rs760670244 Chr.6: 27808059-27808059 on Build GRCh38
rs777558760 Chr.6: 27807765-27807765 on Build GRCh38
rs369136464 Chr.6: 27807929-27807929 on Build GRCh38
rs757721150 Chr.6: 27807940-27807940 on Build GRCh38
rs550217184 Chr.6: 27807804-27807804 on Build GRCh38
rs756970532 Chr.6: 27807778-27807778 on Build GRCh38
rs772348767 Chr.6: 27807799-27807799 on Build GRCh38
rs772565457 Chr.6: 27807867-27807867 on Build GRCh38
rs141811762 Chr.6: 27807897-27807897 on Build GRCh38
rs773743915 Chr.6: 27807906-27807906 on Build GRCh38
rs935478201 Chr.6: 27807912-27807912 on Build GRCh38
rs368194728 Chr.6: 27807849-27807849 on Build GRCh38
rs747534875 Chr.6: 27807935-27807935 on Build GRCh38
rs767500935 Chr.6: 27807878-27807878 on Build GRCh38
rs750172798 Chr.6: 27807837-27807837 on Build GRCh38
rs756467668 Chr.6: 27808180-27808180 on Build GRCh38
rs759193418 Chr.6: 27807805-27807805 on Build GRCh38
rs762291103 Chr.6: 27807797-27807797 on Build GRCh38
rs891488923 Chr.6: 27807767-27807767 on Build GRCh38
rs780475526 Chr.6: 27807882-27807882 on Build GRCh38
rs753606765 Chr.6: 27807920-27807920 on Build GRCh38
rs775106132 Chr.6: 27808192-27808192 on Build GRCh38
rs1046478297 Chr.6: 27807909-27807909 on Build GRCh38
rs778477152 Chr.6: 27808183-27808183 on Build GRCh38
rs493121 Chr.6: 27808169-27808169 on Build GRCh38
rs778250099 Chr.6: 27807901-27807901 on Build GRCh38
rs752263384 Chr.6: 27807810-27807810 on Build GRCh38
rs993999642 Chr.6: 27808127-27808127 on Build GRCh38
rs751429667 Chr.6: 27807828-27807828 on Build GRCh38
rs540640730 Chr.6: 27808150-27808150 on Build GRCh38
rs952848945 Chr.6: 27807900-27807900 on Build GRCh38
rs771732804 Chr.6: 27807952-27807952 on Build GRCh38
rs181697460 Chr.6: 27808185-27808185 on Build GRCh38
rs745957145 Chr.6: 27807832-27807834 on Build GRCh38
rs764424803 Chr.6: 27807918-27807919 on Build GRCh38
rs952674805 Chr.6: 27808144-27808144 on Build GRCh38
rs1011227710 Chr.6: 27807802-27807802 on Build GRCh38
rs570328618 Chr.6: 27808174-27808175 on Build GRCh38
rs773250251 Chr.6: 27807913-27807913 on Build GRCh38
rs965215100 Chr.6: 27808068-27808068 on Build GRCh38
rs542836666 Chr.6: 27808036-27808036 on Build GRCh38
rs201231863 Chr.6: 27807838-27807838 on Build GRCh38
rs372174452 Chr.6: 27807914-27807914 on Build GRCh38
rs755169326 Chr.6: 27807921-27807921 on Build GRCh38
rs192494911 Chr.6: 27808126-27808126 on Build GRCh38
rs750815899 Chr.6: 27808179-27808179 on Build GRCh38
rs199544124 Chr.6: 27808187-27808187 on Build GRCh38
rs747415652 Chr.6: 27807903-27807903 on Build GRCh38
rs148711105 Chr.6: 27807869-27807869 on Build GRCh38
rs559871342 Chr.6: 27808052-27808052 on Build GRCh38
rs142297357 Chr.6: 27807894-27807894 on Build GRCh38
rs780406007 Chr.6: 27807841-27807841 on Build GRCh38
rs536656957 Chr.6: 27807786-27807786 on Build GRCh38
rs769943414 Chr.6: 27807907-27807908 on Build GRCh38
rs202083966 Chr.6: 27808186-27808186 on Build GRCh38
rs200485 Chr.6: 27807919-27807919 on Build GRCh38
rs371492098 Chr.6: 27807760-27807760 on Build GRCh38
rs150000460 Chr.6: 27807812-27807812 on Build GRCh38
rs747110698 Chr.6: 27808176-27808176 on Build GRCh38
rs567071925 Chr.6: 27807823-27807823 on Build GRCh38
rs1019317732 Chr.6: 27808067-27808067 on Build GRCh38
rs759371188 Chr.6: 27808167-27808167 on Build GRCh38
rs550292131 Chr.6: 27807942-27807942 on Build GRCh38
rs763166237 Chr.6: 27808193-27808193 on Build GRCh38
rs538894622 Chr.6: 27807874-27807874 on Build GRCh38
rs375129466 Chr.6: 27807892-27807892 on Build GRCh38
rs754138290 Chr.6: 27807886-27807886 on Build GRCh38
rs139309275 Chr.6: 27807776-27807776 on Build GRCh38
rs769504743 Chr.6: 27807871-27807871 on Build GRCh38
rs772810523 Chr.6: 27807958-27807958 on Build GRCh38
rs761369367 Chr.6: 27807907-27807907 on Build GRCh38
rs780878778 Chr.6: 27807833-27807833 on Build GRCh38
rs374721250 Chr.6: 27807784-27807784 on Build GRCh38
rs779007041 Chr.6: 27807855-27807855 on Build GRCh38
rs1025877796 Chr.6: 27808128-27808128 on Build GRCh38
rs758634123 Chr.6: 27807757-27807757 on Build GRCh38
rs551618764 Chr.6: 27808161-27808161 on Build GRCh38
rs573030664 Chr.6: 27807872-27807872 on Build GRCh38
rs752788097 Chr.6: 27807923-27807923 on Build GRCh38
rs769652233 Chr.6: 27808190-27808190 on Build GRCh38
rs749442063 Chr.6: 27807844-27807844 on Build GRCh38
rs368046141 Chr.6: 27807904-27807904 on Build GRCh38
rs764188269 Chr.6: 27808173-27808173 on Build GRCh38
rs763202728 Chr.6: 27807911-27807912 on Build GRCh38
rs750299249 Chr.6: 27807880-27807880 on Build GRCh38
rs953813898 Chr.6: 27807817-27807817 on Build GRCh38
rs758475262 Chr.6: 27807930-27807930 on Build GRCh38
rs201932219 Chr.6: 27807759-27807759 on Build GRCh38
rs773581237 Chr.6: 27807868-27807868 on Build GRCh38
rs747647947 Chr.6: 27807948-27807948 on Build GRCh38
rs773703254 Chr.6: 27807764-27807764 on Build GRCh38
rs35074066 Chr.6: 27807758-27807759 on Build GRCh38
rs761656522 Chr.6: 27807877-27807877 on Build GRCh38
rs778300859 Chr.6: 27807932-27807932 on Build GRCh38
rs764813903 Chr.6: 27807809-27807809 on Build GRCh38
rs775885933 Chr.6: 27807908-27807909 on Build GRCh38
rs200484 Chr.6: 27807896-27807896 on Build GRCh38
rs372762733 Chr.6: 27807946-27807946 on Build GRCh38
rs928157594 Chr.6: 27807911-27807911 on Build GRCh38
rs1006594923 Chr.6: 27808065-27808065 on Build GRCh38

View Full Product Details