Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36839288-36839787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf89
Entrez Gene ID
221477
Gene Name
chromosome 6 open reading frame 89
Gene Aliases
BRAP
UniGene
Hs.433381
Gene Chromosome Location
Chr.6: 36834886-36928964 on Build GRCh38
Transcripts
XM_017010434.1 NM_001314017.1 NM_020939.1 XM_005249247.2 XM_011514768.1 XM_011514769.1 XM_011514770.1 XM_011514772.1

Gene Symbol
CPNE5
Entrez Gene ID
57699
Gene Name
copine 5
Gene Aliases
COPN5, CPN5
UniGene
Hs.657869
Gene Chromosome Location
Chr.6: 36740773-36839444 on Build GRCh38
Transcripts
XM_017010434.1 NM_001314017.1 NM_020939.1 XM_005249247.2 XM_011514768.1 XM_011514769.1 XM_011514770.1 XM_011514772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6314992 c.7C>A p.Q3K Chr.6: 36839371-36839371 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373616222 Chr.6: 36839329-36839329 on Build GRCh38
rs118013238 Chr.6: 36839547-36839547 on Build GRCh38
rs1010311440 Chr.6: 36839521-36839521 on Build GRCh38
rs987758539 Chr.6: 36839720-36839720 on Build GRCh38
rs768313580 Chr.6: 36839356-36839356 on Build GRCh38
rs935701603 Chr.6: 36839488-36839488 on Build GRCh38
rs950747760 Chr.6: 36839699-36839699 on Build GRCh38
rs770269192 Chr.6: 36839312-36839312 on Build GRCh38
rs904245019 Chr.6: 36839459-36839460 on Build GRCh38
rs929927086 Chr.6: 36839344-36839344 on Build GRCh38
rs201480226 Chr.6: 36839378-36839378 on Build GRCh38
rs780508158 Chr.6: 36839328-36839328 on Build GRCh38
rs865946556 Chr.6: 36839342-36839342 on Build GRCh38
rs763109999 Chr.6: 36839393-36839393 on Build GRCh38
rs371123592 Chr.6: 36839381-36839381 on Build GRCh38
rs1052782877 Chr.6: 36839510-36839510 on Build GRCh38
rs905699516 Chr.6: 36839596-36839596 on Build GRCh38
rs964854051 Chr.6: 36839761-36839761 on Build GRCh38
rs1001336138 Chr.6: 36839626-36839626 on Build GRCh38
rs368691298 Chr.6: 36839334-36839334 on Build GRCh38
rs912106056 Chr.6: 36839741-36839742 on Build GRCh38
rs904418265 Chr.6: 36839555-36839555 on Build GRCh38
rs1006783326 Chr.6: 36839578-36839578 on Build GRCh38
rs910115586 Chr.6: 36839349-36839349 on Build GRCh38
rs766170114 Chr.6: 36839387-36839387 on Build GRCh38
rs988072935 Chr.6: 36839426-36839426 on Build GRCh38
rs1038760623 Chr.6: 36839439-36839439 on Build GRCh38
rs183702303 Chr.6: 36839385-36839385 on Build GRCh38
rs929555611 Chr.6: 36839417-36839417 on Build GRCh38
rs919542393 Chr.6: 36839397-36839397 on Build GRCh38
rs1026300870 Chr.6: 36839668-36839668 on Build GRCh38
rs373299600 Chr.6: 36839656-36839656 on Build GRCh38
rs1033799330 Chr.6: 36839633-36839633 on Build GRCh38
rs1014541065 Chr.6: 36839318-36839318 on Build GRCh38
rs773753997 Chr.6: 36839365-36839365 on Build GRCh38
rs912074949 Chr.6: 36839427-36839427 on Build GRCh38
rs891584771 Chr.6: 36839516-36839516 on Build GRCh38
rs965148301 Chr.6: 36839702-36839702 on Build GRCh38
rs918675982 Chr.6: 36839314-36839314 on Build GRCh38
rs973422124 Chr.6: 36839391-36839391 on Build GRCh38
rs1056865874 Chr.6: 36839486-36839486 on Build GRCh38
rs976557504 Chr.6: 36839706-36839706 on Build GRCh38
rs963455803 Chr.6: 36839642-36839642 on Build GRCh38
rs1003721907 Chr.6: 36839429-36839429 on Build GRCh38
rs776372762 Chr.6: 36839389-36839389 on Build GRCh38
rs576210382 Chr.6: 36839576-36839576 on Build GRCh38
rs1048429683 Chr.6: 36839348-36839348 on Build GRCh38

View Full Product Details