Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36855928-36856435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf89
Entrez Gene ID
221477
Gene Name
chromosome 6 open reading frame 89
Gene Aliases
BRAP
UniGene
Hs.433381
Gene Chromosome Location
Chr.6: 36834886-36928964 on Build GRCh38
Transcripts
XM_017010434.1 NM_016059.4

Gene Symbol
PPIL1
Entrez Gene ID
51645
Gene Name
peptidylprolyl isomerase like 1
Gene Aliases
CGI-124, CYPL1, PPIase, hCyPX
UniGene
Hs.27693
Gene Chromosome Location
Chr.6: 36854829-36875024 on Build GRCh38
Transcripts
XM_017010434.1 NM_016059.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1312154 c.303C>T p.A101A Chr.6: 36856011-36856011 on Build GRCh38
4545156 c.371G>A p.G124D Chr.6: 36855943-36855943 on Build GRCh38
5405148 c.292C>T p.L98F Chr.6: 36856022-36856022 on Build GRCh38
3626294 c.336C>T p.F112F Chr.6: 36855978-36855978 on Build GRCh38
3626295 c.305A>G p.N102S Chr.6: 36856009-36856009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112562781 Chr.6: 36856339-36856339 on Build GRCh38
rs934642672 Chr.6: 36856228-36856228 on Build GRCh38
rs747235472 Chr.6: 36856058-36856058 on Build GRCh38
rs756837992 Chr.6: 36856017-36856017 on Build GRCh38
rs549759641 Chr.6: 36855961-36855961 on Build GRCh38
rs771145903 Chr.6: 36855966-36855966 on Build GRCh38
rs749013271 Chr.6: 36855962-36855962 on Build GRCh38
rs753149952 Chr.6: 36855991-36855991 on Build GRCh38
rs779809974 Chr.6: 36855987-36855988 on Build GRCh38
rs771012391 Chr.6: 36856059-36856059 on Build GRCh38
rs912210443 Chr.6: 36855993-36855993 on Build GRCh38
rs953888780 Chr.6: 36856103-36856103 on Build GRCh38
rs765644535 Chr.6: 36855990-36855990 on Build GRCh38
rs1037344391 Chr.6: 36856008-36856008 on Build GRCh38
rs934923702 Chr.6: 36855970-36855970 on Build GRCh38
rs758695577 Chr.6: 36856089-36856089 on Build GRCh38
rs1029126393 Chr.6: 36856181-36856181 on Build GRCh38
rs756126155 Chr.6: 36856036-36856036 on Build GRCh38
rs28620149 Chr.6: 36856038-36856038 on Build GRCh38
rs796360075 Chr.6: 36856336-36856336 on Build GRCh38
rs776823713 Chr.6: 36856067-36856067 on Build GRCh38
rs544727115 Chr.6: 36856245-36856245 on Build GRCh38
rs912343939 Chr.6: 36856217-36856217 on Build GRCh38
rs754802802 Chr.6: 36856042-36856042 on Build GRCh38
rs985323732 Chr.6: 36856131-36856131 on Build GRCh38
rs1046508565 Chr.6: 36856056-36856056 on Build GRCh38
rs558943116 Chr.6: 36856206-36856206 on Build GRCh38
rs1032774041 Chr.6: 36856090-36856090 on Build GRCh38
rs568130003 Chr.6: 36855965-36855965 on Build GRCh38
rs888578061 Chr.6: 36856279-36856279 on Build GRCh38
rs767270704 Chr.6: 36856258-36856258 on Build GRCh38
rs4604269 Chr.6: 36856142-36856142 on Build GRCh38
rs1037185452 Chr.6: 36856318-36856318 on Build GRCh38
rs772403872 Chr.6: 36856372-36856373 on Build GRCh38
rs949274608 Chr.6: 36856372-36856372 on Build GRCh38
rs544086562 Chr.6: 36856074-36856074 on Build GRCh38
rs774055871 Chr.6: 36856159-36856159 on Build GRCh38
rs111898081 Chr.6: 36856020-36856020 on Build GRCh38
rs778790886 Chr.6: 36856048-36856048 on Build GRCh38
rs1046897318 Chr.6: 36856376-36856376 on Build GRCh38
rs563697602 Chr.6: 36856414-36856414 on Build GRCh38
rs768832306 Chr.6: 36856057-36856058 on Build GRCh38
rs745821421 Chr.6: 36855953-36855953 on Build GRCh38
rs779823888 Chr.6: 36856037-36856037 on Build GRCh38
rs185738522 Chr.6: 36856039-36856039 on Build GRCh38
rs896849850 Chr.6: 36856324-36856324 on Build GRCh38
rs368185019 Chr.6: 36856287-36856287 on Build GRCh38
rs902571765 Chr.6: 36856383-36856383 on Build GRCh38
rs986947754 Chr.6: 36856216-36856216 on Build GRCh38
rs999555154 Chr.6: 36856069-36856069 on Build GRCh38
rs939738054 Chr.6: 36855998-36855998 on Build GRCh38
rs919608650 Chr.6: 36856269-36856269 on Build GRCh38
rs917510534 Chr.6: 36856252-36856252 on Build GRCh38
rs749128143 Chr.6: 36855998-36855998 on Build GRCh38
rs779921408 Chr.6: 36855960-36855960 on Build GRCh38
rs958246899 Chr.6: 36856104-36856104 on Build GRCh38
rs769575987 Chr.6: 36855958-36855958 on Build GRCh38
rs763795089 Chr.6: 36856005-36856005 on Build GRCh38
rs986569193 Chr.6: 36855992-36855992 on Build GRCh38
rs199818754 Chr.6: 36856019-36856019 on Build GRCh38
rs968835343 Chr.6: 36856188-36856188 on Build GRCh38
rs763303819 Chr.6: 36855996-36855996 on Build GRCh38
rs116315976 Chr.6: 36856390-36856390 on Build GRCh38
rs149386685 Chr.6: 36855984-36855984 on Build GRCh38
rs577370748 Chr.6: 36856277-36856277 on Build GRCh38
rs915038091 Chr.6: 36856172-36856172 on Build GRCh38
rs996437477 Chr.6: 36856106-36856106 on Build GRCh38
rs201473369 Chr.6: 36855964-36855964 on Build GRCh38
rs2071822 Chr.6: 36856325-36856325 on Build GRCh38
rs751228235 Chr.6: 36856016-36856016 on Build GRCh38
rs1029567944 Chr.6: 36856100-36856100 on Build GRCh38
rs201822462 Chr.6: 36855978-36855978 on Build GRCh38
rs972404344 Chr.6: 36856246-36856246 on Build GRCh38

View Full Product Details