Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36899118-36899665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf89
Entrez Gene ID
221477
Gene Name
chromosome 6 open reading frame 89
Gene Aliases
BRAP
UniGene
Hs.433381
Gene Chromosome Location
Chr.6: 36834886-36928964 on Build GRCh38
Transcripts
NM_001286635.1 NM_001286636.1 NM_001286637.1 NM_152734.3 XM_011514377.2 XM_017010432.1 XM_017010433.1 XM_017010434.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5472264 c.145C>T p.L49L Chr.6: 36899568-36899568 on Build GRCh38
3076311 c.174C>A p.P58P Chr.6: 36899597-36899597 on Build GRCh38
7113195 c.156G>C p.K52N Chr.6: 36899579-36899579 on Build GRCh38
4765172 c.172_173insC p.Q61fs*10 Chr.6: 36899595-36899596 on Build GRCh38
1734391 c.172_173insC p.Q61fs*10 Chr.6: 36899595-36899596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778504462 Chr.6: 36899466-36899466 on Build GRCh38
rs374490982 Chr.6: 36899462-36899462 on Build GRCh38
rs1015072528 Chr.6: 36899612-36899612 on Build GRCh38
rs768194666 Chr.6: 36899548-36899548 on Build GRCh38
rs939855241 Chr.6: 36899627-36899627 on Build GRCh38
rs11542162 Chr.6: 36899493-36899493 on Build GRCh38
rs745620526 Chr.6: 36899603-36899603 on Build GRCh38
rs1029244927 Chr.6: 36899145-36899145 on Build GRCh38
rs374760313 Chr.6: 36899543-36899543 on Build GRCh38
rs539575159 Chr.6: 36899505-36899505 on Build GRCh38
rs760020887 Chr.6: 36899595-36899595 on Build GRCh38
rs116014494 Chr.6: 36899415-36899415 on Build GRCh38
rs768992037 Chr.6: 36899605-36899605 on Build GRCh38
rs771626432 Chr.6: 36899387-36899387 on Build GRCh38
rs146491196 Chr.6: 36899311-36899311 on Build GRCh38
rs954853564 Chr.6: 36899230-36899230 on Build GRCh38
rs761020486 Chr.6: 36899568-36899568 on Build GRCh38
rs186256883 Chr.6: 36899199-36899199 on Build GRCh38
rs775798976 Chr.6: 36899402-36899402 on Build GRCh38
rs368640375 Chr.6: 36899492-36899492 on Build GRCh38
rs762067759 Chr.6: 36899629-36899629 on Build GRCh38
rs79604474 Chr.6: 36899146-36899146 on Build GRCh38
rs753532631 Chr.6: 36899455-36899455 on Build GRCh38
rs567738213 Chr.6: 36899498-36899498 on Build GRCh38
rs766794131 Chr.6: 36899570-36899570 on Build GRCh38
rs1009038077 Chr.6: 36899232-36899232 on Build GRCh38
rs765729368 Chr.6: 36899636-36899636 on Build GRCh38
rs140541687 Chr.6: 36899602-36899602 on Build GRCh38
rs901880749 Chr.6: 36899332-36899332 on Build GRCh38
rs149702809 Chr.6: 36899607-36899607 on Build GRCh38
rs112935351 Chr.6: 36899418-36899418 on Build GRCh38
rs370949292 Chr.6: 36899444-36899444 on Build GRCh38
rs370774971 Chr.6: 36899530-36899530 on Build GRCh38
rs1015003092 Chr.6: 36899314-36899314 on Build GRCh38
rs763022315 Chr.6: 36899409-36899409 on Build GRCh38
rs780224573 Chr.6: 36899445-36899445 on Build GRCh38
rs928317498 Chr.6: 36899216-36899216 on Build GRCh38
rs140094280 Chr.6: 36899222-36899222 on Build GRCh38
rs764265156 Chr.6: 36899414-36899414 on Build GRCh38
rs760461422 Chr.6: 36899400-36899400 on Build GRCh38
rs935403880 Chr.6: 36899503-36899503 on Build GRCh38
rs748111288 Chr.6: 36899380-36899380 on Build GRCh38
rs754542766 Chr.6: 36899463-36899463 on Build GRCh38
rs777676667 Chr.6: 36899482-36899482 on Build GRCh38
rs757919569 Chr.6: 36899599-36899599 on Build GRCh38
rs144844111 Chr.6: 36899597-36899597 on Build GRCh38
rs376471859 Chr.6: 36899429-36899429 on Build GRCh38
rs945795642 Chr.6: 36899198-36899198 on Build GRCh38
rs772035548 Chr.6: 36899480-36899480 on Build GRCh38
rs1015105196 Chr.6: 36899251-36899251 on Build GRCh38
rs747632694 Chr.6: 36899469-36899469 on Build GRCh38
rs762460277 Chr.6: 36899424-36899424 on Build GRCh38
rs568772621 Chr.6: 36899229-36899229 on Build GRCh38
rs902566591 Chr.6: 36899176-36899176 on Build GRCh38
rs183185724 Chr.6: 36899156-36899156 on Build GRCh38
rs768739302 Chr.6: 36899519-36899519 on Build GRCh38
rs777115998 Chr.6: 36899594-36899595 on Build GRCh38
rs763258481 Chr.6: 36899516-36899516 on Build GRCh38
rs972490550 Chr.6: 36899343-36899343 on Build GRCh38
rs73732050 Chr.6: 36899600-36899600 on Build GRCh38
rs879158913 Chr.6: 36899334-36899334 on Build GRCh38
rs773037331 Chr.6: 36899398-36899398 on Build GRCh38
rs1048228548 Chr.6: 36899258-36899258 on Build GRCh38
rs1029375946 Chr.6: 36899346-36899346 on Build GRCh38
rs1032314151 Chr.6: 36899180-36899180 on Build GRCh38
rs144084583 Chr.6: 36899278-36899278 on Build GRCh38
rs771945567 Chr.6: 36899388-36899388 on Build GRCh38
rs999582287 Chr.6: 36899177-36899177 on Build GRCh38
rs369066579 Chr.6: 36899215-36899215 on Build GRCh38
rs140986907 Chr.6: 36899596-36899596 on Build GRCh38
rs919589559 Chr.6: 36899361-36899361 on Build GRCh38
rs1046222188 Chr.6: 36899160-36899160 on Build GRCh38
rs752265379 Chr.6: 36899598-36899598 on Build GRCh38
rs190776591 Chr.6: 36899567-36899567 on Build GRCh38
rs768108755 Chr.6: 36899428-36899428 on Build GRCh38
rs996422932 Chr.6: 36899218-36899218 on Build GRCh38

View Full Product Details