Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41771235-41771739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRS3
Entrez Gene ID
10817
Gene Name
fibroblast growth factor receptor substrate 3
Gene Aliases
FRS2-beta, FRS2B, FRS2beta, SNT-2, SNT2
UniGene
Hs.194208
Gene Chromosome Location
Chr.6: 41770176-41779905 on Build GRCh38
Transcripts
NM_006653.4 XM_011514254.2 XM_011514255.2 XM_017010193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6230581 c.749G>A p.R250Q Chr.6: 41771349-41771349 on Build GRCh38
484068 c.666G>T p.Q222H Chr.6: 41771432-41771432 on Build GRCh38
6173992 c.771G>T p.Q257H Chr.6: 41771327-41771327 on Build GRCh38
7251381 c.748C>T p.R250W Chr.6: 41771350-41771350 on Build GRCh38
3874004 c.712G>T p.G238C Chr.6: 41771386-41771386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781424408 Chr.6: 41771303-41771303 on Build GRCh38
rs142384930 Chr.6: 41771348-41771348 on Build GRCh38
rs149311369 Chr.6: 41771413-41771413 on Build GRCh38
rs901456824 Chr.6: 41771623-41771623 on Build GRCh38
rs1028352418 Chr.6: 41771703-41771703 on Build GRCh38
rs561289920 Chr.6: 41771315-41771315 on Build GRCh38
rs150084831 Chr.6: 41771360-41771360 on Build GRCh38
rs995757935 Chr.6: 41771687-41771687 on Build GRCh38
rs965414071 Chr.6: 41771374-41771374 on Build GRCh38
rs751299746 Chr.6: 41771462-41771462 on Build GRCh38
rs745616952 Chr.6: 41771255-41771255 on Build GRCh38
rs545948718 Chr.6: 41771702-41771702 on Build GRCh38
rs753719853 Chr.6: 41771454-41771454 on Build GRCh38
rs940232802 Chr.6: 41771608-41771608 on Build GRCh38
rs376907371 Chr.6: 41771289-41771289 on Build GRCh38
rs751103525 Chr.6: 41771561-41771561 on Build GRCh38
rs778553223 Chr.6: 41771480-41771480 on Build GRCh38
rs760799697 Chr.6: 41771532-41771532 on Build GRCh38
rs559997076 Chr.6: 41771717-41771717 on Build GRCh38
rs138079167 Chr.6: 41771428-41771428 on Build GRCh38
rs112111940 Chr.6: 41771475-41771475 on Build GRCh38
rs773366143 Chr.6: 41771526-41771526 on Build GRCh38
rs769181541 Chr.6: 41771429-41771429 on Build GRCh38
rs751765291 Chr.6: 41771500-41771500 on Build GRCh38
rs138673129 Chr.6: 41771398-41771398 on Build GRCh38
rs150886179 Chr.6: 41771601-41771601 on Build GRCh38
rs369514159 Chr.6: 41771659-41771659 on Build GRCh38
rs754863883 Chr.6: 41771469-41771469 on Build GRCh38
rs761378284 Chr.6: 41771284-41771284 on Build GRCh38
rs777337475 Chr.6: 41771484-41771484 on Build GRCh38
rs35310379 Chr.6: 41771435-41771435 on Build GRCh38
rs79413344 Chr.6: 41771613-41771613 on Build GRCh38
rs1015039808 Chr.6: 41771624-41771624 on Build GRCh38
rs777044783 Chr.6: 41771536-41771536 on Build GRCh38
rs745679917 Chr.6: 41771309-41771309 on Build GRCh38
rs376501078 Chr.6: 41771644-41771644 on Build GRCh38
rs746455483 Chr.6: 41771490-41771490 on Build GRCh38
rs745579154 Chr.6: 41771585-41771585 on Build GRCh38
rs368454025 Chr.6: 41771298-41771298 on Build GRCh38
rs769112764 Chr.6: 41771506-41771506 on Build GRCh38
rs751587190 Chr.6: 41771299-41771299 on Build GRCh38
rs912190094 Chr.6: 41771420-41771420 on Build GRCh38
rs780781377 Chr.6: 41771493-41771493 on Build GRCh38
rs754989847 Chr.6: 41771405-41771405 on Build GRCh38
rs770198041 Chr.6: 41771357-41771357 on Build GRCh38
rs913808081 Chr.6: 41771256-41771256 on Build GRCh38
rs200599287 Chr.6: 41771347-41771347 on Build GRCh38
rs137923290 Chr.6: 41771346-41771346 on Build GRCh38
rs532704379 Chr.6: 41771353-41771353 on Build GRCh38
rs200303747 Chr.6: 41771350-41771350 on Build GRCh38
rs202151715 Chr.6: 41771358-41771358 on Build GRCh38
rs758259654 Chr.6: 41771482-41771482 on Build GRCh38
rs760992457 Chr.6: 41771446-41771446 on Build GRCh38
rs751611880 Chr.6: 41771370-41771370 on Build GRCh38
rs963433457 Chr.6: 41771650-41771650 on Build GRCh38
rs771664907 Chr.6: 41771418-41771418 on Build GRCh38
rs3747747 Chr.6: 41771436-41771436 on Build GRCh38
rs140801495 Chr.6: 41771438-41771438 on Build GRCh38
rs764525337 Chr.6: 41771361-41771361 on Build GRCh38
rs753256771 Chr.6: 41771291-41771291 on Build GRCh38
rs766661900 Chr.6: 41771453-41771453 on Build GRCh38
rs748007549 Chr.6: 41771414-41771414 on Build GRCh38
rs753630899 Chr.6: 41771404-41771404 on Build GRCh38
rs767463234 Chr.6: 41771381-41771381 on Build GRCh38
rs756803085 Chr.6: 41771571-41771571 on Build GRCh38
rs953956294 Chr.6: 41771412-41771412 on Build GRCh38
rs144660735 Chr.6: 41771483-41771483 on Build GRCh38
rs369939197 Chr.6: 41771510-41771510 on Build GRCh38
rs780000592 Chr.6: 41771399-41771399 on Build GRCh38
rs750706880 Chr.6: 41771389-41771389 on Build GRCh38
rs763892744 Chr.6: 41771560-41771560 on Build GRCh38
rs200242322 Chr.6: 41771511-41771511 on Build GRCh38
rs769240284 Chr.6: 41771349-41771349 on Build GRCh38
rs749206431 Chr.6: 41771276-41771276 on Build GRCh38
rs199883529 Chr.6: 41771301-41771301 on Build GRCh38
rs776884574 Chr.6: 41771287-41771287 on Build GRCh38
rs766183352 Chr.6: 41771461-41771461 on Build GRCh38
rs780659074 Chr.6: 41771572-41771572 on Build GRCh38
rs370835870 Chr.6: 41771316-41771316 on Build GRCh38
rs761911304 Chr.6: 41771371-41771371 on Build GRCh38
rs775528221 Chr.6: 41771275-41771275 on Build GRCh38
rs887289107 Chr.6: 41771597-41771597 on Build GRCh38
rs767141975 Chr.6: 41771285-41771285 on Build GRCh38
rs779972215 Chr.6: 41771317-41771317 on Build GRCh38
rs772401685 Chr.6: 41771525-41771525 on Build GRCh38
rs374326438 Chr.6: 41771336-41771336 on Build GRCh38
rs768339772 Chr.6: 41771278-41771278 on Build GRCh38
rs1054750288 Chr.6: 41771539-41771539 on Build GRCh38
rs752372768 Chr.6: 41771551-41771551 on Build GRCh38
rs937725881 Chr.6: 41771295-41771295 on Build GRCh38
rs765142756 Chr.6: 41771550-41771550 on Build GRCh38
rs906565723 Chr.6: 41771540-41771541 on Build GRCh38
rs773994466 Chr.6: 41771283-41771283 on Build GRCh38
rs770500682 Chr.6: 41771427-41771427 on Build GRCh38
rs769639339 Chr.6: 41771273-41771273 on Build GRCh38
rs757431291 Chr.6: 41771300-41771300 on Build GRCh38
rs45465093 Chr.6: 41771422-41771422 on Build GRCh38
rs759815229 Chr.6: 41771537-41771537 on Build GRCh38
rs758372745 Chr.6: 41771555-41771555 on Build GRCh38
rs756922101 Chr.6: 41771491-41771491 on Build GRCh38
rs749304627 Chr.6: 41771331-41771331 on Build GRCh38
rs45447496 Chr.6: 41771423-41771423 on Build GRCh38
rs767637639 Chr.6: 41771520-41771520 on Build GRCh38
rs774960801 Chr.6: 41771432-41771432 on Build GRCh38
rs752741564 Chr.6: 41771481-41771481 on Build GRCh38

View Full Product Details