Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151705735-151706235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF3
Entrez Gene ID
11189
Gene Name
CUGBP, Elav-like family member 3
Gene Aliases
BRUNOL1, CAGH4, ERDA4, ETR-1, TNRC4
UniGene
Hs.26047
Gene Chromosome Location
Chr.1: 151700058-151716821 on Build GRCh38
Transcripts
NM_001172648.3 NM_001172649.3 NM_001291106.1 NM_001291107.1 NM_007185.6 XM_006711139.2 XM_006711141.2 XM_011509111.2 XM_011509112.2 XM_011509114.2 XM_017000145.1 XM_017000146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5843627 c.1243C>T p.R415* Chr.1: 151705849-151705849 on Build GRCh38
1295056 c.1240G>A p.D414N Chr.1: 151705852-151705852 on Build GRCh38
895888 c.1265G>A p.C422Y Chr.1: 151705827-151705827 on Build GRCh38
3801839 c.1152C>G p.I384M Chr.1: 151705940-151705940 on Build GRCh38
3473703 c.1210G>C p.G404R Chr.1: 151705882-151705882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544221742 Chr.1: 151706042-151706042 on Build GRCh38
rs780144439 Chr.1: 151706176-151706176 on Build GRCh38
rs770094170 Chr.1: 151705877-151705877 on Build GRCh38
rs192788109 Chr.1: 151706098-151706098 on Build GRCh38
rs762261971 Chr.1: 151705876-151705876 on Build GRCh38
rs530484991 Chr.1: 151706021-151706021 on Build GRCh38
rs112375967 Chr.1: 151705868-151705868 on Build GRCh38
rs375964012 Chr.1: 151706178-151706178 on Build GRCh38
rs776483797 Chr.1: 151705849-151705849 on Build GRCh38
rs766043872 Chr.1: 151705986-151705989 on Build GRCh38
rs762101984 Chr.1: 151705980-151705983 on Build GRCh38
rs774343466 Chr.1: 151705921-151705921 on Build GRCh38
rs778909084 Chr.1: 151705798-151705798 on Build GRCh38
rs55656977 Chr.1: 151705974-151705974 on Build GRCh38
rs746550479 Chr.1: 151705844-151705844 on Build GRCh38
rs549078975 Chr.1: 151706099-151706099 on Build GRCh38
rs1022370720 Chr.1: 151706000-151706000 on Build GRCh38
rs755150109 Chr.1: 151705786-151705786 on Build GRCh38
rs528588465 Chr.1: 151706134-151706134 on Build GRCh38
rs751658069 Chr.1: 151705783-151705783 on Build GRCh38
rs779383582 Chr.1: 151705808-151705808 on Build GRCh38
rs763055886 Chr.1: 151705772-151705772 on Build GRCh38
rs763316916 Chr.1: 151705889-151705889 on Build GRCh38
rs888381440 Chr.1: 151705791-151705791 on Build GRCh38
rs762980314 Chr.1: 151705790-151705790 on Build GRCh38
rs752987071 Chr.1: 151705975-151705975 on Build GRCh38
rs771591135 Chr.1: 151705807-151705807 on Build GRCh38
rs759629716 Chr.1: 151705927-151705927 on Build GRCh38
rs143103619 Chr.1: 151706108-151706108 on Build GRCh38
rs757048866 Chr.1: 151705998-151705998 on Build GRCh38
rs374382164 Chr.1: 151706133-151706133 on Build GRCh38
rs745835393 Chr.1: 151705803-151705803 on Build GRCh38
rs546375948 Chr.1: 151706102-151706102 on Build GRCh38
rs138584652 Chr.1: 151706171-151706171 on Build GRCh38
rs781393952 Chr.1: 151705788-151705788 on Build GRCh38
rs900767063 Chr.1: 151706035-151706035 on Build GRCh38
rs149504358 Chr.1: 151705763-151705763 on Build GRCh38
rs374078133 Chr.1: 151705797-151705797 on Build GRCh38
rs957399495 Chr.1: 151706055-151706055 on Build GRCh38
rs766417038 Chr.1: 151705777-151705777 on Build GRCh38
rs750256689 Chr.1: 151706014-151706014 on Build GRCh38
rs1033409464 Chr.1: 151706050-151706050 on Build GRCh38
rs992261120 Chr.1: 151706155-151706155 on Build GRCh38
rs768357884 Chr.1: 151705848-151705848 on Build GRCh38
rs373583377 Chr.1: 151706210-151706210 on Build GRCh38
rs367603038 Chr.1: 151705878-151705878 on Build GRCh38
rs780959641 Chr.1: 151706188-151706188 on Build GRCh38
rs959539756 Chr.1: 151706118-151706118 on Build GRCh38
rs116471360 Chr.1: 151706091-151706091 on Build GRCh38
rs1308151 Chr.1: 151706165-151706165 on Build GRCh38
rs370547349 Chr.1: 151706179-151706179 on Build GRCh38
rs754602332 Chr.1: 151706187-151706187 on Build GRCh38
rs764058072 Chr.1: 151706017-151706017 on Build GRCh38
rs764992893 Chr.1: 151706003-151706003 on Build GRCh38
rs766937336 Chr.1: 151705909-151705909 on Build GRCh38
rs1010319928 Chr.1: 151706073-151706073 on Build GRCh38

View Full Product Details