Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42822609-42823115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLTSCR1L
Entrez Gene ID
23506
Gene Name
GLTSCR1 like
Gene Aliases
KIAA0240
UniGene
Hs.658033
Gene Chromosome Location
Chr.6: 42746958-42868560 on Build GRCh38
Transcripts
NM_001318819.1 NM_015349.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4849550 c.122C>T p.S41F Chr.6: 42822966-42822966 on Build GRCh38
5357758 c.43G>T p.D15Y Chr.6: 42822797-42822797 on Build GRCh38
1079358 c.100G>T p.D34Y Chr.6: 42822944-42822944 on Build GRCh38
3079305 c.148G>A p.A50T Chr.6: 42822992-42822992 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs145740980 Chr.6: 42822746-42822746 on Build GRCh38
rs759789865 Chr.6: 42822926-42822926 on Build GRCh38
rs1002040361 Chr.6: 42822991-42822991 on Build GRCh38
rs546479168 Chr.6: 42822914-42822914 on Build GRCh38
rs780210952 Chr.6: 42822999-42822999 on Build GRCh38
rs1024393009 Chr.6: 42822644-42822644 on Build GRCh38
rs909898508 Chr.6: 42822708-42822708 on Build GRCh38
rs982790959 Chr.6: 42822663-42822663 on Build GRCh38
rs746856367 Chr.6: 42822749-42822749 on Build GRCh38
rs1054970180 Chr.6: 42823046-42823046 on Build GRCh38
rs371991695 Chr.6: 42822895-42822895 on Build GRCh38
rs752488630 Chr.6: 42822872-42822872 on Build GRCh38
rs759530683 Chr.6: 42822758-42822758 on Build GRCh38
rs987712317 Chr.6: 42823078-42823078 on Build GRCh38
rs762519199 Chr.6: 42822771-42822771 on Build GRCh38
rs1031355021 Chr.6: 42822814-42822814 on Build GRCh38
rs754820160 Chr.6: 42822775-42822776 on Build GRCh38
rs778599858 Chr.6: 42822774-42822775 on Build GRCh38
rs778035209 Chr.6: 42822844-42822844 on Build GRCh38
rs745721447 Chr.6: 42822992-42822992 on Build GRCh38
rs912270232 Chr.6: 42823081-42823081 on Build GRCh38
rs758451407 Chr.6: 42822843-42822843 on Build GRCh38
rs755532791 Chr.6: 42822821-42822821 on Build GRCh38
rs751246348 Chr.6: 42822781-42822781 on Build GRCh38
rs748268713 Chr.6: 42822841-42822841 on Build GRCh38
rs903540754 Chr.6: 42822908-42822908 on Build GRCh38
rs758834669 Chr.6: 42822949-42822949 on Build GRCh38
rs766775775 Chr.6: 42822808-42822808 on Build GRCh38
rs762606829 Chr.6: 42822882-42822882 on Build GRCh38
rs780035075 Chr.6: 42822760-42822762 on Build GRCh38
rs896447221 Chr.6: 42823074-42823074 on Build GRCh38
rs749244962 Chr.6: 42823002-42823002 on Build GRCh38
rs775093316 Chr.6: 42822770-42822770 on Build GRCh38
rs772189428 Chr.6: 42823047-42823050 on Build GRCh38
rs368033120 Chr.6: 42822857-42822857 on Build GRCh38
rs202199143 Chr.6: 42822860-42822860 on Build GRCh38
rs757530367 Chr.6: 42822958-42822958 on Build GRCh38
rs765035790 Chr.6: 42822764-42822764 on Build GRCh38
rs748362046 Chr.6: 42822974-42822979 on Build GRCh38
rs751590419 Chr.6: 42822957-42822957 on Build GRCh38
rs751427347 Chr.6: 42822644-42822644 on Build GRCh38
rs756029201 Chr.6: 42822759-42822759 on Build GRCh38
rs775544520 Chr.6: 42822874-42822874 on Build GRCh38
rs972233016 Chr.6: 42822939-42822939 on Build GRCh38
rs777842176 Chr.6: 42822955-42822955 on Build GRCh38
rs765824109 Chr.6: 42822929-42822929 on Build GRCh38
rs1013548286 Chr.6: 42822833-42822833 on Build GRCh38
rs769460548 Chr.6: 42822996-42822996 on Build GRCh38
rs892346645 Chr.6: 42822828-42822828 on Build GRCh38
rs375868433 Chr.6: 42822975-42822975 on Build GRCh38
rs753191482 Chr.6: 42822946-42822946 on Build GRCh38
rs776221123 Chr.6: 42822751-42822751 on Build GRCh38
rs745606277 Chr.6: 42822871-42822871 on Build GRCh38
rs767032045 Chr.6: 42822912-42822912 on Build GRCh38
rs200455460 Chr.6: 42822775-42822775 on Build GRCh38
rs749055332 Chr.6: 42822775-42822775 on Build GRCh38
rs951582768 Chr.6: 42822656-42822656 on Build GRCh38
rs189069875 Chr.6: 42822856-42822856 on Build GRCh38
rs770753347 Chr.6: 42822750-42822750 on Build GRCh38
rs768660228 Chr.6: 42823049-42823049 on Build GRCh38
rs555900376 Chr.6: 42823083-42823083 on Build GRCh38
rs756616215 Chr.6: 42822783-42822783 on Build GRCh38
rs768280367 Chr.6: 42822885-42822885 on Build GRCh38
rs773933726 Chr.6: 42822902-42822902 on Build GRCh38
rs760280458 Chr.6: 42822827-42822827 on Build GRCh38
rs913637432 Chr.6: 42822868-42822868 on Build GRCh38
rs761545215 Chr.6: 42822911-42822911 on Build GRCh38
rs542579613 Chr.6: 42822997-42822997 on Build GRCh38
rs945141680 Chr.6: 42822870-42822870 on Build GRCh38
rs754097111 Chr.6: 42822809-42822809 on Build GRCh38

View Full Product Details