Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42828882-42829383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLTSCR1L
Entrez Gene ID
23506
Gene Name
GLTSCR1 like
Gene Aliases
KIAA0240
UniGene
Hs.658033
Gene Chromosome Location
Chr.6: 42746958-42868560 on Build GRCh38
Transcripts
NM_001318819.1 NM_015349.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5585574 c.928C>T p.P310S Chr.6: 42829261-42829261 on Build GRCh38
6173955 c.927C>T p.V309V Chr.6: 42829260-42829260 on Build GRCh38
1079360 c.998T>G p.I333S Chr.6: 42829331-42829331 on Build GRCh38
4695191 c.750T>C p.S250S Chr.6: 42829083-42829083 on Build GRCh38
3394205 c.854C>G p.T285R Chr.6: 42829187-42829187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867042538 Chr.6: 42829042-42829042 on Build GRCh38
rs557465503 Chr.6: 42828979-42828979 on Build GRCh38
rs142958072 Chr.6: 42829056-42829056 on Build GRCh38
rs141108186 Chr.6: 42829308-42829308 on Build GRCh38
rs749979308 Chr.6: 42829185-42829185 on Build GRCh38
rs780691468 Chr.6: 42829033-42829033 on Build GRCh38
rs903877718 Chr.6: 42829173-42829173 on Build GRCh38
rs776786692 Chr.6: 42828911-42828911 on Build GRCh38
rs1000799350 Chr.6: 42829184-42829184 on Build GRCh38
rs930119414 Chr.6: 42828933-42828933 on Build GRCh38
rs774165397 Chr.6: 42829265-42829265 on Build GRCh38
rs551087292 Chr.6: 42829041-42829041 on Build GRCh38
rs184041202 Chr.6: 42829078-42829078 on Build GRCh38
rs759857620 Chr.6: 42828928-42828928 on Build GRCh38
rs760174644 Chr.6: 42829314-42829314 on Build GRCh38
rs781758143 Chr.6: 42829227-42829227 on Build GRCh38
rs748514505 Chr.6: 42829057-42829057 on Build GRCh38
rs138455961 Chr.6: 42829015-42829015 on Build GRCh38
rs201246496 Chr.6: 42829121-42829121 on Build GRCh38
rs775364473 Chr.6: 42829241-42829241 on Build GRCh38
rs760896041 Chr.6: 42828907-42828907 on Build GRCh38
rs761565543 Chr.6: 42829284-42829284 on Build GRCh38
rs890953572 Chr.6: 42829161-42829161 on Build GRCh38
rs759656651 Chr.6: 42829105-42829105 on Build GRCh38
rs562256098 Chr.6: 42829209-42829209 on Build GRCh38
rs754524134 Chr.6: 42829213-42829213 on Build GRCh38
rs778298538 Chr.6: 42829214-42829214 on Build GRCh38
rs749889419 Chr.6: 42829043-42829043 on Build GRCh38
rs761759453 Chr.6: 42829170-42829170 on Build GRCh38
rs747335476 Chr.6: 42829217-42829217 on Build GRCh38
rs778211585 Chr.6: 42829086-42829086 on Build GRCh38
rs764949266 Chr.6: 42829355-42829355 on Build GRCh38
rs757876809 Chr.6: 42829225-42829225 on Build GRCh38
rs753231676 Chr.6: 42829342-42829342 on Build GRCh38
rs768710395 Chr.6: 42829264-42829264 on Build GRCh38
rs1009336223 Chr.6: 42829114-42829114 on Build GRCh38
rs769912983 Chr.6: 42829237-42829237 on Build GRCh38
rs769998620 Chr.6: 42829171-42829171 on Build GRCh38
rs368277645 Chr.6: 42828932-42828932 on Build GRCh38
rs747145487 Chr.6: 42829218-42829220 on Build GRCh38
rs113235956 Chr.6: 42828941-42828941 on Build GRCh38
rs763951148 Chr.6: 42829152-42829152 on Build GRCh38
rs752474908 Chr.6: 42829007-42829007 on Build GRCh38
rs370636563 Chr.6: 42829189-42829189 on Build GRCh38
rs377630919 Chr.6: 42829128-42829128 on Build GRCh38
rs969705588 Chr.6: 42828952-42828952 on Build GRCh38
rs1050979811 Chr.6: 42829153-42829153 on Build GRCh38
rs745989826 Chr.6: 42829229-42829229 on Build GRCh38
rs904585656 Chr.6: 42829133-42829133 on Build GRCh38
rs773120901 Chr.6: 42829313-42829313 on Build GRCh38
rs763991181 Chr.6: 42829023-42829023 on Build GRCh38
rs941126959 Chr.6: 42829006-42829006 on Build GRCh38
rs992299949 Chr.6: 42828924-42828924 on Build GRCh38
rs200603056 Chr.6: 42829168-42829168 on Build GRCh38
rs757116152 Chr.6: 42829028-42829028 on Build GRCh38
rs374706410 Chr.6: 42829204-42829204 on Build GRCh38
rs542204774 Chr.6: 42829196-42829196 on Build GRCh38
rs940975885 Chr.6: 42829089-42829089 on Build GRCh38
rs909615359 Chr.6: 42828945-42828945 on Build GRCh38
rs755757552 Chr.6: 42829053-42829053 on Build GRCh38
rs200329698 Chr.6: 42828909-42828909 on Build GRCh38
rs187390779 Chr.6: 42829357-42829357 on Build GRCh38
rs749537034 Chr.6: 42829254-42829254 on Build GRCh38
rs776880847 Chr.6: 42829104-42829104 on Build GRCh38
rs747523790 Chr.6: 42829096-42829096 on Build GRCh38
rs553496430 Chr.6: 42829100-42829100 on Build GRCh38
rs769958866 Chr.6: 42829109-42829109 on Build GRCh38
rs751490097 Chr.6: 42829024-42829024 on Build GRCh38
rs765922473 Chr.6: 42829316-42829316 on Build GRCh38
rs754684568 Chr.6: 42829345-42829345 on Build GRCh38

View Full Product Details