Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42829684-42830185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLTSCR1L
Entrez Gene ID
23506
Gene Name
GLTSCR1 like
Gene Aliases
KIAA0240
UniGene
Hs.658033
Gene Chromosome Location
Chr.6: 42746958-42868560 on Build GRCh38
Transcripts
NM_001318819.1 NM_015349.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6671879 c.1460C>T p.A487V Chr.6: 42829793-42829793 on Build GRCh38
3430681 c.1830C>A p.F610L Chr.6: 42830163-42830163 on Build GRCh38
166626 c.1616G>A p.R539Q Chr.6: 42829949-42829949 on Build GRCh38
5559764 c.1712T>C p.L571P Chr.6: 42830045-42830045 on Build GRCh38
6671877 c.1772C>A p.S591Y Chr.6: 42830105-42830105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199601305 Chr.6: 42829746-42829746 on Build GRCh38
rs773846502 Chr.6: 42829889-42829889 on Build GRCh38
rs766658727 Chr.6: 42829802-42829802 on Build GRCh38
rs774334662 Chr.6: 42830071-42830071 on Build GRCh38
rs371702663 Chr.6: 42829923-42829923 on Build GRCh38
rs771711586 Chr.6: 42829909-42829909 on Build GRCh38
rs751607621 Chr.6: 42829946-42829946 on Build GRCh38
rs149171648 Chr.6: 42830114-42830114 on Build GRCh38
rs925110181 Chr.6: 42829949-42829949 on Build GRCh38
rs767908263 Chr.6: 42830085-42830085 on Build GRCh38
rs899315164 Chr.6: 42830126-42830126 on Build GRCh38
rs144159491 Chr.6: 42829773-42829773 on Build GRCh38
rs753621216 Chr.6: 42830094-42830094 on Build GRCh38
rs777000265 Chr.6: 42829813-42829813 on Build GRCh38
rs540419961 Chr.6: 42829954-42829954 on Build GRCh38
rs1046476694 Chr.6: 42830032-42830032 on Build GRCh38
rs199630219 Chr.6: 42829950-42829950 on Build GRCh38
rs762519324 Chr.6: 42829768-42829768 on Build GRCh38
rs887110854 Chr.6: 42830109-42830109 on Build GRCh38
rs773862714 Chr.6: 42829796-42829796 on Build GRCh38
rs766290946 Chr.6: 42830090-42830090 on Build GRCh38
rs760216450 Chr.6: 42829820-42829820 on Build GRCh38
rs775970222 Chr.6: 42829928-42829928 on Build GRCh38
rs768405641 Chr.6: 42829771-42829771 on Build GRCh38
rs368647251 Chr.6: 42829920-42829920 on Build GRCh38
rs376148515 Chr.6: 42829867-42829867 on Build GRCh38
rs769165870 Chr.6: 42830061-42830061 on Build GRCh38
rs143301030 Chr.6: 42830145-42830145 on Build GRCh38
rs764091195 Chr.6: 42829940-42829940 on Build GRCh38
rs200974412 Chr.6: 42829970-42829970 on Build GRCh38
rs1037911450 Chr.6: 42830117-42830117 on Build GRCh38
rs761169254 Chr.6: 42829797-42829797 on Build GRCh38
rs750671253 Chr.6: 42830086-42830086 on Build GRCh38
rs193921035 Chr.6: 42830111-42830111 on Build GRCh38
rs1025334504 Chr.6: 42829900-42829900 on Build GRCh38
rs780969103 Chr.6: 42829725-42829725 on Build GRCh38
rs775170100 Chr.6: 42829767-42829767 on Build GRCh38
rs755961336 Chr.6: 42829974-42829974 on Build GRCh38
rs760141598 Chr.6: 42830161-42830163 on Build GRCh38
rs767746813 Chr.6: 42829960-42829960 on Build GRCh38
rs765696304 Chr.6: 42829819-42829819 on Build GRCh38
rs770145250 Chr.6: 42830033-42830033 on Build GRCh38
rs975181800 Chr.6: 42829924-42829924 on Build GRCh38
rs757918730 Chr.6: 42830134-42830134 on Build GRCh38
rs141434173 Chr.6: 42829852-42829852 on Build GRCh38
rs560532159 Chr.6: 42830013-42830013 on Build GRCh38
rs771664411 Chr.6: 42830019-42830019 on Build GRCh38
rs754908273 Chr.6: 42830108-42830108 on Build GRCh38
rs780209242 Chr.6: 42829977-42829977 on Build GRCh38
rs752817411 Chr.6: 42829822-42829822 on Build GRCh38
rs746295988 Chr.6: 42830031-42830031 on Build GRCh38
rs371884482 Chr.6: 42829853-42829853 on Build GRCh38
rs951580875 Chr.6: 42829757-42829757 on Build GRCh38
rs780115744 Chr.6: 42829880-42829880 on Build GRCh38
rs777194999 Chr.6: 42830135-42830135 on Build GRCh38
rs150278376 Chr.6: 42829744-42829744 on Build GRCh38
rs777324733 Chr.6: 42830034-42830036 on Build GRCh38
rs376512564 Chr.6: 42829988-42829988 on Build GRCh38
rs147851446 Chr.6: 42829832-42829832 on Build GRCh38
rs775769988 Chr.6: 42830036-42830036 on Build GRCh38
rs576027686 Chr.6: 42829897-42829897 on Build GRCh38
rs749242934 Chr.6: 42829982-42829982 on Build GRCh38
rs763388560 Chr.6: 42830044-42830044 on Build GRCh38
rs369217530 Chr.6: 42829878-42829878 on Build GRCh38
rs544780541 Chr.6: 42829922-42829922 on Build GRCh38
rs912275797 Chr.6: 42829915-42829915 on Build GRCh38
rs768703483 Chr.6: 42829745-42829745 on Build GRCh38
rs778311910 Chr.6: 42829997-42829997 on Build GRCh38
rs111780285 Chr.6: 42830023-42830023 on Build GRCh38
rs539876077 Chr.6: 42829885-42829885 on Build GRCh38
rs951106816 Chr.6: 42829906-42829906 on Build GRCh38
rs983042403 Chr.6: 42829766-42829766 on Build GRCh38
rs745617685 Chr.6: 42829876-42829876 on Build GRCh38
rs533652906 Chr.6: 42829715-42829715 on Build GRCh38
rs780872222 Chr.6: 42829870-42829870 on Build GRCh38
rs145266207 Chr.6: 42829884-42829884 on Build GRCh38
rs751730855 Chr.6: 42829857-42829857 on Build GRCh38
rs752201203 Chr.6: 42830131-42830131 on Build GRCh38
rs772831887 Chr.6: 42830028-42830028 on Build GRCh38
rs760062737 Chr.6: 42829816-42829816 on Build GRCh38
rs1013700019 Chr.6: 42829716-42829716 on Build GRCh38
rs892479395 Chr.6: 42829985-42829985 on Build GRCh38
rs747773730 Chr.6: 42829908-42829908 on Build GRCh38
rs777832712 Chr.6: 42829708-42829708 on Build GRCh38
rs746876231 Chr.6: 42829711-42829711 on Build GRCh38
rs144992246 Chr.6: 42830089-42830089 on Build GRCh38
rs761929434 Chr.6: 42830073-42830073 on Build GRCh38

View Full Product Details