Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43185875-43186380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL9
Entrez Gene ID
23113
Gene Name
cullin 9
Gene Aliases
H7AP1, PARC
UniGene
Hs.485434
Gene Chromosome Location
Chr.6: 43182170-43224587 on Build GRCh38
Transcripts
NM_015089.3 XM_011514422.1 XM_011514423.2 XM_011514424.1 XM_011514425.1 XM_011514426.1 XM_011514427.1 XM_011514428.1 XM_011514430.1 XM_017010589.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6250109 c.1004G>T p.R335L Chr.6: 43186208-43186208 on Build GRCh38
1329674 c.1082G>A p.W361* Chr.6: 43186286-43186286 on Build GRCh38
6226195 c.1168C>T p.R390W Chr.6: 43186372-43186372 on Build GRCh38
1634810 c.829C>G p.P277A Chr.6: 43186033-43186033 on Build GRCh38
4511158 c.859C>T p.P287S Chr.6: 43186063-43186063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200443373 Chr.6: 43185899-43185899 on Build GRCh38
rs975897200 Chr.6: 43186298-43186298 on Build GRCh38
rs953435212 Chr.6: 43186300-43186300 on Build GRCh38
rs754748621 Chr.6: 43186135-43186135 on Build GRCh38
rs764019299 Chr.6: 43186336-43186336 on Build GRCh38
rs770442910 Chr.6: 43185988-43185988 on Build GRCh38
rs147830394 Chr.6: 43186076-43186076 on Build GRCh38
rs779785417 Chr.6: 43185931-43185931 on Build GRCh38
rs767675598 Chr.6: 43186026-43186026 on Build GRCh38
rs772380914 Chr.6: 43186299-43186299 on Build GRCh38
rs1042129393 Chr.6: 43186356-43186356 on Build GRCh38
rs774729669 Chr.6: 43186198-43186198 on Build GRCh38
rs779482945 Chr.6: 43186088-43186088 on Build GRCh38
rs756795985 Chr.6: 43186272-43186272 on Build GRCh38
rs903311397 Chr.6: 43185914-43185914 on Build GRCh38
rs771674728 Chr.6: 43185960-43185960 on Build GRCh38
rs1029937050 Chr.6: 43186170-43186170 on Build GRCh38
rs890751589 Chr.6: 43186008-43186008 on Build GRCh38
rs151050685 Chr.6: 43186134-43186134 on Build GRCh38
rs376344620 Chr.6: 43185908-43185908 on Build GRCh38
rs775345810 Chr.6: 43186328-43186328 on Build GRCh38
rs765095560 Chr.6: 43186258-43186258 on Build GRCh38
rs778635286 Chr.6: 43185955-43185955 on Build GRCh38
rs779981942 Chr.6: 43186035-43186035 on Build GRCh38
rs539870137 Chr.6: 43186295-43186295 on Build GRCh38
rs769112831 Chr.6: 43186091-43186091 on Build GRCh38
rs755456316 Chr.6: 43186294-43186294 on Build GRCh38
rs79828871 Chr.6: 43186326-43186326 on Build GRCh38
rs537623008 Chr.6: 43186205-43186205 on Build GRCh38
rs771649929 Chr.6: 43186058-43186058 on Build GRCh38
rs373891771 Chr.6: 43186004-43186004 on Build GRCh38
rs561502761 Chr.6: 43185898-43185898 on Build GRCh38
rs747887596 Chr.6: 43186053-43186053 on Build GRCh38
rs753566522 Chr.6: 43186041-43186041 on Build GRCh38
rs1012030423 Chr.6: 43186175-43186175 on Build GRCh38
rs1037793089 Chr.6: 43185915-43185915 on Build GRCh38
rs767292472 Chr.6: 43186349-43186349 on Build GRCh38
rs752452800 Chr.6: 43186150-43186150 on Build GRCh38
rs975928307 Chr.6: 43186073-43186073 on Build GRCh38
rs776986537 Chr.6: 43185975-43185975 on Build GRCh38
rs61743561 Chr.6: 43186049-43186049 on Build GRCh38
rs750500338 Chr.6: 43186029-43186029 on Build GRCh38
rs941849 Chr.6: 43186353-43186353 on Build GRCh38
rs780978576 Chr.6: 43185925-43185925 on Build GRCh38
rs777284387 Chr.6: 43186066-43186066 on Build GRCh38
rs750027800 Chr.6: 43185934-43185934 on Build GRCh38
rs766498881 Chr.6: 43186213-43186213 on Build GRCh38
rs764405319 Chr.6: 43185992-43185992 on Build GRCh38
rs779645786 Chr.6: 43186271-43186271 on Build GRCh38
rs760749913 Chr.6: 43186117-43186117 on Build GRCh38
rs145136960 Chr.6: 43186036-43186036 on Build GRCh38
rs150188923 Chr.6: 43186279-43186279 on Build GRCh38
rs755998452 Chr.6: 43186030-43186030 on Build GRCh38
rs113622744 Chr.6: 43185923-43185923 on Build GRCh38
rs773286904 Chr.6: 43186112-43186112 on Build GRCh38
rs958686935 Chr.6: 43186082-43186082 on Build GRCh38
rs147818029 Chr.6: 43186093-43186093 on Build GRCh38
rs765124435 Chr.6: 43186147-43186147 on Build GRCh38
rs1057252064 Chr.6: 43186151-43186151 on Build GRCh38
rs758006872 Chr.6: 43186153-43186153 on Build GRCh38
rs758192043 Chr.6: 43186270-43186270 on Build GRCh38
rs775935398 Chr.6: 43185989-43185989 on Build GRCh38
rs960460553 Chr.6: 43185968-43185968 on Build GRCh38
rs769074644 Chr.6: 43186186-43186186 on Build GRCh38
rs776523576 Chr.6: 43186227-43186227 on Build GRCh38
rs745393413 Chr.6: 43186290-43186290 on Build GRCh38
rs760076216 Chr.6: 43185979-43185979 on Build GRCh38
rs140962096 Chr.6: 43186176-43186176 on Build GRCh38
rs764421817 Chr.6: 43185912-43185912 on Build GRCh38
rs941193898 Chr.6: 43186168-43186168 on Build GRCh38
rs763412061 Chr.6: 43185991-43185991 on Build GRCh38
rs761507783 Chr.6: 43186348-43186348 on Build GRCh38
rs759465036 Chr.6: 43186249-43186249 on Build GRCh38
rs148687309 Chr.6: 43186048-43186048 on Build GRCh38
rs751430285 Chr.6: 43186339-43186339 on Build GRCh38
rs747610788 Chr.6: 43185957-43185957 on Build GRCh38
rs141303988 Chr.6: 43186098-43186098 on Build GRCh38
rs1002971642 Chr.6: 43186131-43186131 on Build GRCh38
rs763215576 Chr.6: 43185909-43185909 on Build GRCh38
rs553582467 Chr.6: 43186083-43186083 on Build GRCh38
rs568642278 Chr.6: 43186199-43186199 on Build GRCh38
rs373211287 Chr.6: 43186319-43186319 on Build GRCh38
rs181783484 Chr.6: 43186166-43186166 on Build GRCh38
rs368973786 Chr.6: 43186269-43186269 on Build GRCh38
rs147065096 Chr.6: 43186133-43186133 on Build GRCh38
rs201241003 Chr.6: 43186243-43186243 on Build GRCh38
rs762056197 Chr.6: 43186096-43186096 on Build GRCh38
rs749851704 Chr.6: 43186177-43186177 on Build GRCh38
rs760733396 Chr.6: 43186208-43186208 on Build GRCh38
rs749105620 Chr.6: 43185932-43185932 on Build GRCh38
rs543872238 Chr.6: 43185916-43185916 on Build GRCh38
rs750422594 Chr.6: 43185929-43185929 on Build GRCh38
rs201425802 Chr.6: 43185937-43185937 on Build GRCh38
rs200409806 Chr.6: 43186087-43186087 on Build GRCh38
rs773493403 Chr.6: 43186207-43186207 on Build GRCh38
rs777291584 Chr.6: 43186165-43186165 on Build GRCh38
rs776630183 Chr.6: 43186318-43186318 on Build GRCh38
rs761776844 Chr.6: 43186005-43186005 on Build GRCh38
rs771179035 Chr.6: 43186304-43186304 on Build GRCh38

View Full Product Details