Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44000885-44001398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf223
Entrez Gene ID
221416
Gene Name
chromosome 6 open reading frame 223
Gene Aliases
-
UniGene
Hs.531926
Gene Chromosome Location
Chr.6: 43999515-44007615 on Build GRCh38
Transcripts
NM_001171992.2 NM_001324369.1 NM_153246.5 XM_017010425.1 NR_125864.1 NR_125865.1

Gene Symbol
LOC101929705
Entrez Gene ID
101929705
Gene Name
uncharacterized LOC101929705
Gene Aliases
-
UniGene
Hs.670378
Gene Chromosome Location
Chr.6: 43995723-44074652 on Build GRCh38
Transcripts
NM_001171992.2 NM_001324369.1 NM_153246.5 XM_017010425.1 NR_125864.1 NR_125865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3627763 c.200C>T p.P67L Chr.6: 44001092-44001092 on Build GRCh38
6482844 c.137C>G p.A46G Chr.6: 44001029-44001029 on Build GRCh38
3627762 c.157C>T p.L53F Chr.6: 44001092-44001092 on Build GRCh38
6616466 c.235G>T p.E79* Chr.6: 44001127-44001127 on Build GRCh38
4667902 c.89C>A p.P30H Chr.6: 44001024-44001024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542757135 Chr.6: 44000970-44000970 on Build GRCh38
rs567338643 Chr.6: 44001213-44001213 on Build GRCh38
rs958078678 Chr.6: 44001205-44001205 on Build GRCh38
rs772777084 Chr.6: 44001173-44001173 on Build GRCh38
rs534691997 Chr.6: 44001214-44001214 on Build GRCh38
rs762797186 Chr.6: 44000958-44000958 on Build GRCh38
rs760404642 Chr.6: 44001175-44001175 on Build GRCh38
rs563883191 Chr.6: 44001303-44001303 on Build GRCh38
rs750159661 Chr.6: 44001178-44001180 on Build GRCh38
rs138433716 Chr.6: 44001106-44001106 on Build GRCh38
rs749306120 Chr.6: 44001166-44001166 on Build GRCh38
rs753361308 Chr.6: 44001138-44001138 on Build GRCh38
rs1016110117 Chr.6: 44001362-44001362 on Build GRCh38
rs958046306 Chr.6: 44000916-44000916 on Build GRCh38
rs991686936 Chr.6: 44000926-44000926 on Build GRCh38
rs753231019 Chr.6: 44001022-44001022 on Build GRCh38
rs757638543 Chr.6: 44001153-44001153 on Build GRCh38
rs569562348 Chr.6: 44001133-44001133 on Build GRCh38
rs199832309 Chr.6: 44000945-44000945 on Build GRCh38
rs575570128 Chr.6: 44001331-44001331 on Build GRCh38
rs557172382 Chr.6: 44001306-44001306 on Build GRCh38
rs557474577 Chr.6: 44001363-44001363 on Build GRCh38
rs761723533 Chr.6: 44001159-44001170 on Build GRCh38
rs1023185907 Chr.6: 44001290-44001290 on Build GRCh38
rs772548835 Chr.6: 44000935-44000935 on Build GRCh38
rs200667175 Chr.6: 44001120-44001120 on Build GRCh38
rs200726704 Chr.6: 44000949-44000949 on Build GRCh38
rs768584777 Chr.6: 44001167-44001167 on Build GRCh38
rs2295333 Chr.6: 44001077-44001077 on Build GRCh38
rs371430022 Chr.6: 44001101-44001101 on Build GRCh38
rs778316720 Chr.6: 44001025-44001025 on Build GRCh38
rs767285933 Chr.6: 44001175-44001180 on Build GRCh38
rs928556015 Chr.6: 44001320-44001320 on Build GRCh38
rs377672754 Chr.6: 44000975-44000975 on Build GRCh38
rs769906631 Chr.6: 44001085-44001085 on Build GRCh38
rs769624686 Chr.6: 44000951-44000951 on Build GRCh38
rs370963797 Chr.6: 44001248-44001248 on Build GRCh38
rs766431366 Chr.6: 44000910-44000910 on Build GRCh38
rs749906099 Chr.6: 44001001-44001001 on Build GRCh38
rs981799088 Chr.6: 44001307-44001307 on Build GRCh38
rs759054673 Chr.6: 44001189-44001189 on Build GRCh38
rs900030603 Chr.6: 44001136-44001136 on Build GRCh38
rs755642676 Chr.6: 44001002-44001002 on Build GRCh38
rs201977991 Chr.6: 44001127-44001127 on Build GRCh38
rs763856535 Chr.6: 44000967-44000967 on Build GRCh38
rs747206661 Chr.6: 44000938-44000938 on Build GRCh38
rs767973975 Chr.6: 44001327-44001327 on Build GRCh38
rs1055980306 Chr.6: 44001042-44001042 on Build GRCh38
rs553332639 Chr.6: 44001361-44001361 on Build GRCh38
rs775457209 Chr.6: 44001088-44001088 on Build GRCh38
rs201283463 Chr.6: 44000939-44000939 on Build GRCh38
rs775402258 Chr.6: 44000953-44000953 on Build GRCh38
rs190852477 Chr.6: 44001259-44001259 on Build GRCh38
rs758871703 Chr.6: 44001024-44001024 on Build GRCh38
rs930994007 Chr.6: 44001100-44001100 on Build GRCh38
rs1001516579 Chr.6: 44001212-44001212 on Build GRCh38
rs200098091 Chr.6: 44001172-44001172 on Build GRCh38
rs370221812 Chr.6: 44001161-44001161 on Build GRCh38
rs767139877 Chr.6: 44000998-44000998 on Build GRCh38
rs774267623 Chr.6: 44001168-44001168 on Build GRCh38
rs144702444 Chr.6: 44001105-44001105 on Build GRCh38
rs749252901 Chr.6: 44001091-44001091 on Build GRCh38
rs528676415 Chr.6: 44001043-44001043 on Build GRCh38
rs781327304 Chr.6: 44001157-44001157 on Build GRCh38
rs761171208 Chr.6: 44001122-44001125 on Build GRCh38
rs561404229 Chr.6: 44000985-44000985 on Build GRCh38
rs748336837 Chr.6: 44000937-44000937 on Build GRCh38
rs148840887 Chr.6: 44001144-44001144 on Build GRCh38
rs551245633 Chr.6: 44001111-44001111 on Build GRCh38
rs776297679 Chr.6: 44001182-44001182 on Build GRCh38
rs779342524 Chr.6: 44001021-44001021 on Build GRCh38
rs147859295 Chr.6: 44001066-44001066 on Build GRCh38
rs768074794 Chr.6: 44000973-44000973 on Build GRCh38
rs919685892 Chr.6: 44001373-44001373 on Build GRCh38
rs746082441 Chr.6: 44001159-44001159 on Build GRCh38
rs762487171 Chr.6: 44000962-44000963 on Build GRCh38
rs779406779 Chr.6: 44000931-44000931 on Build GRCh38
rs766083375 Chr.6: 44001176-44001176 on Build GRCh38
rs771075507 Chr.6: 44001052-44001052 on Build GRCh38
rs545302524 Chr.6: 44000933-44000933 on Build GRCh38
rs201854114 Chr.6: 44001148-44001148 on Build GRCh38
rs747946361 Chr.6: 44001169-44001169 on Build GRCh38

View Full Product Details